Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, Fittings, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
YO-07129-20
Totally enclosed, fan-cooled (TEFC) motor
Click to enlarge
More Product Details
Specifications
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-07129-20
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Motors with 56C Frame
Share
|
56C Frame AC Motor, 1725 rpm, 1/2 hp, totally enclosed fan-cooled (TEF (CL3504)
YO-07129-20
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Specifications
Product Type
AC motors for pumps
hp
1/2
Speed range
1725 rpm max
Power
115/230 VAC
Dimensions
10-3/8"L x 8-1/2"W x 9-1/8"H
Technical Resources
Related technical articles (Roll over for summary)
Technical Expertise: How Cole-Parmer Application Specialists Respond to Customers' Needs
Reducing Pulsation
Peristaltic Tubing Pumps in Dispensing Applications
Show All...
Masterflex Tubing: Questions and Answers
Masterflex Tubing Pump Systems FAQs
Tygon® Flexible Plastic Tubing to Tubing Solvent Sealing
Peristaltic Pumps Not Just for Labs Anymore
Tubing Selection Guide For Viscous Fluids
Pumping Abrasives
Pumping Viscous Fluids
Peristaltic Pumps Offer Custom Fluid Solutions
Peristaltic Pumps Offer Economical Multichannel Pumping
Reducing Pulsation in Peristaltic Pumping Systems
Pumping Abrasives
Masterflex Systems: Questions and Answers
High-Performance Masterflex Pump Head
Six Ways to Simplify Fluid Transfer
Masterflex Pump FAQs
Masterflex Tubing Formulations
Show Less...
Related Searches for 56C Frame AC Motor, 1725 rpm, 1/2 hp, totally enclosed fan-cooled (TEF (CL3504)
Masterflex Motors with 56c Frame
1 Rpm Ac Motor
Low Rpm Ac Motor
1 2-Dichloroethane Acs
Ac Motors
Ac Motors Types
Ac Motor Speed Controller
Leeson Nema Type 56c-Face Dc Motors
Ac Motor Controllers
Cole-Parmer Polyvinylchloride (Pvc) 1-2/3" Od Bailers
Related Products for 56C Frame AC Motor, 1725 rpm, 1/2 hp, totally enclosed fan-cooled (TEF (CL3504)
Image
Product Detail
SKU
56C Frame DC Motor, 1800 rpm, 1/2 hp, explosion-proof (XPRF), 90 VDC (EP3640-1436-7-56BC-CU)
Category: Motors with 56C Frame
Description: DC Motors
Brand:
Masterflex
Model Number:
EP3640-1436-7-56BC-CU
YO-02631-06
More Details
AC Variable-Speed Controller, 1/2 hp, 115/230 VAC, 2.4 amp, 120-240VAC
Category: AC Controllers
Description: Single-Phase Input Variable-Speed Controllers
Brand:
Automation Control
Model Number:
ESV371N01SXB
YO-70101-11
More Details
General-purpose Single-phase TEFC/ODP NEMA Type C-face Motor, 1/2 Hp, (56C34F5301/ D311)
Category: AC Single and Three Motors for Positive Displacement Pumps
Description: General-Purpose Totally Enclosed (TEFC)/Open Drip-Proof (ODP), NEMA Type C-Face Four-In-One Motors
Brand:
Marathon
Model Number:
56C34F5301/ D311
YO-71004-20
More Details
General-purpose Single-phase TEFC/ODP NEMA Type C-face Motor, 1/2 Hp, (56C17F5321/ G571)
Category: AC Single and Three Motors for Positive Displacement Pumps
Description: General-Purpose Totally Enclosed (TEFC)/Open Drip-Proof (ODP), NEMA Type C-Face Four-In-One Motors
Brand:
Marathon
Model Number:
56C17F5321/ G571
YO-71004-25
More Details
General-purpose Single-phase TEFC/ODP NEMA Type C-face Motor, 1 1/2 Hp (56B34F5326/ D314)
Category: AC Single and Three Motors for Positive Displacement Pumps
Description: General-Purpose Totally Enclosed (TEFC)/Open Drip-Proof (ODP), NEMA Type C-Face Four-In-One Motors
Brand:
Marathon
Model Number:
56B34F5326/ D314
YO-71004-50
More Details
General-purpose Single-phase TEFC/ODP NEMA Type C-face Motor, 1 1/2 Hp (56B17F5305/ G574)
Category: AC Single and Three Motors for Positive Displacement Pumps
Description: General-Purpose Totally Enclosed (TEFC)/Open Drip-Proof (ODP), NEMA Type C-Face Four-In-One Motors
Brand:
Marathon
Model Number:
56B17F5305/ G574
YO-71004-55
More Details
General Purpose Three-phase TEFC/ODP NEMA Type C-face Motor, 1/2 Hp, 3
Category: AC Single and Three Motors for Positive Displacement Pumps
Description: General-Purpose Totally Enclosed (TEFC)/Open Drip-Proof (ODP), NEMA Type C-Face Four-In-One Motors
Brand:
Marathon
Model Number:
D391
YO-71005-20
More Details
General Purpose Three-phase TEFC/ODP NEMA Type C-face Motor, 1/2 Hp, 1
Category: AC Single and Three Motors for Positive Displacement Pumps
Description: General-Purpose Totally Enclosed (TEFC)/Open Drip-Proof (ODP), NEMA Type C-Face Four-In-One Motors
Brand:
Marathon
Model Number:
G581
YO-71005-25
More Details
General Purpose Three-phase TEFC/ODP NEMA Type C-face Motor, 1-1/2 Hp, (D394)
Category: AC Single and Three Motors for Positive Displacement Pumps
Description: General-Purpose Totally Enclosed (TEFC)/Open Drip-Proof (ODP), NEMA Type C-Face Four-In-One Motors
Brand:
Marathon
Model Number:
D394
YO-71005-50
More Details
General Purpose Three-phase TEFC/ODP NEMA Type C-face Motor, 1-1/2 Hp, (G584)
Category: AC Single and Three Motors for Positive Displacement Pumps
Description: General-Purpose Totally Enclosed (TEFC)/Open Drip-Proof (ODP), NEMA Type C-Face Four-In-One Motors
Brand:
Marathon
Model Number:
G584
YO-71005-55
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
56C Frame AC Motor, 1725 rpm, 1/2 hp, totally enclosed fan-cooled (TEF (CL3504)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: 569001-8 : Contact Probe (YO-20011-19)
|
Next: 56C Frame DC Motor, 1800 rpm, 1/2 hp, explosion-proof (XPRF), 90 VDC (EP3640-1436-7-56BC-CU)
31125329-p-content_2 31125329pcontent2