Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
YO-07379-90
Vacuum Valve
Click to enlarge
More Product Details
Specifications
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-07379-90
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Vacuum Controllers, Gauges, and Systems
Share
|
Vacuum Valve (674,091.0)
YO-07379-90
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Displays offer both an electronic sweeping dial and numerical readout
Specifications
Product Type
Vacuum gauges & controllers
Technical Resources
Related technical articles (Roll over for summary)
Absolute, Gauge, and Differential Pressure Scales
Common Pressure Terms
Pressure Unit Conversion Constants
Show All...
The Hidden Costs of Mercury-Filled Vacuum Gauges
Show Less...
Related Searches for Vacuum Valve (674,091.0)
Bellows Operated Vacuum Valves
Vacuum Valves (Actuated)
Vacuum Valve Emission System
Cole-Parmer Manual Vacuum Valves
One Way Auto Vacuum Valve
Vacuum Valves on Hot Water Heater
Vacuum Valve Can
High Flow Vacuum Valves
Vacuum Valve Parts & Accessories
Vacuum Valves Types
Related Products for Vacuum Valve (674,091.0)
Image
Product Detail
SKU
Solenoid vacuum valve; right-angle, NW16, aluminum, 110 VAC
Category: Solenoid Operation Vacuum Isolation Valves
Description: Right-angle valves
Brand:
Cole-Parmer
Model Number:
C41203000
YO-79302-73
More Details
Solenoid vacuum valve; right-angle, NW25, aluminum, 110 VAC
Category: Solenoid Operation Vacuum Isolation Valves
Description: Right-angle valves
Brand:
Cole-Parmer
Model Number:
C41303000
YO-79302-77
More Details
Solenoid vacuum valve; right-angle, NW25, aluminum, 240 VAC
Category: Solenoid Operation Vacuum Isolation Valves
Description: Right-angle valves
Brand:
Cole-Parmer
Model Number:
C41301000
YO-79302-78
More Details
Solenoid vacuum valve; in-line, NW16, aluminum, 110 VAC
Category: Solenoid Operation Vacuum Isolation Valves
Description: In-line valves
Brand:
Cole-Parmer
Model Number:
C41611000
YO-79302-88
More Details
Solenoid vacuum valve; in-line, NW25, aluminum, 110 VAC
Category: Solenoid Operation Vacuum Isolation Valves
Description: In-line valves
Brand:
Cole-Parmer
Model Number:
C41631000
YO-79302-90
More Details
Vacuum needle valve, NW 10 connections
Category: Regulators and Valves
Description: Manual Vacuum Valves
Brand:
Cole-Parmer
Model Number:
C371-02-000
YO-79302-40
More Details
Vacuum isolation valve, NW 25 connections
Category: Regulators and Valves
Description: Manual Vacuum Valves
Brand:
Cole-Parmer
Model Number:
C333-55-000
YO-79302-60
More Details
Solenoid Vacuum Valve Acid Proof (31F04027)
Category: Oven Accessories
Description: Salvis Vacucenter Oven Accessories
Brand:
Salvis
Model Number:
31F04027
YO-33959-26
More Details
Solenoid Vacuum Valve Chemical (31F04073)
Category: Oven Accessories
Description: Salvis Vacucenter Oven Accessories
Brand:
Salvis
Model Number:
31F04073
YO-33959-27
More Details
Vacuum ball valve, NW16
Category: Ball Valves
Description: Vacuum Ball Valves
Brand:
Cole-Parmer
Model Number:
C36000100
YO-79302-63
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Vacuum Valve (674,091.0)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Vacuum Switch; 1.6 to 27.0 " Hg decreasing ranges, 2.7 to 28.2 " Hg in
|
Next: Vacuum-Ready Random Orbital Sander Kit, Air powered, 6" pad size, 12,0
31125329-p-content_2 31125329pcontent2