Durac Oven Thermometer, 0/200°C, 405mm Length, 125mm Immersion

YO-08006-57
Loading price...

Usually ships in 14 days or sooner.
Qty

  • Replace mercury thermometers for a specific application with the same accuracy without the any hazards
  • Non-mercury liquid
  • Swell
These thermometers are designed for use in specific applications. These Durac thermometers feature markings that are permanently printed on the glass, individually serialized, and red spirit liquid that has a dark scale contrasts against a white background. Some thermometers are PTFE coated for added safety which may cause a slight change in accuracy up to 1 division.

Oven thermometers - designed for high temperature applications and feature a round glass swell to hold the thermometer in place
Product Type Glass Thermometer
Min temperature (º C) 0
Accuracy ±1°C; ±1.5°C ABOVE 100°C
Length (mm) 405
Division (º C) 1
Immersion depth 125 mm
Max temperature (º C) 200
Fill Red Spirit
Material Glass
Make DURAC
Model B60803-0700
Manufacturer number B60803-0700
Brand H-B Instrument Company

Calibrations
Qty/Price
Availability

EACH
Usually ships in 23 days.




Technical Resources
 
Related Products for Durac Oven Thermometer, 0/200°C, 405mm Length, 125mm Immersion
Image
  Product Detail
SKU

Durac Oven Thermometer, 0/200°C, 405mm Length, 125mm Immersion, PFA Coated

Category: Red Spirit Filled Glass Thermometers
Description: H-B Instrument Durac Red Spirit Oven Glass Thermometers
Brand: H-B Instrument Company
Model Number: B60803-0800


Durac Oven Thermometer, 0/200°C, 280mm Length, 76mm Immersion

Category: Red Spirit Filled Glass Thermometers
Description: H-B Instrument Durac Red Spirit Oven Glass Thermometers
Brand: H-B Instrument Company
Model Number: B60803-0500


Durac Oven Thermometer, 0/200°C, 280mm Length, 76mm Immersion, Pfe Coated

Category: Red Spirit Filled Glass Thermometers
Description: H-B Instrument Durac Red Spirit Oven Glass Thermometers
Brand: H-B Instrument Company
Model Number: B60803-0600


DURAC PLUS, BLUE Spirit, General Purpose Thermometer, -1/51C, 405mm length, partial immersion

Category: H-B Instrument Durac Plus Blue Spirit Laboratory Glass Thermometers
Description: H-B Instrument Durac Plus Blue Spirit Laboratory Precision Accuracy Glass Thermometers, Celsius-scale fractional-degree
Brand: H-B Instrument Company
Model Number: B60700-0000


Enviro-safe General Purpose Thermometer, -1/51c, 405mm Length, 76mm Immersion

Category: 100% Non-Hazardous Liquid Filled Glass Thermometers
Description: H-B Instrument Enviro-Safe
Brand: H-B INSTRUMENT COMPANY
Model Number: 20502


Durac Oven Thermometer, 35/200°C, 180mm Length, Total Immersion

Category: Red Spirit Filled Glass Thermometers
Description: H-B Instrument Durac Red Spirit Oven Glass Thermometers
Brand: H-B Instrument Company
Model Number: B60803-1100


Durac Oven Thermometer, 35/200°C, 180mm Length, Total Immersion, Pfe Coated

Category: Red Spirit Filled Glass Thermometers
Description: H-B Instrument Durac Red Spirit Oven Glass Thermometers
Brand: H-B Instrument Company
Model Number: B60803-1200


Durac Oven Thermometer, 20/130°C, 135mm Length, Total Immersion

Category: Red Spirit Filled Glass Thermometers
Description: H-B Instrument Durac Red Spirit Oven Glass Thermometers
Brand: H-B Instrument Company
Model Number: B60803-0900


Durac Oven Thermometer, 20/130°C, 135mm Length, Total Immersion, PFA Coated

Category: Red Spirit Filled Glass Thermometers
Description: H-B Instrument Durac Red Spirit Oven Glass Thermometers
Brand: H-B Instrument Company
Model Number: B60803-1000


Durac Plus Armored Thermometer, -10/260°C, 405mm Length, 76mm Immersion

Category: H-B Instrument Durac Plus Blue Spirit Armored Glass Thermometers
Description: H-B Instrument Durac Plus Blue Spirit Armored Glass Thermometers, Celsius-scale
Brand: H-B Instrument Company
Model Number: B60740-0400


Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
Questions and Answers

Do you have questions about this product?

get answers from real customers and in-house experts with AnswerBox.

1 Question | 1 Answer
Displaying question 1
  • Q:

    Can this thermometer be used with oven to replace the mercury filled one which is place on a thermomether holder on top?
    Asked on 9/14/2011 by Sylvia Enid from Puerto Rico

    1 answer

    • Staff Reviewer

      A:

      It may be able to replace it. This thermometer has a 125mm immersion depth. So, the tip will need to be in the oven to the specified line in order to provide and accurate reading. If the thermometer tip is not in far enough, or in too far you will not see an accurate temperature reading.

      Answered on 9/15/2011 by Jen from Technical Support
Displaying question 1

Do you have a question about this product? 

Product Reviews