Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Thermo Scientific ERTCO Non Mercury ASTM Glass Thermometer ASTM number 63F
Click to enlarge
More Product Details
Specifications
Calibrations
Technical Resources
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-08007-13
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
YO-17006-03
NIST-Traceable Calibration; 2-4-Point, Liquid-in-Glass Thermometer
$297.84 AUD/EACH/EACH
Usually ships in 22 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Blue Spirit Filled Glass Thermometers
Share
|
Thermo Scientific ERTCO® Non-Mercury ASTM Glass Thermometer, ASTM (063FBLSFC)
YO-08007-13
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Now you can perform ASTM method measurements without the worry of mercury contamination!
Thermometers meet the strict guidelines prescribed by the American Society for Testing and Materials (ASTM)
Non-mercury fill is biodegradable, non-toxic, non-caustic and non-hazardous
Total immersion for highest accuracy
Each thermometer is factory calibrated to standards by the National Institute of Standards and Technology (NIST). The ISO17025 Test Certificate issued by the factory lists an ice-point reading, calibration points, tabulated corrections (to one-tenth of the smallest scale division) for each calibration point, serial and test/identification number of the NIST standard, and explanatory notes defining the conditions under which the test results were made.
What’s included:
Felt-lined hard storage case.
Specifications
Product Type
Glass Thermometer
Min temperature (º F)
18
Max temperature (º F)
90
Length (mm)
379
Division (º F)
0.2
Immersion depth
Total
ASTM number
63F
Calibration points
20, 32, 50, 70, 88°F
Fill
Blue Spirit
Material
Glass
Brand
Ertco
Make
ASTM Glass
Model
063FBLSFC
Manufacturer number
063FBLSFC
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; 2-4-Point, Liquid-in-Glass Thermometer
YO-17006-03
EACH
Usually ships in 22 days.
Add to Cart
Technical Resources
Related Searches for Thermo Scientific ERTCO® Non-Mercury ASTM Glass Thermometer, ASTM (063FBLSFC)
Ertco Blue Spirit Filled Glass Thermometers
Astm Glass Thermometers
Thermo Scientific Ertco Hydrometers
Ertco Red Spirit Filled Glass Thermometers
Thermo Scientific Ertco Thermometer Case 18
Thermo Scientific Ertco Thermometer
Thermo Scientific Ertco
Thermo Scientific Ertco Alcohol Hydrometer
Glass Thermometer with Mercury Inside
Non Mercury Astm Thermometers
Related Products for Thermo Scientific ERTCO® Non-Mercury ASTM Glass Thermometer, ASTM (063FBLSFC)
Image
Product Detail
SKU
Thermo Scientific ERTCO® Non-Mercury ASTM Glass Thermometer, ASTM (062CBBLSFC)
Category: Blue Spirit Filled Glass Thermometers
Description: Thermo Scientific ERTCO
®
Blue Sprirt ASTM Equivalent Glass Thermometers
Brand:
Ertco
Model Number:
062CBBLSFC
YO-08007-10
More Details
Thermo Scientific ERTCO® Ever-Safe® 8" Laboratory Thermometers (1516034)
Category: Blue Spirit Filled Glass Thermometers
Description: Thermo Scientific ERTCO
®
Blue Spirit Ever-Safe
®
Standard Glass Thermometers
Brand:
Ertco
Model Number:
1516034
YO-93909-66
More Details
Thermo Scientific ERTCO® Ever-Safe® 8" Laboratory Thermometers (1516033)
Category: Blue Spirit Filled Glass Thermometers
Description: Thermo Scientific ERTCO
®
Blue Spirit Ever-Safe
®
Standard Glass Thermometers
Brand:
Ertco
Model Number:
1516033
YO-93909-68
More Details
Thermo Scientific ERTCO® Ever-Safe® 8" Laboratory Thermometers (1516036)
Category: Blue Spirit Filled Glass Thermometers
Description: Thermo Scientific ERTCO
®
Blue Spirit Ever-Safe
®
Standard Glass Thermometers
Brand:
Ertco
Model Number:
1516036
YO-93909-70
More Details
Thermo Scientific ERTCO® Ever-Safe® 8" Laboratory Thermometers (1516035)
Category: Blue Spirit Filled Glass Thermometers
Description: Thermo Scientific ERTCO
®
Blue Spirit Ever-Safe
®
Standard Glass Thermometers
Brand:
Ertco
Model Number:
1516035
YO-93909-72
More Details
Thermo Scientific ERTCO® Hydrometer, 0 to 6° API, ASTM 21H, Pl (2021H)
Category: Hydrometers for Degrees API
Description: Thermo Scientific ERTCO
®
Hydrometers, Degrees API (liquids lighter than water)
Brand:
Thermo Scientific Ertco
Model Number:
2021H
YO-08294-00
More Details
Thermo Scientific Nunc Non-treated 700mL Cell Culture Triple Flask 32/
Category: Thermo Scientific Nunc Cell Culture Flasks
Description: Thermo Scientific Nunc Non-Treated Polystyrene Culture Flasks
Brand:
Thermo Scientific Nunc
YO-01928-42
More Details
Thermo Scientific ERTCO® Dual-Scale Hydrometer, 1.000 to 2.000 Spe (2545ADS)
Category: Hydrometers with Dual Scale
Description: Thermo Scientific ERTCO
®
Hydrometers Dual-Scale Specific Gravity/Baume
Brand:
Thermo Scientific Ertco
Model Number:
2545ADS
YO-08287-54
More Details
Thermo Scientific ERTCO® Hydrometer, 0.700 to 0.800 Specific Gravi (2536)
Category: Hydrometers for Specific Gravity
Description: Thermo Scientific ERTCO
®
Specific Gravity Hydrometers, Plain Form. (lighter than water)
Brand:
Thermo Scientific Ertco
Model Number:
2536
YO-08290-10
More Details
Thermo Scientific ERTCO® Enclosed Thermometer, range -90 - 20° (15059145)
Category: Red Spirit Filled Glass Thermometers
Description: Thermo Scientific ERTCO
®
Red Spirit Exact-Temp
™
Bottle Thermometers
Brand:
Ertco
Model Number:
15059145
YO-93907-00
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Thermo Scientific ERTCO® Non-Mercury ASTM Glass Thermometer, ASTM (063FBLSFC)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Thermo Scientific ERTCO® Non-Mercury ASTM Glass Thermometer, ASTM (062CBBLSFC)
|
Next: Thermo Scientific ERTCO® Plastic Hydrometer, 0 to 26.5 Percent Sal (2511-02PL)
31125329-p-content_2 31125329pcontent2