Home  >  Blue Spirit Filled Glass Thermometers   

Thermo Scientific ERTCO Non-Mercury ASTM Glass Thermometer, ASTM number 63F

YO-08007-13
Loading price...

Usually ships in 14 days.
Qty

  • Now you can perform ASTM method measurements without the worry of mercury contamination!
  • Thermometers meet the strict guidelines prescribed by the American Society for Testing and Materials (ASTM)
  • Non-mercury fill is biodegradable, non-toxic, non-caustic and non-hazardous
  • Total immersion for highest accuracy
Each thermometer is factory calibrated to standards by the National Institute of Standards and Technology (NIST). The ISO17025 Test Certificate issued by the factory lists an ice-point reading, calibration points, tabulated corrections (to one-tenth of the smallest scale division) for each calibration point, serial and test/identification number of the NIST standard, and explanatory notes defining the conditions under which the test results were made.
What’s included: Felt-lined hard storage case.
Product Type Glass Thermometer
Min temperature (º F) 18
Max temperature (º F) 90
Length (mm) 379
Division (º F) 0.2
Immersion depth Total
ASTM number 63F
Calibration points 20, 32, 50, 70, 88°F
Fill Blue Spirit
Material Glass
Make ASTM Glass
Model 063FBLSFC
Manufacturer number 063FBLSFC
Brand Ertco

Calibrations
Qty/Price
Availability

EACH
Usually ships in 23 days.




Technical Resources
 
Related Products for Thermo Scientific ERTCO Non-Mercury ASTM Glass Thermometer, ASTM number 63F
Image
  Product Detail
SKU

Thermo Scientific ERTCO Non-Mercury ASTM Glass Thermometer, ASTM number 62C

Category: Blue Spirit Filled Glass Thermometers
Description: Thermo Scientific ERTCO® Blue Sprirt ASTM Equivalent Glass Thermometers
Brand: Ertco


Thermo Scientific ERTCO Dual-Scale Hydrometer, 1.000 to 2.000 Specific Gravity, 0.020 Divisions, 0 to 72° Baume, 1.0° Division, Plain Form

Category: Hydrometers with Dual Scale
Description: Thermo Scientific ERTCO® Hydrometers Dual-Scale Specific Gravity/Baume
Brand: Thermo Scientific Ertco


Thermo Scientific ERTCO Hydrometer, 9 to 21° API, ASTM 2H, Plain Form, 330 mm

Category: Hydrometers for Liquids Lighter than Water
Description: Thermo Scientific ERTCO® Hydrometers, Degrees API (liquids lighter than water)
Brand: Ertco


Thermo Scientific ERTCO Enclosed Thermometer, range -90 - 20°C

Category: Red Spirit Filled Glass Thermometers
Description: Thermo Scientific ERTCO® Red Spirit Exact-Temp Bottle Thermometers
Brand: Ertco


Thermo Scientific ERTCO Enclosed Thermometer, range -30 - 0°C

Category: Red Spirit Filled Glass Thermometers
Description: Thermo Scientific ERTCO® Red Spirit Exact-Temp Bottle Thermometers
Brand: Ertco


Thermo Scientific ERTCO Enclosed Thermometer, range -5 - 15°C

Category: Red Spirit Filled Glass Thermometers
Description: Thermo Scientific ERTCO® Red Spirit Exact-Temp Bottle Thermometers
Brand: Ertco


Thermo Scientific ERTCO Enclosed Thermometer, range 18 - 50°C

Category: Red Spirit Filled Glass Thermometers
Description: Thermo Scientific ERTCO® Red Spirit Exact-Temp Bottle Thermometers
Brand: Ertco


Thermo Scientific ERTCO Enclosed Thermometer, range 20 - 130°C

Category: Red Spirit Filled Glass Thermometers
Description: Thermo Scientific ERTCO® Red Spirit Exact-Temp Bottle Thermometers
Brand: Ertco


Thermo Scientific ERTCO Enclosed Thermometer, range -5 - 20°C

Category: Red Spirit Filled Glass Thermometers
Description: Thermo Scientific ERTCO® Red Spirit Exact-Temp™ Bottle Thermometers
Brand: Ertco


Thermo Scientific ERTCO Enclosed Thermometer, range 35 - 200°C

Category: Red Spirit Filled Glass Thermometers
Description: Thermo Scientific ERTCO® Red Spirit Exact-Temp Bottle Thermometers
Brand: Ertco


Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
Questions and Answers
Product Reviews

REVIEW SNAPSHOT®

by PowerReviews
Thermo Scientific ERTCO<reg> Non-Mercury ASTM Glass Thermometer, ASTM number 63F
 
2.0

(based on 1 review)

Reviewed by 1 customer

Displaying review 1

Back to top

(2 of 3 customers found this review helpful)

 
2.0

EXPENSIVE!

By lab rat

from Peoria, IL

About Me Academic/Professional Researcher

Pros

  • Works Properly

Cons

    Best Uses

    • In The Lab

    Comments about Thermo Scientific ERTCO<reg> Non-Mercury ASTM Glass Thermometer, ASTM number 63F:

    I usually use the ones that you show as discontinued in your catalog. They're also blue, they have the name performatherm. Why don't u keep selling them? they're just as good- better, in fact and 1/2 the cost AND made right here in the good old USA

    Displaying review 1

    Back to top