Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Thermo Scientific ERTCO Hydrometer 9 to 21 API ASTM 2H Plain Form 330 mm
Click to enlarge
More Product Details
Specifications
Accessories
Calibrations
Technical Resources
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-08294-52
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
YO-17107-00
NIST-Traceable Calibration; Hydrometer with test data, one-point (midpoint of Hydrometer's range)
$242.00 AUD/EACH/EACH
Usually ships in 21 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Hydrometers for Liquids Lighter than Water
Share
|
Thermo Scientific ERTCO® Hydrometer, 9 to 21° API, ASTM 2H, Pl (2002H)
YO-08294-52
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
For liquids lighter than water.
Specifications
Product Type
Hydrometer, Degrees API
Manufacturer number
15059599
Form
Plain
Length (mm)
330
Model
2002H
Brand
Thermo Scientific Ertco
Accessories
Qty
/
Price
Availability
Hydrometer Case for 380 mm Length Hygrometer
YO-08285-97
EACH
Call for availability.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Hydrometer with test data, one-point (midpoint of Hydrometer's range)
YO-17107-00
EACH
Usually ships in 21 days.
Add to Cart
Technical Resources
Related Searches for Thermo Scientific ERTCO® Hydrometer, 9 to 21° API, ASTM 2H, Pl (2002H)
Thermo Scientific Ertco Hydrometers
Thermo Scientific Ertco Alcohol Hydrometer
Thermo Scientific Ertco
Thermo Scientific Ertco Thermometer
Thermo Scientific Ertco Thermometer Case 18
H-B Instruments Api Astm Plain
Scientific Products Hydrometer
Thermo Scientific Nalgene General Accessories
Thermo Scientific Nunc Labware
Thermo Scientific Nalgene Fluorinated Jerricans
Related Products for Thermo Scientific ERTCO® Hydrometer, 9 to 21° API, ASTM 2H, Pl (2002H)
Image
Product Detail
SKU
H-B Instrument Hydrometer, 9 to 21° API, ASTM 2H
Category: Hydrometers for Degrees API
Description: H-B Instrument API ASTM Plain Form Hydrometers
Brand:
H-B Instrument Company
Model Number:
2002H
YO-08299-24
More Details
Thermo Scientific ERTCO® Hydrometer, 0 to 6° API, ASTM 21H, Pl (2021H)
Category: Hydrometers for Degrees API
Description: Thermo Scientific ERTCO
®
Hydrometers, Degrees API (liquids lighter than water)
Brand:
Thermo Scientific Ertco
Model Number:
2021H
YO-08294-00
More Details
Thermo Scientific ERTCO® Hydrometer, 0.700 to 0.800 Specific Gravi (2536)
Category: Hydrometers for Specific Gravity
Description: Thermo Scientific ERTCO
®
Specific Gravity Hydrometers, Plain Form. (lighter than water)
Brand:
Thermo Scientific Ertco
Model Number:
2536
YO-08290-10
More Details
Thermo Scientific ERTCO® Dual-Scale Hydrometer, 0.800 to 0.900 Spe (2537DS)
Category: Hydrometers for both Heavier and Lighter than Water
Description: Thermo Scientific ERTCO® Hydrometers Dual-Scale Specific Gravity/Baume
Brand:
Ertco
Model Number:
2537DS
YO-08287-51
More Details
Thermo Scientific ERTCO® Dual-Scale Hydrometer, 1.000 to 2.000 Spe (2545ADS)
Category: Hydrometers with Dual Scale
Description: Thermo Scientific ERTCO
®
Hydrometers Dual-Scale Specific Gravity/Baume
Brand:
Thermo Scientific Ertco
Model Number:
2545ADS
YO-08287-54
More Details
Thermo Scientific ERTCO® Plastic Hydrometer, 0 to 26.5 Percent Sal (2511-02PL)
Category: Hydrometers for Salt
Description: Thermo Scientific ERTCO® Plastic Hydrometers, Salt (percent by weight)
Brand:
Ertco
Model Number:
2511-02PL
YO-08266-30
More Details
Thermo Scientific ERTCO® Sugar Hydrometer, 39 to 51% Brix, Plain F (2520)
Category: Hydrometers for Sugar, Brix Scale
Description: Thermo Scientific ERTCO® Plain Form for Sugar Hydrometers, (brix Scale)
Brand:
Ertco
Model Number:
2520
YO-08286-08
More Details
Thermo Scientific ERTCO® Enclosed Thermometer, range -90 - 20° (15059145)
Category: Red Spirit Filled Glass Thermometers
Description: Thermo Scientific ERTCO
®
Red Spirit Exact-Temp
™
Bottle Thermometers
Brand:
Ertco
Model Number:
15059145
YO-93907-00
More Details
Thermo Scientific ERTCO® Enclosed Thermometer, range -30 - 0°C (15059148)
Category: Red Spirit Filled Glass Thermometers
Description: Thermo Scientific ERTCO
®
Red Spirit Exact-Temp
™
Bottle Thermometers
Brand:
Ertco
Model Number:
15059148
YO-93907-02
More Details
Thermo Scientific ERTCO® Enclosed Thermometer, range -5 - 15°C (15059147)
Category: Red Spirit Filled Glass Thermometers
Description: Thermo Scientific ERTCO
®
Red Spirit Exact-Temp
™
Bottle Thermometers
Brand:
Ertco
Model Number:
15059147
YO-93907-10
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Thermo Scientific ERTCO® Hydrometer, 9 to 21° API, ASTM 2H, Pl (2002H)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Thermo Scientific ERTCO® Hydrometer, 0.700 to 0.800 Specific Gravi (2536)
|
Next: Thermo Scientific ERTCO® Magnifier, thermometer scale magnifier (20905)
31125329-p-content_2 31125329pcontent2