Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
YO-08711-25
Ford Viscosity Cup
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Technical information
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-08711-25
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Viscosity Cups
Share
|
Ford viscosity cup #4 with certificate (SC-0174)
YO-08711-25
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Use accessory stand with bath ports to measure viscosity at various or critcal temperatures
±2% production tolerance
These new Ford viscosity cups are extremely easy to use. Simply pour your sample into the cup and measure the time it takes for the liquid to flow through the orifice until the first break in the liquid stream. Use the included table to convert the elapsed time to centistokes (cSt).
The cups are made from solid aluminum; the orifice is made from brass. All the cups are calibrated to NIST-traceable oils.
An accessory kit, a stand, and a stand with water bath ports are sold separately.
Specifications
Product Type
Viscosity Cups
NIST-traceable
Yes
Cup number
4
Range
70 to 370 cst
Accessories
Qty
/
Price
Availability
Ford cup accessory kit
YO-08711-50
EACH
Call for availability.
Add to Cart
Ford cup stand
YO-08711-60
EACH
Call for availability.
Add to Cart
Technical Resources
Ford viscosity cups conform to ASTM D 333, D 365, and D 1200.
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
L'affaire est ketchup? Pas si simple que ça...les mesures de viscosité
In the Thick of Things: Measuring Viscosity
Technical Expertise: How Cole-Parmer Application Specialists Respond to Customers' Needs
Show All...
Viscous Fluid Behavior
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Related Searches for Ford viscosity cup #4 with certificate (SC-0174)
4 Ford Viscosity Cup Canada
4 Ford Viscosity Cups
Cole-Parmer Ford Viscosity Cups
Dimensions of a Ford Viscosity Cup
Ford Viscosity Cup 2 Millimeter Diameter
Ford Viscosity Cup Stand
Ez Series Viscosity Cup 4
Accessories for Ford Viscosity Cups
4 Ford Viscosity
Number 2 Shell Viscosity Cup
Related Products for Ford viscosity cup #4 with certificate (SC-0174)
Image
Product Detail
SKU
Ford viscosity cup #2 with certificate (SC-7201)
Category: Viscosity Cups
Description: Ford Viscosity Cups
Brand:
Cole-Parmer
Model Number:
SC-7201
YO-08711-05
More Details
Ford viscosity cup #4 (PV-0174)
Category: Viscosity Cups
Description: Ford Viscosity Cups
Brand:
Cole-Parmer
Model Number:
PV-0174
YO-08711-20
More Details
Ford viscosity cup #3 with certificate
Category: Viscosity Cups
Description: Ford Viscosity Cups
Brand:
Cole-Parmer
Model Number:
SC-0173
YO-08711-15
More Details
Ford viscosity cup #2 (PV-7201)
Category: Viscosity Cups
Description: Ford Viscosity Cups
Brand:
Cole-Parmer
Model Number:
PV-7201
YO-08711-00
More Details
Ford viscosity cup #3 (PV-0173)
Category: Viscosity Cups
Description: Ford Viscosity Cups
Brand:
Cole-Parmer
Model Number:
PV-0173
YO-08711-10
More Details
EZ series viscosity cup #4 with certificate
Category: Viscosity Cups
Description: EZ Series Viscosity Cups
Brand:
EZ
Model Number:
EZ4/C
YO-08700-32
More Details
EZ series viscosity cup #1 with certificate
Category: Viscosity Cups
Description: EZ Series Viscosity Cups
Brand:
EZ
Model Number:
EZ1/C
YO-08700-02
More Details
EZ series viscosity cup #2 with certificate
Category: Viscosity Cups
Description: EZ Series Viscosity Cups
Brand:
EZ
Model Number:
EZ2/C
YO-08700-12
More Details
EZ series viscosity cup #3 with certificate
Category: Viscosity Cups
Description: EZ Series Viscosity Cups
Brand:
EZ
Model Number:
EZ3/C
YO-08700-22
More Details
EZ series viscosity cup #4
Category: Viscosity Cups
Description: EZ Series Viscosity Cups
Brand:
EZ
Model Number:
EZ4
YO-08700-30
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Ford viscosity cup #4 with certificate (SC-0174)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Ford viscosity cup #4 (PV-0174)
|
Next: Foreline dry ice cold trap, 1/2" OD straight tube connection
31125329-p-content_2 31125329pcontent2