Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
YO-09200-32
Three heaters (models 09200-30) shown on 55-gallon drum
Click to enlarge
More Product Details
Specifications
Technical Resources
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-09200-32
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Drum and Pail Heaters
Share
|
Sorry, this item is now discontinued but has a direct replacement.
You can compare specifications to the direct replacement YO-03106-69
,
browse similar items
or
Contact us for more information.
3" wide flexible drum heater, 55 gallon, 1000 watt, 240 VAC, 70-425de (YO-09200-32)
YO-09200-32
Loading price...
Discontinued
Qty
:
Save to shopping list
Wrap around drums to maintain temperature or viscosity
Built-in thermostat provides adjustable temperature control
For use with metal drums
These electric heaters combine adjustable temperature control with a variable power output selector. The power output selector provides three wattage levels for a given thermostat setting and voltage rating. High-wattage levels heats drum or pail to set temperature quickly. Medium- and low-wattage levels allow closer temperature control around set point with minimal overshoot. A pilot light indicates when the heating element is activated.
Choose from two thermostate ranges: low (60 to 250ºF). Temperature detector is located on the outside of the drum or pail away from the active heating element. All heaters come with a spring-loaded toggle clamp for a snug fit.
Specifications
Product Type
Drum
Min temperature (º F)
70
Min temperature (º C)
21
Max temperature (º C)
87
Diameter
21.5 in to 23.5 in
Watts
1000
Power (VAC)
240
Drum Size (gal.)
55
Max temperature (º F)
190
Drum Type
Metal
Brand
Cole-Parmer
Manufacturer number
SR4317
Model
SR4317
Technical Resources
Related Searches for 3" wide flexible drum heater, 55 gallon, 1000 watt, 240 VAC, 70-425de (YO-09200-32)
Cole-Parmer Flexible Drum Heaters with Thermostat
Drum Heaters
Ogden Metal Band Drum Heaters
55 Gallon Drums of Heptane
Phosphoric Acid 55 Gallon Drum
Dimensions of a 55 Gallon Drum
Hand Pump for 55 Gallon Drum
55 Gallon Heater
Metal 55 Gallon Drums
55 Gallon Oil Drums
Related Products for 3" wide flexible drum heater, 55 gallon, 1000 watt, 240 VAC, 70-425de (YO-09200-32)
Image
Product Detail
SKU
3" wide flexible drum heater, 55 gallon, 1000 watt, 120 VAC, 70-425de (YO-09200-30)
Category: Drum and Pail Heaters
Description: Cole-Parmer® Flexible Drum Heaters with Thermostat
Brand:
Cole-Parmer
YO-09200-30
More Details
9 1/2" wide flexible drum heater, 55 gallon, 1500 watt, 240 VAC, 70-19 (YO-09203-32)
Category: Drum and Pail Heaters
Description: Cole-Parmer® Flexible Drum Heaters with Thermostat
Brand:
Cole-Parmer
YO-09203-32
More Details
3" wide flexible drum heater, 5 gallon, 300 watt, 120 VAC, 70-425° (YO-09200-00)
Category: Drum and Pail Heaters
Description: Cole-Parmer® Flexible Drum Heaters with Thermostat
Brand:
Cole-Parmer
YO-09200-00
More Details
3" wide flexible drum heater, 15 gallon, 500 watt, 120 VAC, 70-425deg (YO-09200-10)
Category: Drum and Pail Heaters
Description: Cole-Parmer® Flexible Drum Heaters with Thermostat
Brand:
Cole-Parmer
YO-09200-10
More Details
3" wide flexible drum heater, 30 gallon, 750 watt, 120 VAC, 70-425deg (YO-09200-20)
Category: Drum and Pail Heaters
Description: Cole-Parmer® Flexible Drum Heaters with Thermostat
Brand:
Cole-Parmer
YO-09200-20
More Details
9 1/2" wide flexible drum heater, 55 gallon, 750 watt, 240 VAC, 70-140 (YO-09203-36)
Category: Drum and Pail Heaters
Description: Cole-Parmer® Flexible Drum Heaters with Thermostat
Brand:
Cole-Parmer
YO-09203-36
More Details
9 1/2" wide flexible drum heater, 5 gallon, 1000 watt, 120 VAC, 70-190 (YO-09203-00)
Category: Drum and Pail Heaters
Description: Cole-Parmer® Flexible Drum Heaters with Thermostat
Brand:
Cole-Parmer
YO-09203-00
More Details
9 1/2" wide flexible drum heater, 15 gallon, 1000 watt, 120 VAC, 70-19 (YO-09203-10)
Category: Drum and Pail Heaters
Description: Cole-Parmer® Flexible Drum Heaters with Thermostat
Brand:
Cole-Parmer
YO-09203-10
More Details
9 1/2" wide flexible drum heater, 55 gallon, 1500 watt, 120 VAC, 70-19 (YO-09203-30)
Category: Drum and Pail Heaters
Description: Cole-Parmer® Flexible Drum Heaters with Thermostat
Brand:
Cole-Parmer
YO-09203-30
More Details
9 1/2" wide flexible drum heater, 55 gallon, 750 watt, 120 VAC, 70-140 (YO-09203-34)
Category: Drum and Pail Heaters
Description: Cole-Parmer® Flexible Drum Heaters with Thermostat
Brand:
Cole-Parmer
YO-09203-34
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
3" wide flexible drum heater, 55 gallon, 1000 watt, 240 VAC, 70-425de (YO-09200-32)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: 3" wide flexible drum heater, 55 gallon, 1000 watt, 120 VAC, 70-425de (YO-09200-30)
|
Next: 3" x 5" shirt pocket, spiral notebook (135-12)
31125329-p-content_2 31125329pcontent2