Fowler Deburring, Cleaning and Countersink Set (52483888)

YO-09254-15
Loading price...

Call for availability.
Qty

  • A complete kit for all applications
The set has everything needed to clean and prepare delicate work, Set includes: 1 universal handle, 2 adapters, 2 deburring blades, 2 countersinks (20mm/.47" and 30mm/1.18" diameter), 4 cleaner blades.
What's Included:
11 tools and a plastic molded case
Product Type Tool Sets
Qty/pk 10
Brand Fowler
Manufacturer number 52483888





Technical Resources
 
Related Products for Fowler Deburring, Cleaning and Countersink Set (52483888)
Image
  Product Detail
SKU

pHydrion Ultrafine pH Paper Set, Range 0 - 13

Category: pH Test Strips and Indicators
Description: pHydrion Ultrafine pH Paper Set
Brand: Phydrion


Masterflex PTFE-tubing sets, 2mm ID, 4mm OD, set of two.

Category: L/S Spooled Pump Tubing for Masterflex L/S Pumps
Description: L/S PTFE Tube Sets for use with Masterflex L/S PTFE Tubing Pump Head
Brand: Masterflex


Masterflex PTFE-tubing sets, 4mm ID, 6mm OD, set of two.

Category: L/S Spooled Pump Tubing for Masterflex L/S Pumps
Description: L/S PTFE Tube Sets for use with Masterflex L/S PTFE Tubing Pump Head
Brand: Masterflex


Foam Laboratory Brush Set (Universal, 9", and 18"L); 3/Set

Category: Laboratory Brushes
Description: Foam Laboratory Brushes
Brand: Guaranteed In Stock Program


Oakton electrode cleaning solution

Category: pH/Ion Buffers, and Storage Solutions
Description: Oakton Electrode Cleaning and Storage Solutions
Brand: Oakton


Fowler Digi-Level Electronic Protractor, 0 to 360 ° (4 x 90 °) range

Category: Measuring Tools
Description: Fowler Digital Protractor
Brand: Fowler


Fowler 18" Straight Edge (52480050)

Category: Measuring Tools
Description: Fowler Straight Edge
Brand: Fowler


Fowler 24" Straight Edge (52480055)

Category: Measuring Tools
Description: Fowler Straight Edge
Brand: Fowler


52-264012: Fowler 12" /300MM Pre-Certified Flexible Rule

Category: Measuring Tools
Description: Fowler NIST-Certified Rules
Brand: Fowler


52-262-624:6in12in /24in Pre-Cert Rule Set (52-262-624)

Category: Measuring Tools
Description: Fowler NIST-Certified Rules
Brand: Fowler


Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
Questions and Answers
Product Reviews

Fowler Deburring, Cleaning and Countersink Set (52483888)
Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended
Popularity |  Favorites |  Title |  Price |  Part Number |  Category
Title |  Brand |  Category |  Price |  Popularity |  Best Sellers

Previous: Fowler 24" Straight Edge (52480055)  |  Next: Fowler Digi-Level Electronic Protractor, 0 to 360 ° (4 x 90 °) range