55 Gallon, Full Coverage, Drum Heater - 240 V (FGDHC55240)

YO-09300-55
Loading price...

for availability.
Qty

  • Provides safe, uniform heat to 55-gallon metal or plastic drums
  • Built-in adjustable thermoswitch prevents temperature runaway
  • Lightweight flexible design makes installation quick and easy
These insulated drum heaters provide a safe and efficient method of temperature and viscosity control for liquids in 55-gallon drums. Simply wrap insulated drum heater around a 55-gallon drum, adjust the built-in thermostat to desired temperature setting, and uniform heat is evenly distributed around the entire drum. The grounded heating element provides a high-watt density, ensuring rapid heating times—one inch thick fiberglass insulation prevents heat loss. Prevent further heat loss by ordering the drum insulation top cover (optional) below.
Drum heaters feature a VELCRO® seal for easy installation and removal and models are available for both metal and plastic 55-gallon drums. Heaters come with standard three-prong plug (240 VAC models do not come with plugs) and are intended for indoor use in non-hazardous areas only.
Not CSA approved
Product Type Drum
Min temperature (º C) 10
Min temperature (º F) 50
Max temperature (º C) 218
Diameter 22.3 in
Watts 1600
Power (VAC) 240
Power (Amps) 6.6
Max temperature (º F) 425
Drum Size (gal.) 55
Drum Type Metal
Brand BriskHeat

Accessories
Qty/Price
Availability

EACH

for availability.




Technical Resources
 
Related Products for 55 Gallon, Full Coverage, Drum Heater - 240 V (FGDHC55240)
Image
  Product Detail
SKU

55 Gallon, Full Coverage, Drum Heater - 240 V (FGPDHC55240D)

Category: Drum Heaters
Description: BriskHeat Full Coverage Insulated Drum Heaters
Brand: BriskHeat
Model Number: FGPDHC55240D


55 Gallon, Full Coverage, Drum Heater - 120v (FGDHC55120)

Category: Drum Heaters
Description: BriskHeat Full Coverage Insulated Drum Heaters
Brand: BriskHeat
Model Number: FGDHC55120


55 Gallon, Full Coverage, Drum Heater - 120 V (FGPDHC55120)

Category: Drum Heaters
Description: BriskHeat Full Coverage Insulated Drum Heaters
Brand: BriskHeat
Model Number: FGPDHC55120


Drum heater, 55-gallon, 60 to 250°F, 3000 watts, 240 VAC

Category: Drum Heaters
Description: Metal Band Drum Heaters
Brand: Ogden
Model Number: TRX-55-L/R-220V


Drum heater, 55-gallon, 200 to 400°F, 3000 watts, 240 VAC

Category: Drum Heaters
Description: Metal Band Drum Heaters
Brand: Ogden
Model Number: TRX-55-H/R-220V


Drum heater, 55-gallon, 60 to 250°F, 2000 watts, 120 VAC

Category: Drum Heaters
Description: Metal Band Drum Heaters
Brand: Ogden
Model Number: TRX-55-L/R-115V


Drum heater, 55-gallon, 200 to 400°F, 2000 watts, 120 VAC

Category: Drum Heaters
Description: Metal Band Drum Heaters
Brand: Ogden
Model Number: TRX-55-H/R-115V


3" wide flexible drum heater, 55 gallon, 1000 watt, 240 VAC, 70-425°F for metal drums

Category: Drum and Pail Heaters
Description: Cole-Parmer® Flexible Drum Heaters with Thermostat
Brand: Cole-Parmer
Model Number: SR4317


9 1/2" wide flexible drum heater, 55 gallon, 1500 watt, 240 VAC, 70-190°F for metal drums

Category: Drum and Pail Heaters
Description: Cole-Parmer® Flexible Drum Heaters with Thermostat
Brand: Cole-Parmer
Model Number: SR4321


9 1/2" wide flexible drum heater, 55 gallon, 750 watt, 240 VAC, 70-140°F for plastic drums

Category: Drum and Pail Heaters
Description: Cole-Parmer® Flexible Drum Heaters with Thermostat
Brand: Cole-Parmer
Model Number: SR4325


Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
Questions and Answers
Product Reviews

55 Gallon, Full Coverage, Drum Heater - 240 V (FGDHC55240)
Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended
Popularity |  Favorites |  Title |  Price |  Part Number |  Category
Title |  Brand |  Category |  Price |  Popularity |  Best Sellers

Previous: 55 Gallon, Full Coverage, Drum Heater - 120v (FGDHC55120)  |  Next: 55 Gallon, Full Coverage, Drum Heater - 240 V (FGPDHC55240D)