Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
YO-10775-05
Representative photo only. Shown with optional SS stand, sold separately. Market Forge 16" Sterilmatic
®
Benchtop Sterilizer
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-10775-05
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Sterilizers
Share
|
Market Forge 16" Sterilmatic Benchtop Sterilizer, Standard, Set Temper (YO-10775-05)
YO-10775-05
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Fast preheat timereaches 121°C (250°F) in less than 10 minutes
Heating elements on the outside of the sterilizer chamber never come in contact with water
These benchtop sterilizers are ideal for steam sterilization of wrapped or unwrapped dressings, labware, and liquids. Choose from the standard sterilizer with sterilizing temperature set at 121°C (250°F) or the laboratory sterilizer with an adjustable sterilizing temperature from 110 to 121°C (230 to 250°F).
Safety features include a door that locks until steam pressure is completely relieved from the chamber and low water-level cut-off. All models are rated for four-wire, three-phase operation. Sterilizer chambers are ASME-approved. Optional stainless steel stand, perforated tray, and basket sold separately.
Specifications
Product Type
Sterilizers
Capacity (cu ft)
3
Reservoir capacity (Liters)
5.7
Average cycle time
12 minutes (cold)
Chamber size (" D)
26
Chamber size (" Dia)
16
Chamber size (cm)
66 D x 40.6 dia
Max temperature (º C)
121
Max temperature (º F)
250
Dimensions (" W)
18-3/4
Dimensions (" H)
29
Dimensions (" D)
31
Dimensions (cm)
47.6 W x 73.7 H x 78.7 D cm
Timer range
0 - 60 mins
Power (VAC)
220/380
Power (Hz)
50
Power (Amps)
26
Chamber construction
Aluminum
Display
Dial gauge
Max psi
15
Manufacturer number
95-2902, 220/380
Brand
Market Forge
Accessories
Qty
/
Price
Availability
Stainless steel stand for Sterilmatic sterilizers
YO-10775-50
EACH
Call for availability.
Add to Cart
Perforated tray for Sterilimatic sterilizers
YO-10775-55
EACH
Call for availability.
Add to Cart
Basket, 12"L x 20"W x 2-1/2"H
YO-10775-56
EACH
Call for availability.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
13-14 International Catalog
Related technical articles (Roll over for summary)
Equipping a Laboratory: A Study in Customer Service Excellence
Electron zap turns flimsy plastic into sturdy shrink wrap
Basics of Cartridge Filtration
Show All...
Bitter Brew
Show Less...
Related Searches for Market Forge 16" Sterilmatic Benchtop Sterilizer, Standard, Set Temper (YO-10775-05)
Market Forge 16" Sterilmatic Benchtop Sterilizers
Market Forge Sterilmatic Sterilizers
Market Forge Heating Equipment
Benchtop Sterilizers
Sterilization Standards
Sterilmatic Replacement Sterilizer Parts and Components
Sterilmatic Heating Equipment
Heating Mantle Adjustable Size
Cole-Parmer Dry Glass Bead Sterilizer
Dry Glass Bead Sterilizer Suppliers
Related Products for Market Forge 16" Sterilmatic Benchtop Sterilizer, Standard, Set Temper (YO-10775-05)
Image
Product Detail
SKU
Market Forge 16" Sterilmatic Benchtop Sterilizer, Standard, Set Temper (YO-10775-00)
Category: Sterilizers
Description: Market Forge 16" Sterilmatic Benchtop Sterilizers
Brand:
Market Forge
YO-10775-00
More Details
Market Forge 16" Sterilmatic Benchtop Sterilizer, Adjustable Temperatu (YO-10775-10)
Category: Sterilizers
Description: Market Forge 16" Sterilmatic Benchtop Sterilizers
Brand:
Market Forge
YO-10775-10
More Details
Market Forge 16" Sterilmatic Benchtop Sterilizer, Adjustable Temperatu (YO-10775-15)
Category: Sterilizers
Description: Market Forge 16" Sterilmatic Benchtop Sterilizers
Brand:
Market Forge
YO-10775-15
More Details
Thermo Scientific SPECTRONIC™ standards, set of 10 (333150-000)
Category: Calibration Products and Standards
Description: Thermo Scientific SPECTRONIC™ Standards Set for Visible Light Spectroscopy
Brand:
Thermo Scientific
Model Number:
333150-000
YO-02650-57
More Details
Dry Glass Bead Sterilizer, 3" D x 1-3/4" dia sterilizing well; 220 VAC (YO-10779-05)
Category: Sterilizers
Description: Dry Glass Bead Sterilizers
Brand:
Cole-Parmer
YO-10779-05
More Details
Compression Union, 1/16" x 5/8", Stainless Steel, Each (YO-30486-05)
Category: Straight Union Connectors
Description: Straight unions
Brand:
Parker Hannifin
YO-30486-05
More Details
Thermo Scientific Nunc™ Lab-Tek Sterile Petri Dishes, PS, Standard, (YO-14007-05)
Category: Petri Dishes and Contact Plates
Description: Thermo Scientific Nunc Lab-Tek Sterile Petri Dishes, PS
Brand:
Thermo Scientific Nunc
YO-14007-05
More Details
Sterile silicone venting closure (YO-06299-00)
Category: Stoppers
Description: Sterile Venting Closures, Silicone
Brand:
Cole-Parmer
YO-06299-00
More Details
Solid Basket for Yamato Vertical Loading Sterilizers f/10770-series (YO-10770-51)
Category: Yamato Vertical-Loading Laboratory Autoclaves
Description: Yamato Vertical-Loading Sterilizer Baskets
Brand:
Yamato
YO-10770-51
More Details
Solid Basket for Yamato Vertical Loading Sterilizers f/10771-series (YO-10771-51)
Category: Yamato Vertical-Loading Laboratory Autoclaves
Description: Yamato Vertical-Loading Sterilizer Baskets
Brand:
Yamato
YO-10771-51
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Market Forge 16" Sterilmatic Benchtop Sterilizer, Standard, Set Temper (YO-10775-05)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Market Forge 16" Sterilmatic Benchtop Sterilizer, Standard, Set Temper (YO-10775-00)
|
Next: Marsh Bellofram (reg) Type 41 Air Pressure Regulators, 1/4" NPT(F), 0 (960-115-000)
31125329-p-content_2 31125329pcontent2