Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Tuttnauer Heidolph Brinkmann Tabletop Autoclave Digital 2 8 cu ft 220VAC
Click to enlarge
More Product Details
Specifications
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-10780-10
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Sterilizers
Share
|
Tuttnauer/ Heidolph Brinkmann Tabletop Autoclave, Digital 2.8 cu ft, 2 (23210606)
YO-10780-10
Loading price...
Qty
:
Save to shopping list
Offer fast, safe, dependable steam sterilization for your lab
Sterilize liquids, media, instruments, glassware, clothing, or waste
Models available with printer to meet GLP requirements
No plumbing or hard-wiring required
These compact benchtop autoclaves are extremely versatile, offering analog or electronic controls and providing a safe and convenient method for the sterilization of solids and liquids. Both analog and electronic models feature 316 stainless steel sterilization chambers and removable trays that resist corrosion. The continuous pressure purge helps eliminate cool air pockets within the chamber, ensuring thorough, even sterilization. A heatinsulated door with double-locking feature prevents opening while the chamber is under pressure to help minimize dangers to lab personnel. All models feature over-temperature cutoff, over-pressure valve, and a low-water level alarm which helps prevent damage to heating elements by signaling the need to refill the reservoir. A drying cycle helps prevent rusting of delicate instruments and helps dry clothing or absorbent materials.
What's Included:
ASME-certified chamber, stainless steel trays, drain tube, and 6-ft power cord.
Specifications
Product Type
Sterilizer
Capacity (cu ft)
2.8
Reservoir capacity (Liters)
6
Chamber size (" D)
27
Chamber size (" Dia)
15
Max temperature (º C)
134
Min temperature (º C)
100
Min temperature (º F)
212
Max temperature (º F)
273
Dimensions (" W)
34
Dimensions (" H)
21
Dimensions (" D)
26
Timer range
3 - 99 mins
Power (VAC)
220
Power (Hz)
50/60
Power (Amps)
13
Power (watts)
2700
Chamber construction
Stainless Steel
Display
LED
Max psi
40
Shelves
2
Manufacturer number
23210606
Brand
Brinkmann
Model
23210606
Technical Resources
Related technical articles (Roll over for summary)
Equipping a Laboratory: A Study in Customer Service Excellence
Electron zap turns flimsy plastic into sturdy shrink wrap
Basics of Cartridge Filtration
Show All...
Bitter Brew
Show Less...
Related Searches for Tuttnauer/ Heidolph Brinkmann Tabletop Autoclave, Digital 2.8 cu ft, 2 (23210606)
Heidolph Brinkmann
Heidolph Brinkmann Tuttnauer Sterilizers
Heidolph Tabletop Autoclaves
Autoclave Digital Pressure Gauges
Cu Ft Microwave Oven
40 Cu Ft Co2 Incubator
24 Cu Ft Refrigerator
30 Cu Ft Refrigerators
Refrigerator 17 Cu Ft
6 Cu Ft Refrigerator
Related Products for Tuttnauer/ Heidolph Brinkmann Tabletop Autoclave, Digital 2.8 cu ft, 2 (23210606)
Image
Product Detail
SKU
Tuttnauer/ Heidolph Brinkmann Tabletop Autoclave, Digital 0.7 cu ft, 2 (23210258)
Category: Sterilizing Equipment
Description: Heidolph Tabletop Autoclaves
Brand:
Heidolph
Model Number:
23210258
YO-10780-07
More Details
Tuttnauer/ Heidolph Brinkmann Tabletop Autoclave, Digital 0.8 cu ft, 2 (23210452)
Category: Sterilizing Equipment
Description: Heidolph Tabletop Autoclaves
Brand:
Heidolph
Model Number:
23210452
YO-10780-09
More Details
Tuttnauer/ Heidolph Brinkmann Tabletop Autoclave, Digital 0.7 cu ft, 1 (23210207)
Category: Sterilizing Equipment
Description: Heidolph Tabletop Autoclaves
Brand:
Heidolph
Model Number:
23210207
YO-10780-06
More Details
Tuttnauer/ Heidolph Brinkmann Tabletop Autoclave, Digital 0.8 cu ft, 1 (23210401)
Category: Sterilizing Equipment
Description: Heidolph Tabletop Autoclaves
Brand:
Heidolph
Model Number:
23210401
YO-10780-08
More Details
Tuttnauer/ Heidolph Brinkmann Tabletop Autoclave, Digital with Printer (23210410)
Category: Sterilizers
Description: Heidolph Tabletop Autoclaves
Brand:
Brinkmann
Model Number:
23210410
YO-10780-11
More Details
Tuttnauer/ Heidolph Brinkmann Tabletop Autoclave, Digital with Printer (23210461)
Category: Sterilizing Equipment
Description: Heidolph Tabletop Autoclaves
Brand:
Heidolph
Model Number:
23210461
YO-10780-12
More Details
Tuttnauer/ Heidolph Brinkmann Tabletop Autoclave, Digital with Printer (23210568)
Category: Sterilizing Equipment
Description: Heidolph Tabletop Autoclaves
Brand:
Heidolph
Model Number:
23210568
YO-10780-13
More Details
Tuttnauer/ Heidolph Brinkmann Tabletop Autoclave, Analog, 0.7 cu ft, 2 (23210151)
Category: Sterilizing Equipment
Description: Heidolph Tabletop Autoclaves
Brand:
Heidolph
Model Number:
23210151
YO-10780-03
More Details
Tuttnauer/ Heidolph Brinkmann Tabletop Autoclave, Analog 0.7 cu ft, 12 (23210100)
Category: Sterilizing Equipment
Description: Heidolph Tabletop Autoclaves
Brand:
Heidolph
Model Number:
23210100
YO-10780-02
More Details
Tuttnauer/ Heidolph Brinkmann Tabletop Autoclave, Analog 0.8 cu ft, 12 (23210304)
Category: Sterilizing Equipment
Description: Heidolph Tabletop Autoclaves
Brand:
Heidolph
Model Number:
23210304
YO-10780-04
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Tuttnauer/ Heidolph Brinkmann Tabletop Autoclave, Digital 2.8 cu ft, 2 (23210606)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Tuttnauer/ Heidolph Brinkmann Tabletop Autoclave, Digital 0.8 cu ft, 2 (23210452)
|
Next: Tuttnauer/ Heidolph Brinkmann Tabletop Autoclave, Digital with Printer (23210410)
31125329-p-content_2 31125329pcontent2