Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, Fittings, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
YO-68454-30
Pneumatic pump & vacuum, 600 psi to -28 in Hg
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
YO-68454-30
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Calibrators
Share
|
Pneumatic pump & vacuum, 600 psi to -28 in Hg
YO-68454-30
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Hand-held pressure or vacuum generation
Low-cost way to verify performance of pressure instrumentation
Rugged and reliable design
Fine adjustment control
The most efficient pumps available
These pumps are ideal for use with electronic calibrators, digital indicators, analog gauges, pressure transmitters, transducers, relief valves, or similar instrumentation.
The rugged and reliable design of these pumps and the fine adjustment controls added to their design provides a low cost way to verify the performance of pressure instrumentation. Pumps can be purchased independently, or in a kit including carrying case, NPT adapters, seals and hoses.
The SHTP1 hydraulic pump is currently the only hand pump capable of generating pressure up to 15,000 psi, utilizing hydraulic oil, Skydrol or distilled water. The pump includes a unique high pressure or priming selector on the scissor action pump mechanism, making pressure generation above 5,000 psi easier.
The STP1 pneumatic pump generates pressure to 600 psi and vacuum to 28”Hg on the same unit. Made with no plastic parts these pumps are designed to endure harsh environments unlike our competitor’s disposable models.
The SLTP1 pneumatic pump also acts as a dual source of pressure and vacuum over smaller ranges making very fine adjustments easier. The SLTP1 range is 25”Hg vacuum to 30 psi, with 0.00145 psi sensitivity.
Specifications
Product Type
Calibration Pumps
Brand
SI Pressure Instruments
Model
TP1-40
Manufacturer number
TP1-40
Accessories
Qty
/
Price
Availability
Npt Fitting
YO-68454-27
EACH
Call for availability.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
11-12 Catalog
11-12 International Catalog
13-14 Catalog
13-14 International Catalog
Related technical articles (Roll over for summary)
Absolute, Gauge, and Differential Pressure Scales
Common Pressure Terms
Pressure Unit Conversion Constants
Show All...
The Hidden Costs of Mercury-Filled Vacuum Gauges
Show Less...
Related Searches for Pneumatic pump & vacuum, 600 psi to -28 in Hg
Pneumatic Pumps
Si Pressure Instruments Tp1 Pneumatic Pumps
Ryton Pumps - Vacuum Pumps
Dryfast Pumps - Vacuum Pumps
Gast Pumps - Vacuum Pumps
Admiral Pumps - Vacuum Pumps
Welch Pumps - Vacuum Pumps
Cps Pumps - Vacuum Pumps
Ptfe Pumps - Vacuum Pumps
Knf Pumps - Vacuum Pumps
Related Products for Pneumatic pump & vacuum, 600 psi to -28 in Hg
Image
Product Detail
SKU
Pneumatic Pressure and Vacuum Pump, 0-600 PSI; 0-28.5" Hg
Category: Calibrators
Description: GE Druck Pneumatic Pressure and Vacuum Pump
Brand:
Druck
Model Number:
PV211 104
YO-68974-32
More Details
Pump/Vacuum Kit 60
Category: Calibrators
Description: GE (SI Pressure Instruments) TP1 Pneumatic Pumps
Brand:
SI Pressure Instruments
Model Number:
TP1-40-K
YO-68451-17
More Details
PUMP, VACUUM PUMP WITH BRUSHLESS DC MOTOR, 1.5 LPM, 12 VDC, GAS ONLY,
Category: Compact/OEM Vacuum Pumps
Description: Single Head Miniature Diaphragm Pumps
Brand:
Cole-Parmer
YO-79600-21
More Details
PUMP, VACUUM PUMP WITH BRUSHLESS DC MOTOR, 1.5 LPM, 24 VDC, GAS ONLY,
Category: Compact/OEM Vacuum Pumps
Description: Single Head Miniature Diaphragm Pumps
Brand:
Cole-Parmer
YO-79600-22
More Details
PUMP, VACUUM PUMP WITH BRUSHLESS DC MOTOR, 2.5 LPM, 12 VDC, GAS ONLY,
Category: Compact/OEM Vacuum Pumps
Description: Single Head Miniature Diaphragm Pumps
Brand:
Cole-Parmer
YO-79600-23
More Details
PUMP, VACUUM PUMP WITH BRUSHLESS DC MOTOR, 2.5 LPM, 24 VDC, GAS ONLY,
Category: Compact/OEM Vacuum Pumps
Description: Single Head Miniature Diaphragm Pumps
Brand:
Cole-Parmer
YO-79600-24
More Details
Pump, Vacuum Pump, Rotary Vane, 1.5 CFM, 110VAC, 2 Stage Pump (UVP1.5-110)
Category: Rotary Vane Vacuum/Compressor Pumps
Description: Economical Rotary Vane Pumps
Brand:
Cole-Parmer
Model Number:
UVP1.5-110
YO-07164-30
More Details
Pump, Vacuum Pump, Rotary Vane, 1.5 CFM, 220VAC, 2 Stage Pump (UVP1.5-220)
Category: Rotary Vane Vacuum/Compressor Pumps
Description: Economical Rotary Vane Pumps
Brand:
Cole-Parmer
Model Number:
UVP1.5-220
YO-07164-32
More Details
Pump, Vacuum Pump, Rotary Vane, 6.0 CFM, 115/220 VAC, 2 Stage (HVP6)
Category: Rotary Vane Vacuum/Compressor Pumps
Description: Economical Rotary Vane Pumps
Brand:
Cole-Parmer
Model Number:
HVP6
YO-07164-52
More Details
Pump, Vacuum Pump, Rotary Vane, 8.0 CFM, 115/220 VAC, 2 Stage (HVP8)
Category: Rotary Vane Vacuum/Compressor Pumps
Description: Economical Rotary Vane Pumps
Brand:
Cole-Parmer
Model Number:
HVP8
YO-07164-62
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Pneumatic pump & vacuum, 600 psi to -28 in Hg
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Pneumatic Pressure and Vacuum Pump, 0-600 PSI; 0-28.5" Hg
|
Next: Pneumatic Test Pump -13/600psi
31125329-p-content_2 31125329pcontent2