Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
A 314 Bleed Fitting
Click to enlarge
More Product Details
Specifications
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-68462-47
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
General Accessories
Share
|
A-314:Bleed Fitting
YO-68462-47
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Specifications
Product Type
Differential pressure gauge accessories
Related Searches for A-314:Bleed Fitting
Dwyer General Accessories
Colder Metal Quick-Disconnect Fittings
Cole-Parmer Tubing Compatible with Barbed Fittings
Atlas Fit Personal Protective Equipment (Ppe)
Colder all-Plastic Quick-Disconnect Fittings
Cole-Parmer Tubing Compatible with Compression Fittings
Gf Signet Pvdf Tee Fittings
Tefzel Couplings and Tube End Fittings
Tefzel Luer Fittings and Adapters
Cole-Parmer Thermocouple Compression Fittings
Related Products for A-314:Bleed Fitting
Image
Product Detail
SKU
Comark 314 Waterproof Pocket Thermometer with Standard Tip: Range -40
Category: Pocket Thermometers Digital - Food Industry
Description: Comark 314 Waterproof Pocket Thermometer with Standard Tip
Brand:
Comark
Model Number:
314
YO-90025-06
More Details
Sperian® Atlas Fit™ Rubber Coated Cotton/Poly Glove, Medium
Category: Coated Gloves
Description: Sperian
®
Atlas Fit
™
Blue/Gray Rubber Dipped Cotton/Poly Gloves
Brand:
Atlas Fit
Model Number:
300-M
YO-86411-17
More Details
Atlas Fit® Rubber Coated Cotton Poly Glove Black and Gray Sm (320-S)
Category: Coated Gloves
Description: Sperian
®
Atlas Fit
™
Black/Gray Rubber Dipped Cotton/Poly Gloves
Brand:
Atlas Fit
Model Number:
320-S
YO-86411-26
More Details
Atlas Fit® Rubber Coated Cotton Poly Glove Black and Gray Lg (320-L)
Category: Coated Gloves
Description: Sperian
®
Atlas Fit
™
Black/Gray Rubber Dipped Cotton/Poly Gloves
Brand:
Atlas Fit
Model Number:
320-L
YO-86411-28
More Details
Atlas Fit® Nitrile Coated Cotton Poly Glove LG (350-L)
Category: Coated Gloves
Description: Sperian®Atlas Fit™ Nitrile-Dipped Cotton/Poly Gloves
Brand:
Atlas Fit
Model Number:
350-L
YO-86411-38
More Details
Flangeless fittings; 1/16", acetal ferrule; 10/pack (YO-01939-30)
Category: Upchurch Scientific high-pressure chromatography fittings and accessories
Description: Flangeless Fittings
Brand:
UPCHURCH
YO-01939-30
More Details
Fittings; 1/8" x 1/4-28, acetal ferrule; 10/pack (YO-01939-32)
Category: Upchurch Scientific high-pressure chromatography fittings and accessories
Description: Flangeless Fittings
Brand:
UPCHURCH
YO-01939-32
More Details
Flangeless Fittings; 1/16", ETFE male nut; 10/pack (YO-01939-40)
Category: Upchurch Scientific high-pressure chromatography fittings and accessories
Description: Flangeless Fittings
Brand:
UPCHURCH
YO-01939-40
More Details
Fittings; 1/8" x 1/4-28, ETFE male nut; 10/pack (YO-01939-42)
Category: Upchurch Scientific high-pressure chromatography fittings and accessories
Description: Flangeless Fittings
Brand:
UPCHURCH
YO-01939-42
More Details
Stainless Steel fittings; 0.020" bore, tee, ss (YO-01939-50)
Category: Upchurch Scientific high-pressure chromatography fittings and accessories
Description: Stainless Steel Fittings
Brand:
UPCHURCH
YO-01939-50
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
A-314:Bleed Fitting
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: A A A-2-TETRAFLUORO 1G (YO-88282-91)
|
Next: A-BROMO-2 3-DIFLUOROTOLUENE 1G (YO-88285-53)
31125329-p-content_2 31125329pcontent2