Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
YO-11104-41
A&D Orion Analytical Balance, 42/210 g x 0.01 /0.1 mg. Representative Photo Only
Click to enlarge
More Product Details
Specifications
Accessories
Related products
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-11104-41
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
YO-17101-52
NIST Traceable Report With Data, Balances/Scales - all types
$260.61 AUD/EACH/EACH
Usually ships in 21 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
A&D Weighing Analytical Balances
Share
|
A&D Orion Analytical Balance, 42/210 g x 0.01 /0.1 mg
YO-11104-41
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Exceptional combination of remarkable features at affordable prices
Die-cast aluminum alloy housing offers long life and durability
System diagnostics alerts operator of abnormal weighing conditions
Digital tare lets you tare via computer or manually
Parts counting, percent weighing, below-balance weighing, and auto zero tracking
Calibration mass is not included with balance - order separately.
What's Included:
Dust cover and a 120 VAC, 50/60 Hz adapter with 6-ft (1.8 m) cord and two-prong plug. Order optional 220 VAC adapter and RS-232 interface separately below.
Specifications
Product Type
Analytical Balances
Calibration
External
Capacity (g)
51 / 220
Capacity (oz)
2 / 8
Housing dimensions
(21.6 x 30.5 x 31.8 cm)
Dimensions (" W)
8-1/2
Dimensions (" H)
12
Dimensions (" D)
12-1/2
Pan size (dia)
3-3/16" (8.1 cm)
Readability (mg)
0.1 / 0.1
Linearity (mg)
±0.03 / 0.2
Repeatability (mg)
±0.02 / 0.1
Weighing units
g, mg, oz, ozt, ct, dwt, gn, tael
Stability filters
five environmental sensitivity levels
Power (VAC)
120
Output
RS-232
Brand
A&D Weighing
Model
HR-202I
Accessories
Qty
/
Price
Availability
220 VAC Adapter for A&D Balances
YO-01038-65
EACH
Call for availability.
Add to Cart
A&D Pipette Leak Tester Optional Rechargeable Battery
YO-07911-53
EACH
Call for availability.
Add to Cart
Printer, compact
YO-11126-92
EACH
Call for availability.
Add to Cart
Related products
Qty
/
Price
Availability
Cole-Parmer SpatulaBalance
, 300 g x 0.1 g
YO-01019-11
EACH
Usually ships in 14 days or sooner.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST Traceable Report With Data, Balances/Scales - all types
YO-17101-52
EACH
Usually ships in 21 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
13-14 International Catalog
Related technical articles (Roll over for summary)
Gravitational Effects on Balance
Weighing Unit Conversion Table
Common Balance Terms
Show All...
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Related Searches for A&D Orion Analytical Balance, 42/210 g x 0.01 /0.1 mg
A&D Weighing Orion Hr Analytical Balances
A&D Weighing Gemini Autocalibrating Analytical Balances
Sartorius Cubis Semi-Micro and Analytical Balances
Mettler Analytical Balances
Mettler Toledo Excellence Xs Analytical Balance
Ohaus Adventurer Pro Analytical Balances
A&D W and Weighing Analytical Balances
Cole-Parmer Symmetry Pa Analytical Balances
Ohaus Explorer Analytical Balances
Denver Instrument Summit Series Analytical Balances
Related Products for A&D Orion Analytical Balance, 42/210 g x 0.01 /0.1 mg
Image
Product Detail
SKU
A&D Orion Analytical Balance, 210 g
Category: A&D Weighing Analytical Balances
Description: A&D Weighing Orion HR Analytical Balances
Brand:
A&D Weighing
Model Number:
HR-200-C
YO-11104-20
More Details
A&D Gemini Autocalibrating Analytical Balance, 42 g/210 g
Category: A&D Weighing Analytical Balances
Description: A&D Weighing Gemini GR Analytical Balances
Brand:
A&D Weighing
Model Number:
GR-202
YO-11100-80
More Details
A&D Orion Analytical Balance, 60 g
Category: A&D Weighing Analytical Balances
Description: A&D Weighing Orion HR Analytical Balances
Brand:
A&D Weighing
Model Number:
HR-60-C
YO-11104-00
More Details
A&D Orion Analytical Balance, 120 g
Category: A&D Weighing Analytical Balances
Description: A&D Weighing Orion HR Analytical Balances
Brand:
A&D Weighing
Model Number:
HR-120-C
YO-11104-10
More Details
A&D Orion Analytical Balance, 310 g x 0.1 mg, 120 VAC
Category: A&D Weighing Analytical Balances
Description: A&D Weighing Orion HR Analytical Balances
Brand:
A&D Weighing
Model Number:
HR-300I
YO-11104-31
More Details
Mettler Toledo XS Analytical Balance, 41/120 g x 0.1 mg, 115 VAC
Category: Mettler Toledo Analytical Balances
Description: Mettler Toledo Excellence XS Series Analytical Balances
Brand:
Mettler Toledo
Model Number:
XS105DU
YO-11333-08
More Details
Mettler Toledo XS Analytical Balance, 81/220g x 0.01/0.1 mg, 115 VAC
Category: Mettler Toledo Analytical Balances
Description: Mettler Toledo Excellence XS Series Analytical Balances
Brand:
Mettler Toledo
Model Number:
XS205DU
YO-11333-10
More Details
Ohaus Pioneer Analytical Balance, 210 G X 0.1 Mg (115 VAC)
Category: Ohaus Analytical Balances
Description: Ohaus Pioneer Analytical Balances
Brand:
Ohaus
Model Number:
PA 214
YO-11009-58
More Details
A&D Gemini Autocalibrating Analytical Balance, 210 G
Category: A&D Weighing Analytical Balances
Description: A&D Weighing Gemini GR Analytical Balances
Brand:
A&D Weighing
Model Number:
GR-200
YO-11100-70
More Details
Ohaus Discovery Analytical Balances, 210g X 0.1mg
Category: Semi/Micro Analytical Balances
Description: Ohaus Discovery Analytical and Semi-Micro Balances
Brand:
Ohaus
Model Number:
DV214C
YO-11100-75
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
A&D Orion Analytical Balance, 42/210 g x 0.01 /0.1 mg
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: A&D Orion Analytical Balance, 310 g x 0.1 mg, 120 VAC
|
Next: A&D Orion Analytical Balance, 60 g
31125329-p-content_2 31125329pcontent2