Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Spacer
Click to enlarge
More Product Details
Specifications
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-68471-95
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
General Accessories
Share
|
Spacer
YO-68471-95
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Specifications
Product Type
Accessory
Related Searches for Spacer
Cole-Parmer Vertical Slab Gel Spacer Set
Cole-Parmer Dual Vertical Mini-Gel Spacer Sets
Spacers for Gel Electrophoresis
Cole-Parmer Spacers for Gel Electrophoresis
Electrophoresis Set
Gel System
Ptfe Infrared (Ir) Spectroscopy
Thermo Scientific Accessories for Model 28554-15
Ptfe Material Testing & Physical Testing
Beta Cell for Ir Spectroscopy
Related Products for Spacer
Image
Product Detail
SKU
Spacer Set for Cole-Parmer Vertical Slab Gel Systems; 14.5 cm Height, (YO-28573-00)
Category: Cole-Parmer Adjustable Height Single and Dual Vertical Gel Systems
Description: Cole-Parmer Vertical Slab Gel Spacer Set for Height-adjustable units
Brand:
Cole-Parmer
YO-28573-00
More Details
Spacer Set for Cole-Parmer Vertical Slab Gel Systems; 16 cm Height, 0.
Category: Cole-Parmer Adjustable Height Single and Dual Vertical Gel Systems
Description: Cole-Parmer Vertical Slab Gel Spacer Set for Height-adjustable units
Brand:
Cole-Parmer
YO-28573-01
More Details
Spacer Set for Cole-Parmer Vertical Slab Gel Systems; 19 cm Height, 0.
Category: Cole-Parmer Adjustable Height Single and Dual Vertical Gel Systems
Description: Cole-Parmer Vertical Slab Gel Spacer Set for Height-adjustable units
Brand:
Cole-Parmer
YO-28573-02
More Details
Spacer Set for Cole-Parmer Vertical Slab Gel Systems; 22 cm Height, 0.
Category: Cole-Parmer Adjustable Height Single and Dual Vertical Gel Systems
Description: Cole-Parmer Vertical Slab Gel Spacer Set for Height-adjustable units
Brand:
Cole-Parmer
YO-28573-03
More Details
Spacer Set for Cole-Parmer Vertical Slab Gel Systems; 28 cm Height, 0.
Category: Cole-Parmer Adjustable Height Single and Dual Vertical Gel Systems
Description: Cole-Parmer Vertical Slab Gel Spacer Set for Height-adjustable units
Brand:
Cole-Parmer
YO-28573-04
More Details
Spacer Set for Cole-Parmer Vertical Slab Gel Systems; 14.5 cm Height, (YO-28573-05)
Category: Cole-Parmer Adjustable Height Single and Dual Vertical Gel Systems
Description: Cole-Parmer Vertical Slab Gel Spacer Set for Height-adjustable units
Brand:
Cole-Parmer
YO-28573-05
More Details
Spacer Set for Cole-Parmer Vertical Slab Gel Systems; 16 cm Height, 1. (YO-28573-06)
Category: Cole-Parmer Adjustable Height Single and Dual Vertical Gel Systems
Description: Cole-Parmer Vertical Slab Gel Spacer Set for Height-adjustable units
Brand:
Cole-Parmer
YO-28573-06
More Details
Spacer Set for Cole-Parmer Vertical Slab Gel Systems; 19 cm Height, 1. (YO-28573-07)
Category: Cole-Parmer Adjustable Height Single and Dual Vertical Gel Systems
Description: Cole-Parmer Vertical Slab Gel Spacer Set for Height-adjustable units
Brand:
Cole-Parmer
YO-28573-07
More Details
Spacer Set for Cole-Parmer Vertical Slab Gel Systems; 22 cm Height, 1. (YO-28573-08)
Category: Cole-Parmer Adjustable Height Single and Dual Vertical Gel Systems
Description: Cole-Parmer Vertical Slab Gel Spacer Set for Height-adjustable units
Brand:
Cole-Parmer
YO-28573-08
More Details
Spacer Set for Cole-Parmer Vertical Slab Gel Systems; 28 cm Height, 1. (YO-28573-09)
Category: Cole-Parmer Adjustable Height Single and Dual Vertical Gel Systems
Description: Cole-Parmer Vertical Slab Gel Spacer Set for Height-adjustable units
Brand:
Cole-Parmer
YO-28573-09
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Spacer
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Space-saving Desiccator; Polystyrene; exterior dimensions, 9"W x 9"H x (42053-0001)
|
Next: Spacer Set for Cole-Parmer Dual Vertical Mini-Gel System Model 28575-0 (YO-28575-70)
31125329-p-content_2 31125329pcontent2