Costar Stripwell ™ Plates, High Binding

YO-13020-00
Loading price...

Call for availability.
Qty

  • Plates consist of twelve 1 x 8 strips and an ''egg-crate''-style strip holder.
  • Strips have one-way fit into strip holder—prevents misorientation.
  • Flat-bottom strips break apart and can be reassembled into strip holder.
  • Manufactured from high quality polystyrene (PS)—ideal for uniform binding, high optical clarity, and low background absorption.

High-binding plates have a binding capacity of ~500 ng of Mouse IgG/cm2 and offer the lowest CV available.
Product Type Microplates
Color Clear
Well volume (mL) 0.36
Binding property High
Description Stripwell plates
Qty/cs 100
Material Polystyrene
Brand Costar
Manufacturer number 2592

Accessories
Qty/Price
Availability
Acetate sealing tape
YO-01730-00

CASE of 100

Call for availability.

CASE of 100

Call for availability.

CASE of 50

Call for availability.
Strip holder
YO-13020-50

CASE of 20

Call for availability.
Strip ejector
YO-13020-52

CASE of 5

Call for availability.




Technical Resources
 
Related Products for Costar Stripwell ™ Plates, High Binding
Image
  Product Detail
SKU

Costar Stripwell ™ Plates, Medium Binding

Category: Stripwell Plates
Description: Costar Stripwell Plates
Brand: Costar


Costar Stripwell ™ Plates, 1 x 8 strips

Category: Stripwell Plates
Description: Costar Stripwell Plates
Brand: Costar


Corning 96-Well White Solid Plates, High binding

Category: 96-Well Plates
Description: Corning 96-Well Black and White Solid Plates
Brand: Corning


Corning 96-Well Black Solid Plates, High binding

Category: 96-Well Plates
Description: Corning 96-Well Black and White Solid Plates
Brand: Corning


Strip holder (2572)

Category: Stripwell Plates
Description: Costar Stripwell Plate Stip Holder and Strip Ejector
Brand: Costar


Strip ejector (2578)

Category: Stripwell Plates
Description: Costar Stripwell Plate Stip Holder and Strip Ejector
Brand: Costar


Costar 96-Well Microplates, Immunology, PS, Round, High Binding

Category: Assay Plates
Description: Corning 96-Well Solid Plates
Brand: Corning


Costar 96-Well Microplates, Immunology, PS, Flat, High Binding

Category: Assay Plates
Description: Corning 96-Well Solid Plates
Brand: Corning


Costar 6-well Multiple-well cell culture plates with lid, treated, sterile, 5/pk

Category: Multi-Well Cell Culture Plates
Description: Costar Multi-Well Cell Culture Plates
Brand: Costar


Costar 6-well Multiple-well cell culture plates with lid, treated, sterile, 1/pk

Category: Multi-Well Cell Culture Plates
Description: Costar Multi-Well Cell Culture Plates
Brand: Costar


Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
Questions and Answers
Product Reviews

Costar Stripwell ™ Plates, High Binding
Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended
Popularity |  Favorites |  Title |  Price |  Part Number |  Category
Title |  Brand |  Category |  Price |  Popularity |  Best Sellers

Previous: Costar Stripwell ™ Plates, 1 x 8 strips  |  Next: Costar Stripwell ™ Plates, Medium Binding