Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Standard Methods for the Examination of Water and Wastewater 22nd edition
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-15504-00
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Environmental Books
Share
|
Standard Methods for the Examination of Water and Wastewater, 22nd edi
YO-15504-00
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
More than 340 methods detailing the step-by-step analysis of water and wastewater approved by the EPA. Includes new procedures for laboratory occupational health and safety, and much more.
Specifications
Product Type
Environmental Books
Author
Edited by L. Clesceri et al.
No. of pages
1368
ISBN
9780875530130
Edition
22nd Edition
Published date
2012
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related Searches for Standard Methods for the Examination of Water and Wastewater, 22nd edi
Standard Methods
Water and Wastewater Laboratory Supply Wholesalers
Standard Methods Caseinate Agar
Cole-Parmer Standard Methods Agar
Ph Meter Standards Methods
Environmental Water
Environmental Water Testing Supplies
Laboratories Environmental
Cole-Parmer Environmental Books
Wastewater Laboratory Supplies
Related Products for Standard Methods for the Examination of Water and Wastewater, 22nd edi
Image
Product Detail
SKU
Cole-Parmer Standard methods agar, 500 g jar
Category: Cole-Parmer Industrial Grade Dehydrated Culture Media
Description: Cole-Parmer Standard Methods Agar
Brand:
Cole-Parmer
YO-14200-32
More Details
Whirl-Pak stand-up sodium thiosulfate bags for potable water sampling, (YO-06499-77)
Category: Whirl-Pak Sterile Sampling Bags
Description: Whirl-Pak Sodium Thiosulfate Bags for Water and Wastewater Sampling
Brand:
Whirl-Pak
YO-06499-77
More Details
Whirl-Pak Coli-Test sodium thiosulfate bags for water sampling, 4 oz
Category: Whirl-Pak Sterile Sampling Bags
Description: Whirl-Pak Sodium Thiosulfate Bags for Water and Wastewater Sampling
Brand:
Whirl-Pak
YO-06499-88
More Details
Whirl-Pak sodium thiosulfate bags for potable water sampling, 4 oz
Category: Whirl-Pak Sterile Sampling Bags
Description: Whirl-Pak Sodium Thiosulfate Bags for Water and Wastewater Sampling
Brand:
Whirl-Pak
YO-06499-90
More Details
Whirl-Pak sodium thiosulfate bags for potable water sampling, 18 oz
Category: Whirl-Pak Sterile Sampling Bags
Description: Whirl-Pak Sodium Thiosulfate Bags for Water and Wastewater Sampling
Brand:
Whirl-Pak
YO-06499-95
More Details
Whirl-Pak stand-up sodium thiosulfate bags for potable water sampling, (YO-06499-97)
Category: Whirl-Pak Sterile Sampling Bags
Description: Whirl-Pak Sodium Thiosulfate Bags for Water and Wastewater Sampling
Brand:
Whirl-Pak
YO-06499-97
More Details
Prepared agar plate, Standard Methods agar, 10/pk
Category: Prepared and Contact Media Plates
Description: Prepared Media Plates
Brand:
Cole-Parmer
YO-14201-44
More Details
Prepared media tube, Standard methods agar—prepared tube, 20 pa
Category: Prepared and Contact Media Plates
Description: Prepared Media Tubes
Brand:
Cole-Parmer
YO-14203-92
More Details
Standard methods agar, prepared; 200 mL bottle, 12/pk (YO-14204-88)
Category: Prepared and Contact Media Plates
Description: Prepared Media Bottles
Brand:
Cole-Parmer
YO-14204-88
More Details
Standard Methods Agar 500g
Category: Culture Media
Description: Thermo Scientific Dehydrated Culture Media
Brand:
Remel
YO-14206-16
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Standard Methods for the Examination of Water and Wastewater, 22nd edi
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Standard methods agar, prepared; 200 mL bottle, 12/pk (YO-14204-88)
|
Next: Standard micro tubing valves; 5 ports; one port to inlet; tube OD 1 to
31125329-p-content_2 31125329pcontent2