Home  >  Bottletop Dispensers >
  

30 mL Bottle and glass dropper (15040G-30)

YO-16602-06
Loading price...

Usually ships in 14 days or sooner.
Qty

  • Heat and chemically resistant borosilicate glass
  • Autoclavable; except rubber nipples
Dropper bottles are used in analytical, QC and environmental labs. Amber dropper bottles protect light sensitive chemicals, and are made to ASTM E438 specifications. Replacement droppers both glass and plastic are available below.
Product Type Bottles
Volume (mL) 30
Range (mL) 30
Wetted parts Borosilicate glass
Qty/cs 12
Includes Glass dropper
Autoclavable Yes, except rubber nipples





Technical Resources
 
Related Products for 30 mL Bottle and glass dropper (15040G-30)
Image
  Product Detail
SKU

Kimax Glass Dropping Bottle, 30 mL, 12/Cs

Category: Glass Dropping Bottles
Description: Kimax Glass Dropping Bottles
Brand: Kimble


Fill solution for chlorine dioxide sensors, 30-mL bottle (CLDA-2015)

Category: Cole-Parmer® Free Chlorine and Chlorine Dioxide Sensors
Description: Accessories for Cole-Parmer Free Chlorine and Chlorine Dioxide Sensors


Fill solution for Free Chlorine sensors, 30-mL bottle (FCLA-4015)

Category: Cole-Parmer® Free Chlorine and Chlorine Dioxide Sensors
Description: Accessories for Cole-Parmer Free Chlorine and Chlorine Dioxide Sensors


Thermo Scientific Orion pH electrode cleaning solution A, 4 x 30 mL bottles

Category: pH/Ion Buffers, and Storage Solutions
Description: Thermo Scientific Orion Electrode Cleaning Solutions
Brand: Thermo Scientific Orion


Thermo Scientific Orion pH electrode cleaning solution C, 4 x 30 mL bottles

Category: pH/Ion Buffers, and Storage Solutions
Description: Thermo Scientific Orion Electrode Cleaning Solutions
Brand: Thermo Scientific Orion


Inserts for swing-out bucket rotor; 1 100-mL bottle (YO-02550-74)

Category: Multi-Purpose Universal Centrifuge
Description: Inserts for Multi Purpose Universal Centrifuge
Brand: Cole-Parmer


30 mL Amber Glass Dropping Bottle, Glass dropper

Category: Glass Dropping Bottles
Description: Kimax Amber Glass Dropping Bottles
Brand: Kimble


30 mL Amber Glass Dropping Bottle, Plastic dropper

Category: Glass Dropping Bottles
Description: Kimax Amber Glass Dropping Bottles
Brand: Kimble


Bottle, Precleaned, Clear Glass, Narrow Mouth, Qorpak, 30 mL, 20 mm Neck, PTFE-lined Cap, Case Of 48

Category: Qorpak Sample Jars
Description: Qorpak Clear Narrow-Mouth Glass Bottles Precleaned by Vacuum and Ionization
Brand: Qorpak


Qorpack Precleaned Square Glass Bottle, WM, PTFE Cap; 30 mL, 48/Cs

Category: Qorpak Sample Jars
Description: Qorpak Clear Wide-Mouth Square Glass Bottles Precleaned by Vacuum and Ionization
Brand: Qorpak


Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
Questions and Answers

Do you have questions about this product?

get answers from real customers and in-house experts with AnswerBox.

2 Questions | 2 Answers
Displaying questions 1-2
  • Q:

    ow many bottles in a case?
    Thanks,
    Sandy
    Asked on 11/22/2013 by Sandy from West Midlands

    1 answer

    • STAFF EXPERT

      A:

      There are 12 bottles and droppers per case.

      Answered on 11/22/2013 by Jack from Technical Support
  • Q:

    what are the dimensions of the bottle height and circumference
    Asked on 8/28/2013 by dale from las vegas

    1 answer

    • STAFF EXPERT

      A:

      Approx. Diameter (mm) 31
      Approx. Height (mm) 102
      Dropper Capacity (mL) 1.0

      Answered on 8/28/2013 by Dan from Technical Support
Displaying questions 1-2

Do you have a question about this product? 

Product Reviews

REVIEW SNAPSHOT®

by PowerReviews
30 mL Bottle and glass dropper
 
3.0

(based on 1 review)

Reviewed by 1 customer

Displaying review 1

Back to top

(1 of 1 customers found this review helpful)

 
3.0

great bottles

By seriouslyenoughrequiredfields

from whywouldyouneedthis, MA

About Me Academic/Professional Researcher

Pros

  • Accurate
  • Clear Markings
  • Easy To Use
  • Sturdy

Cons

    Best Uses

    • Chemical Storage
    • In The Lab
    • Research

    Comments about 30 mL Bottle and glass dropper:

    The only drawback to this product is that this company will never stop emailing you for feedback. It is pretty annoying to be honest.

    Displaying review 1

    Back to top


    30 mL Bottle and glass dropper (15040G-30)
    Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended
    Popularity |  Favorites |  Title |  Price |  Part Number |  Category
    Title |  Brand |  Category |  Price |  Popularity |  Best Sellers

    Previous: 30 mL Amber Glass Dropping Bottle, Plastic dropper  |  Next: 30 Piece Deluxe Insulated Tool Kit (TK30)