Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
NIST Traceable Calibration High Accuracy Humidity and Temperature
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
More info
Service & Support
Related media
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
YO-17030-20
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Calibration Services
Share
|
NIST-Traceable Calibration; High Accuracy Humidity and Temperature (YO-17030-20)
YO-17030-20
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
All calibration certificates contain as-found and as-left data , estimated measurement uncertainties (emu’s) and test uncertainty ratios (TUR’s) at no additional cost!
NIST-traceable calibration allows you to confirm that the specific asset you purchase does in fact meet the manufacturer’s published specification
FDA, EPA, USDA, ISO and GLP/GMPs standards require regular NIST-Traceable calibration of all test equipment being used in critical applications
Receive your product calibrated and ready to use rather than sending it to a third-party or internal calibration lab and incurring additional delays
Testing range from 5 to 90% RH at uncertainties as low as
(K=2) ±0.59% RH
Testing at three points: 30, 60, and 80% RH with one temperature point
at ambient (22 to 25C)
All measurements reported are traceable to SI units via national standards maintained by NIST and performed in compliance with ANSI Z540, ISO17025 and MIL-STD-45662A. See the sample certificate under the “More Info” section to see how InnoCal’s certificates provide more information in a concise manner than the rest of the industry.
InnoCal is accredited to ISO/IEC 17025 by the American Association for Laboratory Accreditation (A2LA) under certificate number 1746.01. This accreditation is your assurance that the calibration is performed in a qualified, controlled laboratory which is technically proficient and committed to quality and customer-focused continuous improvement. Our scope of A2LA accreditation can be found under the “More Info” section.
Our highly-trained technicians provide NIST-traceable accredited humidity
calibrations using Condensation Chilled Mirror Hygrometer technology in
a dedicated environmental chamber.
Specifications
Product Type
Humidity Calibrations
Calibration points
Three humidity test points (30, 60, and 80% RH) and one temperature test point at ambient (22 to 25°C)
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
13-14 International Catalog
More info
InnoCal's A2LA Scope of Accreditation
An interview with ACLASS Accreditation Services
See an example of InnoCal's NIST-traceable calibration report
Why calibrate test equipment
Related Searches for NIST-Traceable Calibration; High Accuracy Humidity and Temperature (YO-17030-20)
Cole-Parmer Timers with Nist-Traceable Calibration Report
Nist Traceable Calibration
Nist-Traceable Calibrators/Simulators
Dissolved Oxygen Nist-Traceable Calibration
Nist Traceable Sieve Calibration Glass Beads
Nist Traceable Pressure Gauges
Cole-Parmer Traceable High-Accuracy Conductivity Meters
Nist Traceable Temperature
Nist Traceable Digital Thermometer
Ashcroft High-Accuracy Digital Test Gauge
Related Products for NIST-Traceable Calibration; High Accuracy Humidity and Temperature (YO-17030-20)
Image
Product Detail
SKU
NIST-Traceable Calibration; Standard Accuracy Humidity and Temperature (YO-17030-28)
Category: Calibration Services
Description: Humidity Calibrations
Brand:
InnoCal
YO-17030-28
More Details
Cole-Parmer Sieve with NIST-Traceable Calibration Report, Brass Frame/ (YO-59980-50)
Category: 8" Diameter Testing Sieves with Certificate to NIST-Traceable Standards
Description: Cole-Parmer Sieves with NIST-Traceable Calibration Report, 8" Brass Frame/Brass Wire
Brand:
Cole-Parmer
YO-59980-50
More Details
Cole-Parmer Sieve with NIST-Traceable Calibration Report, Brass Frame/ (YO-59980-51)
Category: 8" Diameter Testing Sieves with Certificate to NIST-Traceable Standards
Description: Cole-Parmer Sieves with NIST-Traceable Calibration Report, 8" Brass Frame/Brass Wire
Brand:
Cole-Parmer
YO-59980-51
More Details
Cole-Parmer Sieve with NIST-Traceable Calibration Report, Brass Frame/ (YO-59980-52)
Category: 8" Diameter Testing Sieves with Certificate to NIST-Traceable Standards
Description: Cole-Parmer Sieves with NIST-Traceable Calibration Report, 8" Brass Frame/Brass Wire
Brand:
Cole-Parmer
YO-59980-52
More Details
Cole-Parmer Sieve with NIST-Traceable Calibration Report, Brass Frame/ (YO-59980-53)
Category: 8" Diameter Testing Sieves with Certificate to NIST-Traceable Standards
Description: Cole-Parmer Sieves with NIST-Traceable Calibration Report, 8" Brass Frame/Brass Wire
Brand:
Cole-Parmer
YO-59980-53
More Details
Cole-Parmer Sieve with NIST-Traceable Calibration Report, Brass Frame/ (YO-59980-54)
Category: 8" Diameter Testing Sieves with Certificate to NIST-Traceable Standards
Description: Cole-Parmer Sieves with NIST-Traceable Calibration Report, 8" Brass Frame/Brass Wire
Brand:
Cole-Parmer
YO-59980-54
More Details
Cole-Parmer Sieve with NIST-Traceable Calibration Report, Brass Frame/ (YO-59980-55)
Category: 8" Diameter Testing Sieves with Certificate to NIST-Traceable Standards
Description: Cole-Parmer Sieves with NIST-Traceable Calibration Report, 8" Brass Frame/Brass Wire
Brand:
Cole-Parmer
YO-59980-55
More Details
Cole-Parmer Sieve with NIST-Traceable Calibration Report, Brass Frame/ (YO-59980-56)
Category: 8" Diameter Testing Sieves with Certificate to NIST-Traceable Standards
Description: Cole-Parmer Sieves with NIST-Traceable Calibration Report, 8" Brass Frame/Brass Wire
Brand:
Cole-Parmer
YO-59980-56
More Details
Cole-Parmer Sieve with NIST-Traceable Calibration Report, Brass Frame/ (YO-59980-57)
Category: 8" Diameter Testing Sieves with Certificate to NIST-Traceable Standards
Description: Cole-Parmer Sieves with NIST-Traceable Calibration Report, 8" Brass Frame/Brass Wire
Brand:
Cole-Parmer
YO-59980-57
More Details
Cole-Parmer Sieve with NIST-Traceable Calibration Report, Brass Frame/ (YO-59980-58)
Category: 8" Diameter Testing Sieves with Certificate to NIST-Traceable Standards
Description: Cole-Parmer Sieves with NIST-Traceable Calibration Report, 8" Brass Frame/Brass Wire
Brand:
Cole-Parmer
YO-59980-58
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Related media
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
NIST-Traceable Calibration; High Accuracy Humidity and Temperature (YO-17030-20)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: NIST-Traceable Calibration; Height Gauge, 0-24" (YO-17050-14)
|
Next: NIST-Traceable Calibration; High-Accuracy Loop Calibrators (YO-17090-22)
31125329-p-content_2 31125329pcontent2