Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
YO-17425-10
Gerber-method Butyrometer; milk test
Click to enlarge
More Product Details
Specifications
Accessories
Build a complete system
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-17425-10
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Gerber-Method Fat Analysis System
Share
|
Gerber-method Butyrometer; milk test (YO-17425-10)
YO-17425-10
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Get test results in 15 minutessaves you time and money
Use these butyrometers to measure the percent fat in dairy products. Butyrometers are precision calibrated and feature permanent graduations. Made of resistant borosilicate glass.
Order glass cups for Roeder and Van-Gulik method testing. Stoppers with special acid resistant rubber are available for all butyrometers. Both stopper 17425-15 and stopper pusher 17425-16 are required for milk and Kohler method testing; stoppers 17425-14 and -17 are required for Roeder and Van-Gulik method testing.
Specifications
Product Type
Extraction bottles
Material
Glass
Capacity (Liters)
0.011
Capacity (mL)
11
Accessories
Qty
/
Price
Availability
Stopper For Butyrometers 17425-10 And -11. Order Stopper Pusher 17425-16. Box Of 100.
YO-17425-15
BOX of 100
Call for availability.
Add to Cart
Stopper Pusher For Stopper 17425-15. Box Of Five.
YO-17425-16
BOX of 5
Call for availability.
Add to Cart
Build a complete system
1
.
Butyrometers for Gerber-Method Fat Analysis
Details
2
.
Pipette for Gerber-Method Fat Analysis
Details
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
13-14 International Catalog
Related technical articles (Roll over for summary)
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Related Searches for Gerber-method Butyrometer; milk test (YO-17425-10)
Cole-Parmer Butyrometers for Gerber-Method Fat Analysis
Cole-Parmer Gerber-Method Fat Analysis System
Hydrogen Peroxide Milk Test
Testing Milk
Gerber Test
Mojonnier Fat Analysis Method Application
Fat Extraction System
Fat Analysis
Fat Extraction
Equipment Fat Analysis
Related Products for Gerber-method Butyrometer; milk test (YO-17425-10)
Image
Product Detail
SKU
Gerber-method Butyrometer; cheese test (Van-Gulik) (YO-17425-13)
Category: Gerber-Method Fat Analysis System
Description: Butyrometers for Gerber-Method Fat Analysis
Brand:
Cole-Parmer
YO-17425-13
More Details
E. coli Swabcheck kit, 125 tests (YO-14019-10)
Category: Bacteria test kits
Description:
Escherichia coli
SwabCheck Kits
Brand:
Cole-Parmer
YO-14019-10
More Details
Babcock bottle, skim milk test (YO-34590-01)
Category: Babcock System for Fat Analysis
Description: Babcock and Paley Bottles
Brand:
Cole-Parmer
YO-34590-01
More Details
Babcock bottle, milk test (YO-34590-06)
Category: Babcock System for Fat Analysis
Description: Babcock and Paley Bottles
Brand:
Cole-Parmer
YO-34590-06
More Details
Pipette for Gerber test, 11-mL capacity. Case of 12. (YO-34590-63)
Category: Gerber-Method Fat Analysis System
Description: Pipette for Gerber-Method Fat Analysis
Brand:
Cole-Parmer
YO-34590-63
More Details
Cole-Parmer Testing Sieve, Brass Frame/SS Wire, 8" OD, full height, No (YO-59940-10)
Category: 8" Diameter Testing Sieves
Description: Cole-Parmer Testing Sieves, Brass Frame/Stainless Steel Wire
Brand:
Cole-Parmer
YO-59940-10
More Details
Cole-Parmer Testing Sieve, Brass Frame/SS Wire, 8" OD, half height, No (YO-59941-10)
Category: 8" Diameter Testing Sieves
Description: Cole-Parmer Testing Sieves, Brass Frame/Stainless Steel Wire
Brand:
Cole-Parmer
YO-59941-10
More Details
Cole-Parmer Testing Sieve, 8" OD Brass Frame/Brass Wire, Full height, (YO-59980-10)
Category: 8" Diameter Testing Sieves
Description: Cole-Parmer Testing Sieves, 8" Brass Frame/Brass Wire
Brand:
Cole-Parmer
YO-59980-10
More Details
Cole-Parmer Testing Sieve, 8" OD Brass Frame/Brass Wire, Half height, (YO-59981-10)
Category: 8" Diameter Testing Sieves
Description: Cole-Parmer Testing Sieves, 8" Brass Frame/Brass Wire
Brand:
Cole-Parmer
YO-59981-10
More Details
Cole-Parmer Testing Sieve, Brass Frame/Brass Wire, 12" OD, Full height (YO-59990-10)
Category: 12" Diameter Testing Sieves
Description: Cole-Parmer Testing Sieves, 12" Brass Frame/Brass Wire
Brand:
Cole-Parmer
YO-59990-10
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Gerber-method Butyrometer; milk test (YO-17425-10)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Gerber-method Butyrometer; cheese test (Van-Gulik) (YO-17425-13)
|
Next: Germanium Plate for iD5 ATR for the Nicolet iS5 FT-IR Spectrometer
31125329-p-content_2 31125329pcontent2