Home  >  Cleaning Detergents >
  

Liqui-Nox Cleaning Detergent, one gallon bottle

YO-17778-40
Loading price...

Usually ships in 14 days or sooner.
Qty

  • Order new bulk packages for added convenience
White powder Alconox®, Alcojet®, and Terg-A-Zyme® cleaners rinses off completely—with no residue. Alconox® cleaner is biodegradable; Alcojet® cleaner is low-sudsing; Terg-A-Zyme® cleaner combines the original Alconox® power with protein enzymes. Liqui-Nox® cleaner is biodegradable—use to remove soil, grease, blood, chemicals, and solvents. All cleaner are safe, odorless, and FDA compliant.
ALCOJET, ALCONOX, LIQUI-NOX, TERG-A-ZYME-Reg TM Alconox Inc.
Product Type Equipment Cleaner
Format Liquid
Volume (gal.) 1
Packaging 1 Gallon
Uses Manual or ultrasonic cleaning
Brand Liqui-Nox
Model 1201-1
Manufacturer number 1201-1





Technical Resources
 
Related Products for Liqui-Nox Cleaning Detergent, one gallon bottle
Image
  Product Detail
SKU

Citranox Cleaning Detergent, one-gallon bottle

Category: Cleaning Detergents
Description: Alconox Cleaning Detergents
Brand: Citranox


Luminox Cleaning Detergent, one-gallon bottle

Category: Cleaning Detergents
Description: Alconox Cleaning Detergents
Brand: Luminox


Liqui-Nox Cleaning Detergent, case of four 1-gallon bottles

Category: Cleaning Detergents
Description: Alconox Cleaning Detergents
Brand: Liqui-Nox


Liqui-Nox cleaner for general purpose cleaning, 1 quart bottle, case of 12 bottles

Category: Cleaning Detergents
Description: Alconox Cleaning Detergents
Brand: Liqui-Nox


Liqui-Nox cleaner for general purpose cleaning, 1 quart bottle

Category: Cleaning Detergents
Description: Alconox Cleaning Detergents
Brand: Liqui-Nox


Detergent 8 Cleaning Detergent, case of four 1-gallon bottles

Category: Cleaning Detergents
Description: Alconox Cleaning Detergents
Brand: Detergent 8


Cole-Parmer Micro-90 cleaning solution, 5 gallon bottle

Category: Surface Cleaners
Description: Cole-Parmer Micro-90 Cleaning Solution
Brand: Micro-90


Industrial strength ultrasonic cleaning solution, 5 gallon bottle (000-955-107)

Category: Ultrasonic Cleaning Solutions
Description: Industrial strength cleaning solution
Brand: Branson


Citranox Cleaning Detergent, case of four 1-gallon bottles

Category: Cleaning Detergents
Description: Alconox Cleaning Detergents
Brand: Citranox


I-CHEM Certified Pre-Cleaned Wide-Mouth Plastic Bottles, 125 mL

Category: Environmental Bottles
Description: I-CHEM Certified Pre-Cleaned Wide-Mouth Plastic Bottles
Brand: I-Chem


Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
Questions and Answers

Do you have questions about this product?

get answers from real customers and in-house experts with AnswerBox.

1 Question | 1 Answer
Displaying question 1
  • Q:

    what is the difference between alconox nd liqui-nox
    Asked on 8/10/2011 by Anonymous

    1 answer

    • Staff Reviewer

      A:

      Alconox is the manufacturer, Liqui-Nox is one of their products.

      Answered on 8/12/2011 by Jen from Technical Support
Displaying question 1

Do you have a question about this product? 

Product Reviews
Customers Who Bought This Item Also Bought