Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Cole Parmer Zymit enzyme cleaner 1 liter bottle case of twelve
Click to enlarge
More Product Details
Specifications
Technical Resources
MSDS
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-18101-20
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Ultrasonic Cleaning Solutions
Share
|
Cole-Parmer Zymit™ enzyme cleaner, 1 liter bottle, case of twelve
YO-18101-20
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Ideal for use in ultrasonic cleaners and glassware washers
This enzyme-based liquid cleaner removes biological materials from chemical, food, and pharmaceutical equipment. Cleaner has pH of 7.5 allowing you to use it with delicate materials and metals. Solution is USDA approved.Zymit Low-foam is a liquid dual-enzyme and detergent formula for removal of protein and starch based soils.
Zymit Low-foam is used to clean diagnostic and precision instruments, labware, photographic film equipment, and daycare furnishings and many other surfaces. Target soils include food, grass, fat, sweat, mucous, tissue, feces and sebum. Solution is safe for workers and the environment - contains no phosphates, CFCs, ODCs, phenols or borates.
Designed for use in ultrasonic baths, automatic washers and high-agitation applications. A 1% to 2% solution is effective in most applications. Typical pH: 7.5. NSF-registered for USDA A1 use.
Specifications
Product Type
Equipment Cleaner
Description
Case of 12 1-L bottles
Format
Liquid
Volume (Liters)
1
Packaging
1 Liter
Uses
Manual or ultrasonic; dissolves protein and starch-based soils; may use for delicate materials
Brand
Zymit
Manufacturer number
Z-9701
Technical Resources
MSDS 1 (135 B)
Related technical articles (Roll over for summary)
ChemicalsSpecial Ordering Information
Show Less...
Related Searches for Cole-Parmer Zymit™ enzyme cleaner, 1 liter bottle, case of twelve
Cole-Parmer Zymit Enzyme Cleaner
Cole-Parmer Ultrasonic Jewelry Cleaners
Cole-Parmer Compact Ultrasonic Cleaners
Cole-Parmer Wheaton Safety-Coated Narrow-Mouth Bottles
1 Liter Bottles
Cole-Parmer Standard Ultrasonic Cleaners
Cole-Parmer Test Tube Rack
Cole-Parmer Conductivity/Tds/Temperature 1/4-Din Controllers
Cole-Parmer Ph/Orp/Temperature 1/8-Din Controller and Transmitter
Cole-Parmer Conductivity/Resistivity/Temperature 1/8-Din Transmitter and Controller
Related Products for Cole-Parmer Zymit™ enzyme cleaner, 1 liter bottle, case of twelve
Image
Product Detail
SKU
Cole-Parmer Zymit™ enzyme cleaner, 1 liter bottle
Category: Ultrasonic Cleaning Solutions
Description: Cole-Parmer Zymit Enzyme Cleaner
Brand:
Zymit
YO-18101-30
More Details
Cole-Parmer 300 pH/ORP/Temperature 1/8-DIN Transmitter
Category: 1/8-DIN pH/ORP Control
Description: Cole-Parmer pH/ORP/Temperature 1/8-DIN Controller and Transmitter
Brand:
Cole-Parmer
Model Number:
300PH
YO-05802-12
More Details
Cole-Parmer 350 pH/ORP/Temperature 1/8-DIN Controller
Category: 1/8-DIN pH/ORP Control
Description: Cole-Parmer pH/ORP/Temperature 1/8-DIN Controller and Transmitter
Brand:
Cole-Parmer
Model Number:
350PH
YO-05802-14
More Details
Cole-Parmer 500 pH/Temperature 1/4-DIN Controller
Category: 1/4-DIN pH/ORP Control
Description: Cole-Parmer pH/Temperature and pH/ORP/Temperature 1/4-DIN Controllers
Brand:
Cole-Parmer
Model Number:
500 PH
YO-05802-16
More Details
Cole-Parmer 550 pH/Temperature 1/4-DIN Controller
Category: 1/4-DIN pH/ORP Control
Description: Cole-Parmer pH/Temperature and pH/ORP/Temperature 1/4-DIN Controllers
Brand:
Cole-Parmer
Model Number:
500 PH
YO-05802-18
More Details
Cole-Parmer 500 Cond/TDS/Temperature 1/4-DIN Controller
Category: 1/4-DIN Conductivity Control
Description: Cole-Parmer Conductivity/TDS/Temperature 1/4-DIN Controllers
Brand:
Cole-Parmer
Model Number:
Cond 500
YO-05802-06
More Details
Cole-Parmer 550 Cond/TDS/Temperature 1/4 DIN Controller
Category: 1/4-DIN Conductivity Control
Description: Cole-Parmer Conductivity/TDS/Temperature 1/4-DIN Controllers
Brand:
Cole-Parmer
Model Number:
Cond 550
YO-05802-08
More Details
Cole-Parmer 300 Conductivity/Resistivity/Temperature 1/8-DIN Transmitt
Category: 1/8-DIN Conductivity Control
Description: Cole-Parmer Conductivity/Resistivity/Temperature 1/8-DIN Transmitter and Controller
Brand:
Cole-Parmer
Model Number:
300 Cond
YO-05802-02
More Details
Cole-Parmer 350 Conductivity/Resistivity/Temperature 1/8- DIN Controll
Category: 1/8-DIN Conductivity Control
Description: Cole-Parmer Conductivity/Resistivity/Temperature 1/8-DIN Transmitter and Controller
Brand:
Cole-Parmer
Model Number:
350 Cond
YO-05802-04
More Details
Cole-Parmer 500 DO (Boiler) 1/4-DIN Controller
Category: 1/4-DIN Dissolved Oxygen Control
Description: Cole-Parmer 1/4-DIN Dissolved Oxygen Controllers
Brand:
Cole-Parmer
YO-05808-12
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Cole-Parmer Zymit™ enzyme cleaner, 1 liter bottle, case of twelve
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Cole-Parmer Zymit™ enzyme cleaner, 1 liter bottle
|
Next: Cole-Parmer, Barbed Sanitary Clamp Adapter, 304L SS, 1.5" Clamp X 1.5"
31125329-p-content_2 31125329pcontent2