Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, Fittings, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Diaphragm Sensor 1 1500 Torr 1 2 O d
Click to enlarge
More Product Details
Specifications
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-68808-62
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
YO-17040-12
NIST-Traceable Calibration; Pressure / Vacuum (For gauges with accuracy 0.75% or < 0.75%)
$260.61 AUD/EACH/EACH
Usually ships in 20 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Vacuum Controllers, Gauges, and Systems
Share
|
Diaphragm Sensor, 1-1500 Torr, 1/2" O.d.
YO-68808-62
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Use with Hastings
®
DV-6 or VRC gauge tubes
Ranges from 10
-5 Torr to 2 atmospheres
Analog output for PLC or computer interface
These active vacuum switch/transmitters provide an inexpensive and straightforward way to control your vacuum system. Use the Pirani sensor switch directly to initiate crossover after roughing, or the diaphragm sensor switch to control backfill pressure on pump and purge operations. Feed the switch output into your PC or PLC and let the digital system make the control decisions based on the output signal.To create a complete vacuum system, choose your vacuum range and sensor type from the table below, and then choose your sensor connection style. Switch/transmitter plugs directly into sensors. Measure high, medium, and low vacuum with Pirani, thermocouple, and diaphragm sensors.
Specifications
Product Type
Vacuum controllers
Wetted materials
300 SS
Process connection
1/2" OD
Manufacturer number
912210
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Pressure / Vacuum (For gauges with accuracy 0.75% or < 0.75%)
YO-17040-12
EACH
Usually ships in 20 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
11-12 Catalog
11-12 International Catalog
13-14 Catalog
13-14 International Catalog
Related technical articles (Roll over for summary)
Absolute, Gauge, and Differential Pressure Scales
Common Pressure Terms
Pressure Unit Conversion Constants
Show All...
The Hidden Costs of Mercury-Filled Vacuum Gauges
Show Less...
Related Searches for Diaphragm Sensor, 1-1500 Torr, 1/2" O.d.
Id 1 2 Od Surgical Tubing
Diaphragm Sensors
2 Od Y Fitting
2 Od 16 Ga Ss Tubing
2 Od Pipe Fittings
Rubber Tubing 1 2 Od
1 Hose to 1 Flange Fitting
1 Nptm X 1 Brass Barb
Diaphragm Pressure Switches
Pressure Sensor Transmitter
Related Products for Diaphragm Sensor, 1-1500 Torr, 1/2" O.d.
Image
Product Detail
SKU
1 to 1500 torr Cole-Parmer Pressure/Vacuum Diaphragm-Type Sensor, NW 1
Category: Cole-Parmer Pressure / Vacuum Systems
Description: Cole-Parmer Pressure/Vacuum Systems with silicon diaphragm sensor
Brand:
Cole-Parmer
YO-68801-47
More Details
1 to 1500 torr Cole-Parmer Pressure/Vacuum Diaphragm-Type Sensor, 1/8"
Category: Cole-Parmer Pressure / Vacuum Systems
Description: Cole-Parmer Pressure/Vacuum Systems with silicon diaphragm sensor
Brand:
Cole-Parmer
YO-68801-57
More Details
1 to 1500 torr Cole-Parmer Pressure/Vacuum Meter for Diaphragm-Type Se
Category: Cole-Parmer Pressure / Vacuum Systems
Description: Cole-Parmer Pressure/Vacuum Systems with 316 SS diaphragm sensor
Brand:
Cole-Parmer
YO-68801-03
More Details
1 to 1500 torr Cole-Parmer Pressure/Vacuum Controller for Diaphragm-Ty
Category: Cole-Parmer Pressure / Vacuum Systems
Description: Cole-Parmer Pressure/Vacuum Systems with 316 SS diaphragm sensor
Brand:
Cole-Parmer
YO-68801-23
More Details
1 To 1500 Torr Cole-parmer Pressure/vacuum Pirani-type Sensor, Nw 16 C
Category: Cole-Parmer Pressure / Vacuum Systems
Description: Cole-Parmer Pressure/Vacuum Systems with 316 SS diaphragm sensor
Brand:
Cole-Parmer
YO-68801-43
More Details
1 to 1500 torr Cole-Parmer Pressure/Vacuum Pirani-Type Sensor, 1/4" NP
Category: Cole-Parmer Pressure / Vacuum Systems
Description: Cole-Parmer Pressure/Vacuum Systems with 316 SS diaphragm sensor
Brand:
Cole-Parmer
YO-68801-53
More Details
1 to 1500 torr Cole-Parmer Pressure/Vacuum Calibrator for Diaphragm-Ty
Category: Cole-Parmer Pressure / Vacuum Systems
Description: Cole-Parmer Pressure/Vacuum Systems with 316 SS diaphragm sensor
Brand:
Cole-Parmer
YO-68801-63
More Details
Diaphragm sensors, 1/8" NPT(M)
Category: Vacuum Controllers, Gauges, and Systems
Description: Vacuum Research Pirani and Diaphragm Integral Sensor Gauges
Brand:
Vacuum Research
YO-68808-42
More Details
30" Hg To 100 PSI Cole-parmer Pressure/vacuum Diaphragm-type Sensor, 1
Category: Cole-Parmer Pressure / Vacuum Systems
Description: Cole-Parmer Pressure/Vacuum Systems with silicon diaphragm sensor
Brand:
Cole-Parmer
YO-68801-54
More Details
0 to 200 psi Cole-Parmer Pressure/Vacuum Diaphragm-Type Sensor, 1/4" N
Category: Cole-Parmer Pressure / Vacuum Systems
Description: Cole-Parmer Pressure/Vacuum Systems with silicon diaphragm sensor
Brand:
Cole-Parmer
YO-68801-55
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Diaphragm Sensor, 1-1500 Torr, 1/2" O.d.
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Diamond Plate for iD5 ATR for the Nicolet iS5 FT-IR Spectrometer
|
Next: Diaphragm Sensor, 30" Hg To Atm, 1/4" NPT (m)
31125329-p-content_2 31125329pcontent2