Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
YO-24908-00
Hach digital titrator
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
YO-24908-00
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Digital Burette
Share
|
Hach digital titrator (16900-01)
YO-24908-00
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Faster and more accurate than a conventional burette
Portable, precision dispensing with automatic calculation of titrant - works in field or lab settings
Interchangeable, sealed cartridges with concentrated titrants save time and eliminate glassware cleanup
The Hach digital titrator produces repeatable results through the use of concentrated titrants packaged in compact interchangeable cartridges. To titrate, simply attach a cartridge to the titrator and insert a delivery tube. Each turn of the delivery knob dispenses a calculated amount of titrant. The numerical counter records the volume. When the end point is reached, calculate the sample concentration using the number displayed on the counter. Any remaining reagent may be capped and stored for future use.
What's Included:
plastic case and five straight delivery tubes. Order interchangeable titrator cartridges separately.
Specifications
Product Type
Digital Titrators
Dispensing accuracy
800 digits/mL or 0.00125 mL/digit
Accuracy
±1%
CE Compliance
Yes
Brand
Hach
Manufacturer number
16900-01
Model
16900-01
Accessories
Qty
/
Price
Availability
Delivery tubes, straight with J-hook, 5/PK
YO-24908-50
PACKAGE
Call for availability.
Add to Cart
Delivery tubes, 90 degree with J-hook, 5/PK
YO-24908-55
PACKAGE
Call for availability.
Add to Cart
Acid-Base titrator cartridge with range of 1 to 10 meq/L Acid. Titrant is 1.6N NaOH
YO-24908-60
EACH
Call for availability.
Add to Cart
Alkalinity titrator cartridge with range of 10 to 160 mg/L CaCO3 and titrant of 0.1600N H2SO4
YO-24908-64
EACH
Call for availability.
Add to Cart
Alkalinity titrator cartridge with range of 10 to 4000 mg/L CaCO3 and titrant of1.600N H2SO4
YO-24908-66
EACH
Call for availability.
Add to Cart
Chloride titrator cartridge with range of 2.5 to 150 g/L C-; 100 to 10,000 mg/L C- and titrant of 1.128N AgNO3
YO-24908-68
EACH
Call for availability.
Add to Cart
Chloride titrator cartridge with range of 10 to 100 mg/L C- and titrant of 0.2256N AgNO3
YO-24908-72
EACH
Call for availability.
Add to Cart
Hardness, Calcium titrator cartridge with range 10 to 160 mg/L CaCO3 of and titrant of 0.0800M EDTA
YO-24908-76
EACH
Call for availability.
Add to Cart
Hardness, Calcium titrator cartridge with range 100 to 4000 mg/L CaCO3 of and titrant of 0.800M EDTA
YO-24908-78
EACH
Call for availability.
Add to Cart
Technical Resources
24908-00.pdf
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Taking the “Dry” Out of Moisture Determination
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Related Searches for Hach digital titrator (16900-01)
Hach Digital Titrator
Hach Lab Equipment
Hach Titration Cartridges
Digital Scales 01
Digital Hot Plate Hp50 01
Cooper Temperature Digital Thermometers
Fischer Titrators
Digital Torque Screwdriver
Digital Torque Wrench Tester
Hardness Titrators
Related Products for Hach digital titrator (16900-01)
Image
Product Detail
SKU
Acid-Base titrator cartridge with range of 1 to 10 meq/L Acid. Titran (14379-01)
Category: Digital Burette
Description: Hach Titration Cartridges
Brand:
Hach
Model Number:
14379-01
YO-24908-60
More Details
Alkalinity titrator cartridge with range of 10 to 160 mg/L CaCO3 and t (14388-01)
Category: Digital Burette
Description: Hach Titration Cartridges
Brand:
Hach
Model Number:
14388-01
YO-24908-64
More Details
Alkalinity titrator cartridge with range of 10 to 4000 mg/L CaCO3 and (14389-01)
Category: Digital Burette
Description: Hach Titration Cartridges
Brand:
Hach
Model Number:
14389-01
YO-24908-66
More Details
Chloride titrator cartridge with range of 2.5 to 150 g/L C-; 100 to 10 (14397-01)
Category: Digital Burette
Description: Hach Titration Cartridges
Brand:
Hach
Model Number:
14397-01
YO-24908-68
More Details
Chloride titrator cartridge with range of 10 to 100 mg/L C- and titra (14396-01)
Category: Digital Burette
Description: Hach Titration Cartridges
Brand:
Hach
Model Number:
14396-01
YO-24908-72
More Details
Hardness, Calcium titrator cartridge with range 10 to 160 mg/L CaCO3 o (14364-01)
Category: Digital Burette
Description: Hach Titration Cartridges
Brand:
Hach
Model Number:
14364-01
YO-24908-76
More Details
Hardness, Calcium titrator cartridge with range 100 to 4000 mg/L CaCO3 (14399-01)
Category: Digital Burette
Description: Hach Titration Cartridges
Brand:
Hach
Model Number:
14399-01
YO-24908-78
More Details
Hirschmann opus Titrator, 0.1 to 200 mL, 120 VAC
Category: Digital Burette
Description: Hirschmann opus Titrator
Brand:
Hirschmann
Model Number:
Opus
YO-24910-00
More Details
Hach StablCal 0.1 NTU Turbidity Standard, 500-mL
Category: Turbidity Standards and Calibration Sets
Description: Hach StablCal Turbidity Standard Solutions
Brand:
Hach
Model Number:
26597-49
YO-99512-54
More Details
Digital mass Flowmeter, Aluminum, For Air, 0 To 10 mL/min (XFM17A-VADL9A5-01-AIR/N2)
Category: Thermal Dispersion / Gas Mass Flowmeters
Description: Digital Gas Mass Flowmeter
Brand:
Cole-Parmer
Model Number:
XFM17A-VADL9A5-01-AIR/N2
YO-33400-01
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Hach digital titrator (16900-01)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Hach Cyanide Test Kit (24302-00)
|
Next: Hach Dissolved Oxygen Test Kit (25150-25)
31125329-p-content_2 31125329pcontent2