Safetypette pipette filler (475-165)

YO-25200-30
Loading price...

Usually ships in 14 days or sooner.
Qty

  • Egg-shaped bulb provides a comfortable grip and fast suction for both right- and left-handed users
  • Powerful suction quickly draws fluids
  • Filter protects bulb from damage by caustic liquids and filters air to prevent cross contamination of pipettes.
Uniquely designed, lightweight pipette filler fits comfortably in either hand enabling smooth, variable speed control when aspirating or dispensing. Simple thumb-button operation provides precise control. Safetypette works with any pipette between 1 mL and 100 mL, and is compatible with both blow-out and to-deliver pipettes. Autoclavable.
WhatÕs included: integral 0.45 µm filter, silicone pipette holder, and three silicone colored pipette cones for safety color coding.
Product Type Pipetting Accessory
Color three silicone colored pipette cones
Brand Cole-Parmer
Manufacturer number 475-165

Accessories
Qty/Price
Availability

PACKAGE

Usually ships in 14 days or sooner.

EACH

Usually ships in 14 days.




Technical Resources
 
Related Products for Safetypette pipette filler (475-165)
Image
  Product Detail
SKU

Jencons Smoothie Pipette Filler, Vanilla (475-360)

Category: Pipette Fillers
Description: Jencons Smoothie Pipette Fillers
Brand: Jencons Smoothie


Jencons Smoothie Pipette Filler, Strawberry (475-361)

Category: Pipette Fillers
Description: Jencons Smoothie Pipette Fillers
Brand: Jencons Smoothie


Jencons Smoothie Pipette Filler, Banana (475-362)

Category: Pipette Fillers
Description: Jencons Smoothie Pipette Fillers
Brand: Jencons Smoothie


Jencons Smoothie Pipette Filler, Lemon Lime (475-363)

Category: Pipette Fillers
Description: Jencons Smoothie Pipette Fillers
Brand: Jencons Smoothie


Jencons Smoothie Pipette Filler, Blueberry (475-364)

Category: Pipette Fillers
Description: Jencons Smoothie Pipette Fillers
Brand: Jencons Smoothie


Safety Pipette Filler bulb with removable Valve (FLIP-001)

Category: Pipette Fillers
Description: Safety Pipette Filler with Removable Valve
Brand: Cole-Parmer


50.0 mL Silicone pipette filler (SILRED-001)

Category: Pipette Fillers
Description: Silicone Pipette Filler
Brand: Cole-Parmer


Cole-Parmer Pipette Controller, filler, fits standard pipettes; 50 mL capacity

Category: Pipette Fillers
Description: Cole-Parmer Pipette Filler
Brand: Cole-Parmer


Pipette Pumps, yellow, 0.2 mL (37896)

Category: Pipette Fillers
Description: Pipette Pumps
Brand: Scienceware


Pipette Pumps, blue, 2 mL (37897)

Category: Pipette Fillers
Description: Pipette Pumps
Brand: Scienceware


Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
Questions and Answers
Product Reviews

REVIEW SNAPSHOT®

by PowerReviews
Cole-ParmerSafetypette pipette filler
 
3.0

(based on 2 reviews)

Reviewed by 2 customers

Sort by

Displaying reviews 1-2

Back to top

 
5.0

Pipette filler

By JC

from Malibu, Ca

About Me Academic/Professional Researcher

Verified Reviewer

Pros

  • Accurate
  • Clear Markings
  • Easy To Use
  • Sturdy

Cons

    Best Uses

    • In The Lab

    Comments about Cole-Parmer Safetypette pipette filler:

    I use the pipette for various titrations that do not require a micropipette

     
    1.0

    sucks or does it

    By haha

    from Denver, CO

    About Me Lab Assistant

    Pros

    • Easy To Use

    Cons

    • Breaks Easily
    • Inaccurate

    Best Uses

    • In The Lab

    Comments about Cole-Parmer Safetypette pipette filler:

    breaks really easily.

    Displaying reviews 1-2

    Back to top

    Customers Who Bought This Item Also Bought