Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Zippette Dispenser 0 2 5 mL
Click to enlarge
More Product Details
Specifications
Calibrations
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-25204-10
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
YO-17104-00
NIST-Traceable Calibration; Pipettors, with 10 measurements at 3 volumes
$122.86 AUD/EACH/EACH
Usually ships in 23 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Bottletop Dispensers
Share
|
Zippette Dispenser, 0-2.5 mL (263-000)
YO-25204-10
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
These ZIPPETTE dispensers are simple and durable enough to use in the lab, the field, or the classroom.
Redesigned twist-to-lock volume adjuster stays secure to ensure accuracy. To dispense, simply set volume, lock in adjuster, then lift piston and push down. A delivery tube of PTFE lets you safely fill handheld narrow-neck containers. Piston assembly is easy to disassemble for cleaning.
All models have a polypropylene-encased body, borosilicate glass syringe, chemical-resistant plunger, and a safety valve system contained within a PVDF pedestal that holds a primeprevents reagent loss during re-priming. Wetted parts are borosilicate glass and PTFE. Each dispenser fits bottles with 30-mm necks and includes one 300-mm inlet feed tube and adapters for 38-, 40-, and 45-mm bottle necks. Order bottles separately.
DIGITRATE,ZIPPETTE—Reg TM Jencons Ltd.
Specifications
Product Type
Dispenser (Bottletop)
Volume (mL)
2.5
Accuracy
±0.3%
Division (mL)
0.05
Range (mL)
.05 - 2.5
Reproducibility
±0.1%
Wetted parts
Borosilicate glass and PTFE
Autoclavable
Yes
Brand
Zippette
Manufacturer number
263-000
Model
263-000
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Pipettors, with 10 measurements at 3 volumes
YO-17104-00
EACH
Usually ships in 23 days.
Add to Cart
Technical Resources
Related technical articles (Roll over for summary)
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Related Searches for Zippette Dispenser, 0-2.5 mL (263-000)
25 Ml Burette Fischer
2.5 Ml Burette
25 Ml Graduated Cylinder
25 Ml Pipette
25 Ml Pipet
25 Ml Beaker
25 Ml Erlenmeyer Flask
25 Ml Volumetric Flask
25 Ml Graduated Cylinder Plastic
25 Ml Disposable Pipettes
Related Products for Zippette Dispenser, 0-2.5 mL (263-000)
Image
Product Detail
SKU
Zippette Dispenser, 0-5.0 mL (263-001)
Category: Bottletop Dispensers
Description: Zippette Dispensers
Brand:
Zippette
Model Number:
263-001
YO-25204-11
More Details
Zippette Dispenser, 0-10.0 mL (263-002)
Category: Bottletop Dispensers
Description: Zippette Dispensers
Brand:
Zippette
Model Number:
263-002
YO-25204-12
More Details
Zippette Dispenser, 0-30.0 mL (263-003)
Category: Bottletop Dispensers
Description: Zippette Dispensers
Brand:
Zippette
Model Number:
263-003
YO-25204-13
More Details
Zippette Dispenser, 0-50.0 mL (263-004)
Category: Bottletop Dispensers
Description: Zippette Dispensers
Brand:
Zippette
Model Number:
263-004
YO-25204-14
More Details
Chempette digital repetitive dispenser, 1-5 mL (A3-21DG-5)
Category: Repetitive Dispensers/Pipettors
Description: Cole-Parmer Chempette Digital Repetitive Dispensers
Brand:
Chempette
Model Number:
A3-21DG-5
YO-07848-30
More Details
Chempette digital repetitive dispenser, 2-10 mL (A3-21DG-10)
Category: Repetitive Dispensers/Pipettors
Description: Cole-Parmer Chempette Digital Repetitive Dispensers
Brand:
Chempette
Model Number:
A3-21DG-10
YO-07848-40
More Details
Brinkmann ChemSaver Dispenser 1-5 mL
Category: Bottletop Dispensers
Description: Brinkmann ChemSaver Dispensers
Brand:
Brinkmann
Model Number:
22 22 100-1
YO-07932-01
More Details
Brinkmann ChemSaver Dispenser 2-10 mL
Category: Bottletop Dispensers
Description: Brinkmann ChemSaver Dispensers
Brand:
Brinkmann
Model Number:
22 22 110-8
YO-07932-02
More Details
GLI 1000 Ohm electrodeless conductivity sensor, PP, 3/4 (37 2 5 E2T XX)
Category: Conductivity Electrodes - Industrial
Description: GLI 1000 Ω Pt RTD Electrodeless Conductivity Cells
Brand:
GLI
Model Number:
37 2 5 E2T XX
YO-19065-54
More Details
Brand Positive Displacement Pipettor Tips, PP, 1-5 Ml, 10/pk
Category: Pipette Tips
Description: Brand Positive Displacement Pipettor Tips
Brand:
Cole-Parmer
Model Number:
702858
YO-24550-77
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Zippette Dispenser, 0-2.5 mL (263-000)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Zippette Dispenser, 0-10.0 mL (263-002)
|
Next: Zippette Dispenser, 0-30.0 mL (263-003)
31125329-p-content_2 31125329pcontent2