Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Zippette Dispenser 0 50 0 mL
Click to enlarge
More Product Details
Specifications
Calibrations
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-25204-14
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
YO-17104-00
NIST-Traceable Calibration; Pipettors, with 10 measurements at 3 volumes
$122.86 AUD/EACH/EACH
Usually ships in 23 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Bottletop Dispensers
Share
|
Zippette Dispenser, 0-50.0 mL (263-004)
YO-25204-14
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
These ZIPPETTE dispensers are simple and durable enough to use in the lab, the field, or the classroom.
Redesigned twist-to-lock volume adjuster stays secure to ensure accuracy. To dispense, simply set volume, lock in adjuster, then lift piston and push down. A delivery tube of PTFE lets you safely fill handheld narrow-neck containers. Piston assembly is easy to disassemble for cleaning.
All models have a polypropylene-encased body, borosilicate glass syringe, chemical-resistant plunger, and a safety valve system contained within a PVDF pedestal that holds a primeprevents reagent loss during re-priming. Wetted parts are borosilicate glass and PTFE. Each dispenser fits bottles with 30-mm necks and includes one 300-mm inlet feed tube and adapters for 38-, 40-, and 45-mm bottle necks. Order bottles separately.
DIGITRATE,ZIPPETTE—Reg TM Jencons Ltd.
Specifications
Product Type
Dispenser (Bottletop)
Volume (mL)
50
Accuracy
±0.3%
Division (mL)
1
Range (mL)
1 - 50
Reproducibility
±0.1%
Wetted parts
Borosilicate glass and PTFE
Autoclavable
Yes
Brand
Zippette
Manufacturer number
263-004
Model
263-004
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Pipettors, with 10 measurements at 3 volumes
YO-17104-00
EACH
Usually ships in 23 days.
Add to Cart
Technical Resources
Related technical articles (Roll over for summary)
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Related Searches for Zippette Dispenser, 0-50.0 mL (263-004)
Tutwiler Burette 500 Ml
Corning Fritted 500 Ml Buchner Funnel
500 Ml Glass Bottles for Labs
500 Ml Glass Vial
500 Ml Glass Beakers
500 Ml Plastic Beaker
500 Ml Erlenmeyer Flask
500 Ml Beaker
500 Ml Volumetric Flask
Pyrex Beaker 500 Ml
Related Products for Zippette Dispenser, 0-50.0 mL (263-004)
Image
Product Detail
SKU
Zippette Dispenser, 0-5.0 mL (263-001)
Category: Bottletop Dispensers
Description: Zippette Dispensers
Brand:
Zippette
Model Number:
263-001
YO-25204-11
More Details
Zippette Dispenser, 0-2.5 mL (263-000)
Category: Bottletop Dispensers
Description: Zippette Dispensers
Brand:
Zippette
Model Number:
263-000
YO-25204-10
More Details
Zippette Dispenser, 0-10.0 mL (263-002)
Category: Bottletop Dispensers
Description: Zippette Dispensers
Brand:
Zippette
Model Number:
263-002
YO-25204-12
More Details
Zippette Dispenser, 0-30.0 mL (263-003)
Category: Bottletop Dispensers
Description: Zippette Dispensers
Brand:
Zippette
Model Number:
263-003
YO-25204-13
More Details
Centrifuge Tube Rack for 50-mL centrifuge tubes (18794)
Category: Tube Racks
Description: 50-mL Centrifuge Tube Rack
Brand:
Scienceware
Model Number:
18794
YO-06696-00
More Details
Cole-Parmer Chempette Ultramacropipettors Pipette Tips, 50 mL, 5/pk
Category: Pipette Tips
Description: Cole-Parmer Chempette Ultramacropipettors Pipette Tips
Brand:
Cole-Parmer
Model Number:
51TL
YO-07859-62
More Details
Thermo Scientific Finnpipette ® Dispenser, 1.0 to 5.0 mL (4421140)
Category: Bottletop Dispensers
Description: Thermo Scientific Finnpipette Dispensers
Brand:
Thermo Scientific
Model Number:
4421140
YO-07903-35
More Details
Thermo Scientific Finnpipette ® Dispenser, 5.0 to 30.0 mL (4421160)
Category: Bottletop Dispensers
Description: Thermo Scientific Finnpipette Dispensers
Brand:
Thermo Scientific
Model Number:
4421160
YO-07903-55
More Details
Adjustable-Volume analog dispenser - 5 - 50 mL (4701161)
Category: Brand Dispensette III Bottletop Dispensers
Description: Dispensette III Adjustable-Volume Dispensers, analog volume setting
Brand:
Dispensette
Model Number:
4701161
YO-07902-24
More Details
Brinkmann ChemSaver Dispenser 10-50 mL
Category: Bottletop Dispensers
Description: Brinkmann ChemSaver Dispensers
Brand:
Brinkmann
Model Number:
22 22 130-2
YO-07932-04
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Zippette Dispenser, 0-50.0 mL (263-004)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Zippette Dispenser, 0-5.0 mL (263-001)
|
Next: Zirconia grinding media, 1/2" x 1/2" (YO-04158-20)
31125329-p-content_2 31125329pcontent2