Compact 10-Outlet (5 x 2) Power Strip with 15 Ft Cord
YO-26870-04
Loading price...
Usually ships in 14 days.
Qty
  • Centralize your power supply with these UL-listed switched outlet strips
Each model features an aluminum housing and impact-resistant plastic end caps. All outlet strips meet UL standard 1363 and have a built-in circuit breaker.
Compact outlet strips let you supply power to areas where space is limited. Outlet strips have a combination ON/OFF switch/pilot light. Keyhole mount on the wall or below a workbench. CSA-certified
Product Type Outlet Strips
No. of outlets 10
Line voltage 120 V
Dimensions (" W) 3.5
Dimensions (" H) 2
Dimensions (" L) 12
Cord length (ft) 15
Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Related Products for Compact 10-Outlet (5 x 2) Power Strip with 15 Ft Cord
    Image
      Product Detail
    SKU
    Compact 10-Outlet (5 x 2) Power Strip with 6 Ft Cord
    Category: Outlet Strips
    Description: Compact Outlet Strips
    Brand: Cole-Parmer
    Bench-Mount 10-Outlet Power Strip with 15 Ft Cord
    Category: Outlet Strips
    Description: Bench-Mount Outlet Strips
    Brand: Cole-Parmer
    Heavy-Duty, 20 Amp, 6-Outlet Power Strip with 15 Ft Cord
    Category: Outlet Strips
    Description: Heavy-Duty 20 A Outlet Strips
    Brand: Cole-Parmer
    Standard 6-Outlet Power Strip with 15 Ft Cord
    Category: Outlet Strips
    Description: Standard Outlet Strips
    Brand: Cole-Parmer
    Bench-Mount 12-Outlet Power Strip with 15 Ft Cord
    Category: Outlet Strips
    Description: Bench-Mount Outlet Strips
    Brand: Cole-Parmer
    Bench-Mount 10-Outlet Power Strip with 6 Ft Cord
    Category: Outlet Strips
    Description: Bench-Mount Outlet Strips
    Brand: Cole-Parmer
    Heavy-Duty, 20 Amp, 6-Outlet Power Strip with 6 Ft Cord
    Category: Outlet Strips
    Description: Heavy-Duty 20 A Outlet Strips
    Brand: Cole-Parmer
    Standard 6-Outlet Power Strip with 6 Ft Cord
    Category: Outlet Strips
    Description: Standard Outlet Strips
    Brand: Cole-Parmer
    Bench-Mount 12-Outlet Power Strip with 6 Ft Cord
    Category: Outlet Strips
    Description: Bench-Mount Outlet Strips
    Brand: Cole-Parmer
    Ethyl 5-bromo-2-chlorobenzoate 97% (10ml) (AC250030100)
    Category: Ethylb to Ethyld
    Description: Ethyl 5-bromo-2-chlorobenzoate
    Brand: Cole-Parmer
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
    Questions and Answers
    Product Reviews
    Compact 10-Outlet (5 x 2) Power Strip with 15 Ft Cord
    Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended
    Popularity |  Favorites |  Title |  Price |  Part Number |  Category
    Title |  Brand |  Category |  Price |  Popularity |  Best Sellers
    Previous: Communications Module (3560)  |  Next: Compact 10-Outlet (5 x 2) Power Strip with 6 Ft Cord