Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
ZoneMaster Surge Protectors 180ka 3 phase surge 120
Click to enlarge
More Product Details
Specifications
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-26874-12
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Surge and Noise Suppressors, and Power Conditioners
Share
|
ZoneMaster Surge Protectors, 180ka 3-phase surge 120
YO-26874-12
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Provide up to 300,000 amps of protection per phase for sub-panels to main distribution panels
Full surge protection for all modes (L-N, L-G, L-L, and N-G)
Constructed of a durable, high-impact plastic, these surge protectors conform to NEMA 4x. NO/NC contacts rated for 125 VAC, 2 A provide remote indication of alarm status. Internal protection modules feature both mechanical and LED status indication. Each module also provides dual thermal and short circuit fusing protection mechanisms.
Models 26874-00 to –04 designed specifically for applications at distribution boards and small service locations. Units provide maximum surge capacity of 90,000 A per phase and 45,000 A per mode.
Models 26874-10 to –24 provide protection at large service entrances and distribution panels. The dual protection circuits provide redundancy ensuring the site is always protected. Models 26874-10 to -14 provide maximum surge capacity of 180,000 A per phase and 90,000 A per mode. Models 26874-20 to –24 provide maximum surge capacity 300,000 A per phase and 150,000 A per mode.
Specifications
Product Type
Surge Protectors
Surge suppression
400 V (UL1449)
Stages of protection
3
Line voltage
120/208 VAC
Dimensions (" W)
12
Dimensions (" H)
12
Dimensions (" D)
6
Max current capacity (A/phase)
180,000
Max current capacity (A/mode)
90,000
Brand
ZoneMaster
Manufacturer number
ZB120Y
Model
ZB120Y
Technical Resources
Related technical articles (Roll over for summary)
Technical Expertise: How Cole-Parmer Application Specialists Respond to Customers' Needs
ABCs of DMMs
Show Less...
Related Searches for ZoneMaster Surge Protectors, 180ka 3-phase surge 120
Wiremold / Legrand Hard-Wired Surge Protectors
Surge Protector 3
Air Compressor 3 Phase
4 Wire 3 Phase
3 Phase Synchronous Motor
3 Phase Inverter
Extech 3-Phase Power & Harmonics Analyzers
3 Phase Rotation Tester
Single Surge Suppressor
Zonemaster Process Equipment & Instrumentation
Related Products for ZoneMaster Surge Protectors, 180ka 3-phase surge 120
Image
Product Detail
SKU
ZoneMaster Surge Protectors 180ka 3-phase surge 277 (ZB277Y)
Category: Surge and Noise Suppressors, and Power Conditioners
Description: ZoneMaster Surge Protectors
Brand:
ZoneMaster
Model Number:
ZB277Y
YO-26874-14
More Details
ZoneMaster Surge Protectors 3-phase surge 120 (ZC120Y)
Category: Surge and Noise Suppressors, and Power Conditioners
Description: ZoneMaster Surge Protectors
Brand:
ZoneMaster
Model Number:
ZC120Y
YO-26874-22
More Details
ZoneMaster Surge Protectors, single phase, 3 wire, 90,000 A per phase (ZA120T)
Category: Surge and Noise Suppressors, and Power Conditioners
Description: ZoneMaster Surge Protectors
Brand:
ZoneMaster
Model Number:
ZA120T
YO-26874-00
More Details
ZoneMaster Surge Protectors, 4 wire, 90,000 A per phase device 90ka 3- (ZA120Y)
Category: Surge and Noise Suppressors, and Power Conditioners
Description: ZoneMaster Surge Protectors
Brand:
ZoneMaster
Model Number:
ZA120Y
YO-26874-02
More Details
ZoneMaster Surge Protectors, 4 wire, 90,000 A per phase device 90ka 3- (ZA277Y)
Category: Surge and Noise Suppressors, and Power Conditioners
Description: ZoneMaster Surge Protectors
Brand:
ZoneMaster
Model Number:
ZA277Y
YO-26874-04
More Details
ZoneMaster Surge Protectors, single phase, 3 wire, 180,000 A per phase (ZB120T)
Category: Surge and Noise Suppressors, and Power Conditioners
Description: ZoneMaster Surge Protectors
Brand:
ZoneMaster
Model Number:
ZB120T
YO-26874-10
More Details
ZoneMaster Surge Protectors, single phase, 3 wire, 300,000 A per phase (ZC120T)
Category: Surge and Noise Suppressors, and Power Conditioners
Description: ZoneMaster Surge Protectors
Brand:
ZoneMaster
Model Number:
ZC120T
YO-26874-20
More Details
ZoneMaster Surge Protectors 3-phase surge 277 (ZC277Y)
Category: Surge and Noise Suppressors, and Power Conditioners
Description: ZoneMaster Surge Protectors
Brand:
ZoneMaster
Model Number:
ZC277Y
YO-26874-24
More Details
Single Phase, 3 Wire Surge Protector (80,000 A per phase), 120/240 VAC (PA120T)
Category: Surge and Noise Suppressors, and Power Conditioners
Description: WIREMOLD / LEGRAND Hard-Wired Surge Protectors
Brand:
Sentrex
Model Number:
PA120T
YO-26873-00
More Details
Single Phase, 3 Wire Surge Protector (160,000 A per phase), 120/240 VA (PB120T)
Category: Surge and Noise Suppressors, and Power Conditioners
Description: WIREMOLD / LEGRAND Hard-Wired Surge Protectors
Brand:
Sentrex
Model Number:
PB120T
YO-26873-10
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
ZoneMaster Surge Protectors, 180ka 3-phase surge 120
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: ZoneMaster Surge Protectors 3-phase surge 277 (ZC277Y)
|
Next: ZoneMaster Surge Protectors, 4 wire, 90,000 A per phase device 90ka 3- (ZA120Y)
31125329-p-content_2 31125329pcontent2