Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
YO-68930-10
Representative Photo Only. Ashcroft
®
Dual-Scale Low-Pressure Gauge.
Click to enlarge
More Product Details
Specifications
Calibrations
Technical Resources
Virtual Catalog (PDFs)
More info
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-68930-10
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
YO-17040-10
NIST-Traceable Calibration; Pressure / Vacuum (For gauges with accuracy 1% or >1%)
$184.29 AUD/EACH/EACH
Usually ships in 18 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Mechanical Pressure Gauges
Share
|
Ashcroft Dual-Scale Low-Pressure Gauge, 0 to 3 psi and 0 to 20 kPa
YO-68930-10
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Effective low-pressure and low-vacuum response with diaphragm
True zero indication identifies damaged gauges increasing safety and ensuring accuracy
Built-in snubber minimizes rapid pressure fluctuations
Gauges have brass socket, beryllium copper diaphragm capsule, painted steel case, and polycarbonate lens. Bottom connection is 1/4" NPT(M).
Specifications
Product Type
Commercial gauges
Media compatibility
any media not corrosive to beryllium copper, brass, polysulfone, and RTV silicone
Wetted materials
beryllium copper, brass, polysulfone, RTV silicone
Accuracy
±1% (ANSI/ASME grade A)
Range
0 to 3 psi; 0 to 20 kPa
Process connection
1/4" NPT(M), lower
Liquid fill
none
Dial size (" Dia)
2.5
Case
Polysulphone
Lens material
Polycarbonate
Display
analog dial
Brand
Ashcroft
Manufacturer number
25-1490-A-02L-XTU-3#/KPA
Model
14902.5
Make
Type1490
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Pressure / Vacuum (For gauges with accuracy 1% or >1%)
YO-17040-10
EACH
Usually ships in 18 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
13-14 International Catalog
More info
Dimensional Drawing
Pressure Gauge Selection Made Easy!
Related technical articles (Roll over for summary)
Absolute, Gauge, and Differential Pressure Scales
Common Pressure Terms
Pressure Unit Conversion Constants
Show All...
The Hidden Costs of Mercury-Filled Vacuum Gauges
Show Less...
Related Searches for Ashcroft Dual-Scale Low-Pressure Gauge, 0 to 3 psi and 0 to 20 kPa
Low Pressure Gauge 0 5 Psi
Ashcroft Low-Pressure Gauges
Low Pressure Gauge
Differential Low Pressure Gauges
Water Pressure Gauges 2 125 Psi
Ashcroft Dual-Scale Gauges
Low Pressure Gauge 35 20
Low Scale Pressure Gauge
Ashcroft Low-Pressure and Low-Vacuum Gauges
Low Oil Pressure Gauges
Related Products for Ashcroft Dual-Scale Low-Pressure Gauge, 0 to 3 psi and 0 to 20 kPa
Image
Product Detail
SKU
Ashcroft Dual-Scale Low-Pressure Gauge, 0 to 5 psi and 0 to 35 kPa
Category: Mechanical Pressure Gauges
Description: Ashcroft Low-Pressure and Low-Vacuum Gauges
Brand:
Ashcroft
Model Number:
14902.5
YO-68930-11
More Details
Ashcroft Dual-Scale Low-Pressure Gauge, 0 to 10 psi and 0 to 70 kPa
Category: Mechanical Pressure Gauges
Description: Ashcroft Low-Pressure and Low-Vacuum Gauges
Brand:
Ashcroft
Model Number:
14902.5
YO-68930-12
More Details
Ashcroft Dual-Scale Low-Pressure Gauge, 0 to 10" H2O and 0
Category: Mechanical Pressure Gauges
Description: Ashcroft Low-Pressure and Low-Vacuum Gauges
Brand:
Ashcroft
Model Number:
14902.5
YO-68930-00
More Details
Ashcroft Dual-Scale Low-Pressure Gauge, 0 to 15" H2O and 0
Category: Mechanical Pressure Gauges
Description: Ashcroft Low-Pressure and Low-Vacuum Gauges
Brand:
Ashcroft
Model Number:
14902.5
YO-68930-01
More Details
Ashcroft Dual-Scale Low-Pressure Gauge, 0 to 30" H2O and 0
Category: Mechanical Pressure Gauges
Description: Ashcroft Low-Pressure and Low-Vacuum Gauges
Brand:
Ashcroft
Model Number:
14902.5
YO-68930-02
More Details
Ashcroft Dual-Scale Low-Pressure Gauge, 0 to 60" H2O and 0
Category: Mechanical Pressure Gauges
Description: Ashcroft Low-Pressure and Low-Vacuum Gauges
Brand:
Ashcroft
Model Number:
14902.5
YO-68930-03
More Details
Ashcroft Dual-Scale Low-Pressure Gauge, 0 to 100" H2O and 0
Category: Mechanical Pressure Gauges
Description: Ashcroft Low-Pressure and Low-Vacuum Gauges
Brand:
Ashcroft
Model Number:
14902.5
YO-68930-04
More Details
Ashcroft Dual-Scale Low-Pressure Gauge, 0 to 160" H2O and 0
Category: Mechanical Pressure Gauges
Description: Ashcroft Low-Pressure and Low-Vacuum Gauges
Brand:
Ashcroft
Model Number:
14902.5
YO-68930-05
More Details
Ashcroft Dual-Scale Low-Pressure Gauge, 0 to 200" H2O and 0
Category: Mechanical Pressure Gauges
Description: Ashcroft Low-Pressure and Low-Vacuum Gauges
Brand:
Ashcroft
Model Number:
14902.5
YO-68930-06
More Details
Ashcroft Dual-Scale Low-Vacuum Gauge, 15" H2O to 0 and 0 to
Category: Mechanical Pressure Gauges
Description: Ashcroft Low-Pressure and Low-Vacuum Gauges
Brand:
Ashcroft
Model Number:
14902.5
YO-68930-30
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Ashcroft Dual-Scale Low-Pressure Gauge, 0 to 3 psi and 0 to 20 kPa
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Ashcroft Dual-Scale Low-Pressure Gauge, 0 to 200" H2O and 0
|
Next: Ashcroft Dual-Scale Low-Pressure Gauge, 0 to 30" H2O and 0
31125329-p-content_2 31125329pcontent2