Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Microbial systems
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-29200-10
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Cole-Parmer Modular Fermentation Start-Up Kits
Share
|
Microbial systems (YO-29200-10)
YO-29200-10
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Modular design makes customization easy
Choose start-up kits for microbial or mammalian cell culture applications
Scale-up is easy—all kits feature the same dimensional geometry for easy duplication of conditions
Start-up kits come with a nonwelded headplate and a lipseal stirrer assembly for maximum versatility
These fermentation kits, made exclusively for Cole-Parmer by Applikon, feature a modular design which allows you to interchange vessels, headplates, stirrer assemblies, and optional accessories.
The component chart, which can be accessed by downloading the catalog page in .pdf format (see below right) indicates the basic components included with each fermentation start-up kit.
Complete your system by ordering: 1) Meters and controllers for your application and 2) Fermentation accessories required by your application. A magnetic stirrer assembly is available by special order when contamination-free work is critical.
Systems for Microbial and Bacterial Applications
—High speed shearing impeller drives large volumes of oxygen into the solution while at the same time breaking up large clumps of cells—ensuring that your samples receive the proper aeration.
Microbial systems include: two six-bladed flat disc impellers, one nonwelded headplate, one lipseal stirrer assembly, and one heat exchanger to control temperature.
Specifications
Product Type
Fermentation, Microbial kits
Working volume (Liters)
2.7
Vessel volume (Liters)
3
Motor speed
0 to 1250
Port size (mm)
6, 10, 12, 18, 19.05
Power (VAC)
110
Manufacturer number
Z61103MCPF
Brand
Cole-Parmer
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
13-14 International Catalog
Related technical articles (Roll over for summary)
Cole-Parmer Introduces Enhancements to Life Science Offering
Show Less...
Related Searches for Microbial systems (YO-29200-10)
Systems for Microbial and Bacterial Applications
Cole Systems for Microbial An
Microbial Fermentation
Microbial Kits
Cole-Parmer Soxhlet Extraction Glassware Systems
Cole-Parmer Conical-Bottom Tank Systems
Cole-Parmer all-in-One Gas Purification Systems
Cole-Parmer Systems for Cell Culture Applications
Innocal Temperature System Calibrations
Video Microscopy Systems
Related Products for Microbial systems (YO-29200-10)
Image
Product Detail
SKU
Microbial systems (YO-29200-11)
Category: Cole-Parmer Modular Fermentation Start-Up Kits
Description: Systems for Microbial and Bacterial Applications
Brand:
Cole-Parmer
YO-29200-11
More Details
Microbial systems (YO-29200-30)
Category: Cole-Parmer Modular Fermentation Start-Up Kits
Description: Systems for Microbial and Bacterial Applications
Brand:
Cole-Parmer
YO-29200-30
More Details
Microbial systems (YO-29200-31)
Category: Cole-Parmer Modular Fermentation Start-Up Kits
Description: Systems for Microbial and Bacterial Applications
Brand:
Cole-Parmer
YO-29200-31
More Details
Microbial systems (YO-29200-40)
Category: Cole-Parmer Modular Fermentation Start-Up Kits
Description: Systems for Microbial and Bacterial Applications
Brand:
Cole-Parmer
YO-29200-40
More Details
Microbial systems (YO-29200-41)
Category: Cole-Parmer Modular Fermentation Start-Up Kits
Description: Systems for Microbial and Bacterial Applications
Brand:
Cole-Parmer
YO-29200-41
More Details
Spacer Set for Cole-Parmer Vertical Slab Gel Systems; 14.5 cm Height, (YO-28573-10)
Category: Cole-Parmer Adjustable Height Single and Dual Vertical Gel Systems
Description: Cole-Parmer Vertical Slab Gel Spacer Set for Height-adjustable units
Brand:
Cole-Parmer
YO-28573-10
More Details
Cole-Parmer Isocratic HPLC Systems with fixed wavelength detectors, 30 (YO-42661-10)
Category: High Pressure Liquid Chromatography (HPLC) Systems
Description: Isocratic HPLC Systems
Brand:
Cole-Parmer
YO-42661-10
More Details
PhotoDoc-It 50 Imaging System, 115VAC (YO-97700-10)
Category: Imaging Products for Gel Electrophoresis
Description: PhotoDoc-It Imaging Systems
Brand:
PhotoDoc-It
YO-97700-10
More Details
Soxhlet extraction glassware systems, 125 mL (YO-99036-10)
Category: Extraction Apparatus
Description: Soxhlet Extraction Systems
Brand:
Cole-Parmer
YO-99036-10
More Details
Lab-Tek™ Chambered Cover Glass System; material, glass; number of w (YO-01838-10)
Category: Lab-Tek Chamber Slide Systems
Description: Lab-Tek Chambered Cover Glass Systems
Brand:
Lab-Tek
YO-01838-10
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Microbial systems (YO-29200-10)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Microbial air sampler, digital, stainless steel head, 115 VAC (18198-00)
|
Next: Microbial systems (YO-29200-11)
31125329-p-content_2 31125329pcontent2