Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Systems for cell culture applications 220 VAC
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-29200-43
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Cole-Parmer Modular Fermentation Start-Up Kits
Share
|
Systems for cell culture applications, 220 VAC (YO-29200-43)
YO-29200-43
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Modular design makes customization easy
Choose start-up kits for microbial or mammalian cell culture applications
Scale-up is easy—all kits feature the same dimensional geometry for easy duplication of conditions
Start-up kits come with a nonwelded headplate and a lipseal stirrer assembly for maximum versatility
These fermentation kits, made exclusively for Cole-Parmer by Applikon, feature a modular design which allows you to interchange vessels, headplates, stirrer assemblies, and optional accessories.
The component chart, which can be accessed by downloading the catalog page in .pdf format (see below right) indicates the basic components included with each fermentation start-up kit.
Complete your system by ordering: 1) Meters and controllers for your application and 2) Fermentation accessories required by your application. A magnetic stirrer assembly is available by special order when contamination-free work is critical.
Systems for Cell Culture Applications
—A gentle environment for optimal cell growth—air inlet pipe with porous sparger gently aerates your cultures.
—Scoping marine impeller efficiently mixes components within the media while minimizing shear.
—Use the sample pipe assembly with 25-µm filter (included) to extract samples from your cell cultures.
—Available in water jacketed (flat bottom) and non-water jacketed (round bottom) versions
Maintain the system temperature in the non-water jacketed vessels by using the heat exchanger, or by wrapping the entire vessel in an optional heating blanket.
All cell culture systems include: one scoping marine impeller, one nonwelded headplate, and one lipseal stirrer assembly.
Specifications
Product Type
Fermentation, Cell Culture kits
Working volume (Liters)
10
Vessel volume (Liters)
15
Motor speed
0 to 200
Port size (mm)
10, 12, 27
Power (VAC)
220
Manufacturer number
Z61110CCPR-220VAC
Brand
Cole-Parmer
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
13-14 International Catalog
Related technical articles (Roll over for summary)
Cole-Parmer Introduces Enhancements to Life Science Offering
Show Less...
Related Searches for Systems for cell culture applications, 220 VAC (YO-29200-43)
Cole-Parmer Systems for Cell Culture Applications
Cole Systems for Cell Culture
Cell Culture Fermentation
Cell Culture Kits
Cell Culture Water Bath
Cell Culture Fermentation Kits
Smallest Cover Glasses Cell Cultures
Glass Cell Culture Spinner Flasks
Wheaton Cell Culture Spin Flasks
Integra Biosciences Tissue and Cell Culture
Related Products for Systems for cell culture applications, 220 VAC (YO-29200-43)
Image
Product Detail
SKU
Systems for cell culture applications, 220 VAC (YO-29200-13)
Category: Cole-Parmer Modular Fermentation Start-Up Kits
Description: Systems for Cell Culture Applications
Brand:
Cole-Parmer
YO-29200-13
More Details
Systems for cell culture applications, 220 VAC (YO-29200-33)
Category: Cole-Parmer Modular Fermentation Start-Up Kits
Description: Systems for Cell Culture Applications
Brand:
Cole-Parmer
YO-29200-33
More Details
Systems for cell culture applications, 115 VAC (YO-29200-12)
Category: Cole-Parmer Modular Fermentation Start-Up Kits
Description: Systems for Cell Culture Applications
Brand:
Cole-Parmer
YO-29200-12
More Details
Systems for cell culture applications, 115 VAC (YO-29200-32)
Category: Cole-Parmer Modular Fermentation Start-Up Kits
Description: Systems for Cell Culture Applications
Brand:
Cole-Parmer
YO-29200-32
More Details
Systems for cell culture applications, 115 VAC (YO-29200-42)
Category: Cole-Parmer Modular Fermentation Start-Up Kits
Description: Systems for Cell Culture Applications
Brand:
Cole-Parmer
YO-29200-42
More Details
Systems for cell culture applications, water-jacketed, 220 VAC (YO-29200-15)
Category: Cole-Parmer Modular Fermentation Start-Up Kits
Description: Systems for Cell Culture Applications
Brand:
Cole-Parmer
YO-29200-15
More Details
Systems for cell culture applications, water-jacketed, 220 VAC (YO-29200-35)
Category: Cole-Parmer Modular Fermentation Start-Up Kits
Description: Systems for Cell Culture Applications
Brand:
Cole-Parmer
YO-29200-35
More Details
Systems for cell culture applications, water-jacketed, 115 VAC (YO-29200-14)
Category: Cole-Parmer Modular Fermentation Start-Up Kits
Description: Systems for Cell Culture Applications
Brand:
Cole-Parmer
YO-29200-14
More Details
Systems for cell culture applications, water-jacketed, 115 VAC (YO-29200-34)
Category: Cole-Parmer Modular Fermentation Start-Up Kits
Description: Systems for Cell Culture Applications
Brand:
Cole-Parmer
YO-29200-34
More Details
Cell culture flask, 500 ml (YO-29300-00)
Category: Mini Suspension Culture Systems
Description: Cell Culture Spin Flasks
Brand:
Wheaton
YO-29300-00
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Systems for cell culture applications, 220 VAC (YO-29200-43)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Systems for cell culture applications, 220 VAC (YO-29200-33)
|
Next: Systems for cell culture applications, water-jacketed, 115 VAC (YO-29200-14)
31125329-p-content_2 31125329pcontent2