Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
KST47 WASHER
Click to enlarge
More Product Details
Specifications
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-29902-16
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Pressure Filtration Holders
Share
|
Kst47 Washer (301514)
YO-29902-16
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Specifications
Manufacturer number
301514
Model
301514
Related Searches for Kst47 Washer (301514)
Stainless Steel Washers
Pressure Washer
96 Well Microplate Washer
Masterflex Thrust Washers
96 Well Plate Washer
Washer Rubber
Biotek Microplate Washers
Thermo Scientific Nunc Nunc-Immuno Washers
Biotek Cole-Parmer Automatic Strip Washers
Bod Bottle Washers
Related Products for Kst47 Washer (301514)
Image
Product Detail
SKU
Undercounter Glassware Washer 115V (YO-98950-01)
Category: Glassware Washers
Description: Cole-Parmer
®
Laboratory Glassware Washers
Brand:
Cole-Parmer
YO-98950-01
More Details
Undercounter, General-Purpose Washer, 230V (YO-98950-05)
Category: Glassware Washers
Description: Cole-Parmer
®
Laboratory Glassware Washers
Brand:
Cole-Parmer
YO-98950-05
More Details
Freestanding Glassware Washer, 115V (YO-98950-11)
Category: Glassware Washers
Description: Cole-Parmer
®
Laboratory Glassware Washers
Brand:
Cole-Parmer
YO-98950-11
More Details
Undercounter Spindle-ready Washer, 230V (YO-98950-20)
Category: Glassware Washers
Description: Cole-Parmer
®
Laboratory Glassware Washers
Brand:
Cole-Parmer
YO-98950-20
More Details
Free Standing Spindle-Ready Washer, 230V
Category: Glassware Washers
Description: Cole-Parmer
®
Laboratory Glassware Washers
Brand:
Cole-Parmer
YO-98950-30
More Details
Free Standing Spindle-ready Washer, 230V (CLW-1285I-FS) (YO-98950-35)
Category: Glassware Washers
Description: Cole-Parmer
®
Laboratory Glassware Washers
Brand:
Cole-Parmer
YO-98950-35
More Details
Cuvette washer
Category: Cuvette Accessories
Description: Vakuwash Cuvette Washer
Brand:
Scienceware
Model Number:
38960-0000
YO-83300-00
More Details
Heated Dry Cycle for Cole-Parmer® Glassware Washers (YO-98950-40)
Category: Glassware Washers
Description: Cole-Parmer
®
Laboratory Glassware Washers
Brand:
Cole-Parmer
YO-98950-40
More Details
FlaskScrubber Undercounter Glassware Washer, 115V 60HZ (4420320)
Category: Labconco Laboratory Glassware Washers & Accessories
Description: Labconco FlaskScrubber Laboratory Glassware Washers
Brand:
FlaskScrubber
Model Number:
4420320
YO-98963-00
More Details
FlaskScrubber Undercounter Hi-Temperature Glassware Washer, 230V 50/60 (4420321)
Category: Labconco Laboratory Glassware Washers & Accessories
Description: Labconco FlaskScrubber Laboratory Glassware Washers
Brand:
FlaskScrubber
Model Number:
4420321
YO-98963-05
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Kst47 Washer (301514)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Kst47 Butterfly Bolts (301512)
|
Next: Kuderna-Danish Concentrator without springs, 1L capacity, 720mm H
31125329-p-content_2 31125329pcontent2