Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, Fittings, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
MF Millipore Membrane MCE 0 22 m 47 mm dia
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-29959-00
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Membranes
Share
|
MF Millipore Membrane, MCE, 0.22 µm, 47 mm dia (YO-29959-00)
YO-29959-00
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
The laboratory standard for pressure or vacuum-driven filtration
MF-Millipore
®
(mixed cellulose esters) is the most widely used membrane for sterilization
This item is considered a hazardous material.
Please note: there may be additional shipping restrictions like freight collect or no priority air service, extra freight charges may apply. This item(s) may be restricted from shipping to a residential address, sold to private individuals or exported from the United States. For additional information or questions please call us at 1-800-323-4340 (US) or 847-549-7600 (International).
Specifications
Product Type
Mixed Cellulose Ester Membranes
Material
Mixed Cellulose Ester
Flow rate (mL/min/sq cm)
18
Diameter (mm)
47
Pore size (µm)
0.22
Qty/pk
100
Bubble point (bar sq)
3.52
Brand
Millipore
Manufacturer number
GSTF04700
Technical Resources
Virtual Catalog (PDFs)
11-12 Catalog
11-12 International Catalog
13-14 Catalog
13-14 International Catalog
Related technical articles (Roll over for summary)
Laboratory Filtration Selection Guide
Science in the Small Dairy Industry: Milk Sediment Testing
Basics of Cartridge Filtration
Show All...
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Related Searches for MF Millipore Membrane, MCE, 0.22 µm, 47 mm dia (YO-29959-00)
Millipore Membranes
47 Mm Ca Filters
47 Mm Mce Filters
47 Mm Mce Membranes
Advantec 47 Mm Mce Membrane
47 Mm Membrane for Asbestos Monitoring
Advantec 47 Mm Pfa Membrane Holders
47 Mm Ptfe Membrane Filter
47 Mm Mixed Cellulose Ester Membranes
47 Mm Cellulose Acetate Membranes
Related Products for MF Millipore Membrane, MCE, 0.22 µm, 47 mm dia (YO-29959-00)
Image
Product Detail
SKU
MF Millipore Membrane, MCE, 0.45 µm, 47 mm dia (YO-29959-02)
Category: Membranes
Description: Millipore Membranes
Brand:
Millipore
YO-29959-02
More Details
MF Millipore Membrane, MCE, 0.22 µm, 90 mm dia (YO-29959-10)
Category: Membranes
Description: Millipore Membranes
Brand:
Millipore
YO-29959-10
More Details
MF Millipore Membrane, MCE, 0.45 µm, 90 mm dia (YO-29959-12)
Category: Membranes
Description: Millipore Membranes
Brand:
Millipore
YO-29959-12
More Details
Millipore Fluoropore Membrane, PTFE, 0.22 µm, 47 mm dia
Category: Membranes
Description: Millipore Membranes
Brand:
Millipore
YO-29958-00
More Details
Millipore Fluoropore Membrane, PTFE, 0.50 µm, 47 mm dia
Category: Membranes
Description: Millipore Membranes
Brand:
Millipore
YO-29958-02
More Details
Millipore Fluoropore Membrane, PTFE, 1.0 µm, 47 mm dia
Category: Membranes
Description: Millipore Membranes
Brand:
Millipore
YO-29958-06
More Details
Millipore Durapore Membrane, PVDF, 0.22 µm, 47 mm dia
Category: Membranes
Description: Millipore Membranes
Brand:
Millipore
YO-29960-00
More Details
Millipore Durapore Membrane, PVDF, 0.45 µm, 47 mm dia
Category: Membranes
Description: Millipore Membranes
Brand:
Millipore
YO-29960-02
More Details
Millipore Express Membrane, PES, 0.22 µm, 47 mm dia
Category: Membranes
Description: Millipore Membranes
Brand:
Millipore
YO-29960-20
More Details
Net filter, nylon, 11 µm, 47 mm dia (YO-29959-20)
Category: Mesh Screen Discs, Sheets and Net Filters
Description: Millipore Net Filters
Brand:
Millipore
YO-29959-20
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
MF Millipore Membrane, MCE, 0.22 µm, 47 mm dia (YO-29959-00)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Metzenbaum dissecting scissors, Surgical-grade, blunt point, curved (YO-10921-12)
|
Next: MF Millipore Membrane, MCE, 0.22 µm, 90 mm dia (YO-29959-10)
31125329-p-content_2 31125329pcontent2