MF Millipore Membrane, MCE, 0.22 µm, 47 mm dia

YO-29959-00
Loading price...

Call for availability.
Qty

  • The laboratory standard for pressure or vacuum-driven filtration
  • MF-Millipore® (mixed cellulose esters) is the most widely used membrane for sterilization

This item is considered a hazardous material.Please note: there may be additional shipping restrictions like freight collect or no priority air service, extra freight charges may apply. This item(s) may be restricted from shipping to a residential address, sold to private individuals or exported from the United States. For additional information or questions please call us at 1-800-323-4340 (US) or 847-549-7600 (International).
Product Type Mixed Cellulose Ester Membranes
Material Mixed Cellulose Ester
Flow rate (mL/min/sq cm) 18
Diameter (mm) 47
Pore size (µm) 0.22
Qty/pk 100
Bubble point (bar sq) 3.52
Brand Millipore
Manufacturer number GSTF04700





Technical Resources
 
Related Products for MF Millipore Membrane, MCE, 0.22 µm, 47 mm dia
Image
  Product Detail
SKU

MF Millipore Membrane, MCE, 0.45 µm, 47 mm dia

Category: Membranes
Description: Millipore Membranes
Brand: Millipore


MF Millipore Membrane, MCE, 0.22 µm, 90 mm dia

Category: Membranes
Description: Millipore Membranes
Brand: Millipore


MF Millipore Membrane, MCE, 0.45 µm, 90 mm dia

Category: Membranes
Description: Millipore Membranes
Brand: Millipore


Millipore Fluoropore Membrane, PTFE, 0.22 µm, 47 mm dia

Category: Membranes
Description: Millipore Membranes
Brand: Millipore


Millipore Fluoropore Membrane, PTFE, 0.50 µm, 47 mm dia

Category: Membranes
Description: Millipore Membranes
Brand: Millipore


Millipore Fluoropore Membrane, PTFE, 1.0 µm, 47 mm dia

Category: Membranes
Description: Millipore Membranes
Brand: Millipore


Millipore Durapore Membrane, PVDF, 0.22 µm, 47 mm dia

Category: Membranes
Description: Millipore Membranes
Brand: Millipore


Millipore Durapore Membrane, PVDF, 0.45 µm, 47 mm dia

Category: Membranes
Description: Millipore Membranes
Brand: Millipore


Advantec MCE Solid White Filters, 0.8um, 47mm Dia; 100/Pk

Category: Advantec 37-mm Monitors for Microbial Contamination Analysis
Description: Advantec MCE Membranes for Asbestos Monitoring
Brand: Advantec


Advantec MCE Gridded Filters, 0.8um, 47mm Dia; 100/Pk

Category: Advantec 37-mm Monitors for Microbial Contamination Analysis
Description: Advantec MCE Membranes for Asbestos Monitoring
Brand: Advantec


Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
Questions and Answers

Do you have questions about this product?

get answers from real customers and in-house experts with AnswerBox.

1 Question | 1 Answer
Displaying question 1
  • Q:

    Hello,


    Can these membranes (29959-00) be sterilized with an autoclave? As they are flammable, what is the proper procedure?

    Thank you
    Asked on 7/21/2009 by Anonymous

    1 answer

    • STAFF EXPERT

      A:

      These membranes are for use to 55 degrees C. These are not filters to be sterilized and used multiple times. Please contact our technical application specialists for additional assistance (contact information can be found on the product web page).

      Answered on 7/21/2009 by Jen from Technical Support
Displaying question 1

Do you have a question about this product? 

Product Reviews