Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Hayward double length bag filter housing 2 NPT F
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-29990-04
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Bag Filter Systems
Share
|
Hayward double-length bag filter housing, 2" NPT(F)
YO-29990-04
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
All-plastic constructionno need to worry about rust and corrosion!
Ideal for water, chemical, pharmaceutical, food and other general fluid filtration applications
Feature hand-removable cover and built-in basket/bag handlessimply lift out the basket and replace the filter bag
External cover threads are not in contact with the process mediaeliminates the need for cleaning
Cover features buttress style threads for extra strength
Available in single- and double-length sizes; maximum flow rate for single-length size is 50 GPM, double-length size is 100 GPM
Each filter includes an integral mounting flange and Viton
®
seal; order bags at right
These bag filter systems are ideal for use as an economical and easy-to-use duplex filtering system. A complete system consists of a 10" or 20" 100% polypropylene (PP) bag and PP vessel (sold separately below right). Bags are available in seven pore sizes.
Specifications
Product Type
Bag Filter Housing
Housing dimensions
50.10 in L x 18.30 in Dia
Port size
2 in NPT(F)
Inlet/outlet NPT
2" NPT(F)
Brand
Hayward
Manufacturer number
FLV4P2S20T4VS
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
13-14 International Catalog
Related technical articles (Roll over for summary)
Compressed Air Purification Selection Guide
Industrial Filtration Selection Guide
Science in the Small Dairy Industry: Milk Sediment Testing
Show All...
Basics of Cartridge Filtration
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Related Searches for Hayward double-length bag filter housing, 2" NPT(F)
Hayward Single and Double-Length Bag Filters
Screw Top Bag Filter Housing
Diy Pvc Bag Filter Housing
Cole-Parmer Heavy-Duty Bag Filter Housings
Cole-Parmer Polypropylene Bag Filter Housings
Cole-Parmer Double Open End Filter Housings
Bag Filter Housing
Bag Filter Housing System
Polypropylene Bag Filter Housings
Process Bag Filter Housings
Related Products for Hayward double-length bag filter housing, 2" NPT(F)
Image
Product Detail
SKU
Hayward double-length bags; 1 µm, 30/pack
Category: Hayward Single- and Double-Length Bag Filters
Description: Hayward Bags
Brand:
Hayward
Model Number:
F31B36372E
YO-29990-70
More Details
Hayward double-length bags; 5 µm, 50/pack
Category: Hayward Single- and Double-Length Bag Filters
Description: Hayward Bags
Brand:
Hayward
Model Number:
F31B37455E
YO-29990-74
More Details
Hayward double-length bags; 25 µm, 50/pack
Category: Hayward Single- and Double-Length Bag Filters
Description: Hayward Bags
Brand:
Hayward
Model Number:
F3AB00002E
YO-29990-78
More Details
Hayward double-length bags; 100 µm, 50/pack
Category: Hayward Single- and Double-Length Bag Filters
Description: Hayward Bags
Brand:
Hayward
Model Number:
F3AA01128E
YO-29990-82
More Details
Process Bag Filter Housing, 2" NPT
Category: High-Flow Filter Bag Housings
Description: Process Bag Filter Housings
Brand:
Cole-Parmer
YO-01523-31
More Details
Hayward single-length bags; 1 µm, 50/pack
Category: Hayward Single- and Double-Length Bag Filters
Description: Hayward Bags
Brand:
Hayward
Model Number:
F3AB00013E
YO-29990-50
More Details
Hayward single-length bags; 25 µm, 50/pack
Category: Hayward Single- and Double-Length Bag Filters
Description: Hayward Bags
Brand:
Hayward
Model Number:
F3AB00020E
YO-29990-58
More Details
Hayward single-length bags; 100 µm, 50/pack
Category: Hayward Single- and Double-Length Bag Filters
Description: Hayward Bags
Brand:
Hayward
Model Number:
F31A37288E
YO-29990-62
More Details
Process Bag Filter Housing, 2" Flange
Category: High-Flow Filter Bag Housings
Description: Process Bag Filter Housings
Brand:
Cole-Parmer
YO-01523-33
More Details
Polypropylene bag filter housing, 1-1/2" NPT(F) (150338)
Category: Polypropylene Bag Filter Systems
Description: Polypropylene Bag Filter Housings
Brand:
Cole-Parmer
Model Number:
150338
YO-01510-04
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Hayward double-length bag filter housing, 2" NPT(F)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Hayward CPVC True Union Solenoid Valve, 3/4", 12 VDC
|
Next: Hayward double-length bags; 1 µm, 30/pack
31125329-p-content_2 31125329pcontent2