Threaded plug, nylon, 1/2" male NPT, 100/pack (YO-30401-40)

YO-30401-40
Loading price...

Usually ships in 14 days.
Qty
    Good chemical resistance. Rated to 200 psi at 77ºF (25ºC). Usable from -40 to 200ºF (-40 to 93ºC).
    Product Type Threaded Fitting
    Material Nylon
    NPT(M) (in.) 0.5
    Quantity 100
    Pipe size 1/2"





    Technical Resources
     
    Related Products for Threaded plug, nylon, 1/2" male NPT, 100/pack (YO-30401-40)
    Image
      Product Detail
    SKU

    Threaded plug, nylon, 1/2" male NPT, 10/pack (YO-30401-38)

    Category: Threaded plugs
    Description: Nylon Threaded Plugs, NPT(M)
    Brand: Cole-Parmer


    Threaded plug, nylon, 1/4" male NPT, 100/pack (YO-30401-44)

    Category: Threaded plugs
    Description: Nylon Threaded Plugs, NPT(M)
    Brand: Cole-Parmer


    Threaded plug, nylon, 1/8" male NPT, 10/pack (YO-31701-91)

    Category: Threaded plugs
    Description: Nylon Threaded Plugs, NPT(M)
    Brand: Cole-Parmer


    Threaded plug, nylon, 1/4" male NPT, 10/pack (YO-30401-42)

    Category: Threaded plugs
    Description: Nylon Threaded Plugs, NPT(M)
    Brand: Cole-Parmer


    Threaded plug, nylon, 3/4" male NPT, 100/pack (YO-30401-48)

    Category: Threaded plugs
    Description: Nylon Threaded Plugs, NPT(M)
    Brand: Cole-Parmer


    Threaded plug, nylon, 3/8" male NPT, 100/pack (YO-30401-52)

    Category: Threaded plugs
    Description: Nylon Threaded Plugs, NPT(M)
    Brand: Cole-Parmer


    Threaded plug, polyethylene, 1/2" male NPT, 100/pack (YO-30401-41)

    Category: Threaded plugs
    Description: HDPE Threaded Plugs, NPT(M)
    Brand: Cole-Parmer


    Threaded plug, polyethylene, 1/8" male NPT, 100/pack (YO-31701-92)

    Category: Threaded plugs
    Description: HDPE Threaded Plugs, NPT(M)
    Brand: Cole-Parmer


    Threaded plug, polyethylene, 1/4" male NPT, 100/pack (YO-30401-45)

    Category: Threaded plugs
    Description: HDPE Threaded Plugs, NPT(M)
    Brand: Cole-Parmer


    Threaded plug, nylon, 3/4" male NPT, 10/pack (YO-30401-46)

    Category: Threaded plugs
    Description: Nylon Threaded Plugs, NPT(M)
    Brand: Cole-Parmer


    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
    Questions and Answers
    Product Reviews

    Threaded plug, nylon, 1/2" male NPT, 100/pack (YO-30401-40)
    Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended
    Popularity |  Favorites |  Title |  Price |  Part Number |  Category
    Title |  Brand |  Category |  Price |  Popularity |  Best Sellers

    Previous: Threaded plug, nylon, 1/2" male NPT, 10/pack (YO-30401-38)  |  Next: Threaded plug, nylon, 1/4" male NPT, 10/pack (YO-30401-42)