Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
YO-30527-60
Parker
®
three-way ball valve, stainless steel, 1/4" NPT(F)
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Technical information
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-30527-60
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Parker Small Metal Ball Valves
Share
|
Parker three-way ball valve, stainless steel, 1/4" NPT(F)
YO-30527-60
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Specifications
Product Type
Ball Valves (manual)
Dimensions (" L)
2
Description
Three-way valves
Connections
0.25 in
Cv
0.7
Material
Stainless Steel
Max temperature (º F)
300
Max temperature (º C)
149
Max pressure (psi)
2500
Orifice (" Dia)
0.187
Brand
Parker
Model
4F-MB6XPFA-SSP
Manufacturer number
4F-MB6XPFA-SSP
Accessories
Qty
/
Price
Availability
Tri-Flow Food-Grade Anti-Seize Thread Compound, 9-oz jar
YO-08809-20
EACH
Call for availability.
Add to Cart
General-Purpose Anti-Seize Thread Compound, 8-oz jar
YO-08809-22
EACH
Call for availability.
Add to Cart
Technical Resources
Cv to Flow Conversion Technical Bulletin.pdf
Virtual Catalog (PDFs)
13-14 Catalog
13-14 International Catalog
Related technical articles (Roll over for summary)
Basics of Cartridge Filtration
Valve Selection Guide
Electron zap turns flimsy plastic into sturdy shrink wrap
Show All...
Bitter Brew
Show Less...
Related Searches for Parker three-way ball valve, stainless steel, 1/4" NPT(F)
Parker Three-Way Ball Valves
Hayward Electronically Actuated Three-Way Ball Valves
Electric Actuated Ball Valve Stainless Steel
Parker Small Metal Ball Valves
Three Way Check Valve
Three Way Luer Valve
Stainless Two Way Ball Valve
Cole-Parmer Three-Way Pinch Valves
Cole-Parmer Parker Metal Industrial Ball Valves
Cole-Parmer Chemically Inert Two-Way Ball Valves
Related Products for Parker three-way ball valve, stainless steel, 1/4" NPT(F)
Image
Product Detail
SKU
Parker three-way ball valve, stainless steel, 1/4" compression connect
Category: Parker Small Metal Ball Valves
Description: Parker Three-way ball valves
Brand:
Parker
Model Number:
4A-MB4XPFA-SSP
YO-30527-58
More Details
Parker three-way ball valve, stainless steel, 1/8" compression connect
Category: Parker Small Metal Ball Valves
Description: Parker Three-way ball valves
Brand:
Parker
Model Number:
2A-MB2XPFA-SSP
YO-30527-54
More Details
Parker three-way ball valve, stainless steel, 1/8" NPT(F)
Category: Parker Small Metal Ball Valves
Description: Parker Three-way ball valves
Brand:
Parker
Model Number:
2F-MB4XPFA-SSP
YO-30527-56
More Details
Parker three-way ball valve, stainless steel, 3/8" compression connect
Category: Parker Small Metal Ball Valves
Description: Parker Three-way ball valves
Brand:
Parker
Model Number:
6A-MB6XPFA-SSP
YO-30527-62
More Details
Parker three-way ball valve, brass, 1/8" compression connection
Category: Parker Small Metal Ball Valves
Description: Parker Three-way ball valves
Brand:
Parker
Model Number:
2A-MB2XPFA-BP
YO-30527-80
More Details
Parker three-way ball valve, brass, 1/8" NPT(F)
Category: Parker Small Metal Ball Valves
Description: Parker Three-way ball valves
Brand:
Parker
Model Number:
2F-MB4XPFA-BP
YO-30527-82
More Details
Parker three-way ball valve, brass, 1/4" compression connection
Category: Parker Small Metal Ball Valves
Description: Parker Three-way ball valves
Brand:
Parker
Model Number:
4A-MB4XPFA-BP
YO-30527-84
More Details
Parker three-way ball valve, brass, 1/4" NPT(F)
Category: Parker Small Metal Ball Valves
Description: Parker Three-way ball valves
Brand:
Parker
Model Number:
4F-MB6XPFA-BP
YO-30527-86
More Details
Parker three-way ball valve, brass, 3/8" compression connection
Category: Parker Small Metal Ball Valves
Description: Parker Three-way ball valves
Brand:
Parker
Model Number:
6A-MB6XPFA-BP
YO-30527-88
More Details
Parker two-way straight ball valve, stainless steel, 1/4" compression
Category: Parker Small Metal Ball Valves
Description: Parker Two-way straight ball valves
Brand:
Parker
Model Number:
4A-MB4LPFA-SSP
YO-30527-34
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Parker three-way ball valve, stainless steel, 1/4" NPT(F)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Parker three-way ball valve, stainless steel, 1/4" compression connect
|
Next: Parker three-way ball valve, stainless steel, 1/8" compression connect
31125329-p-content_2 31125329pcontent2