Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Sanitary Gasket Flanged Epdm 2 1 2 Clamp Size
Click to enlarge
More Product Details
Specifications
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-30564-68
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Gaskets for Sanitary Clamp Fittings
Share
|
Sanitary Gasket, Flanged, Epdm, 2-1/2" Clamp Size (40MPFE-250)
YO-30564-68
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Hands-free installation for difficult-to-reach locations!
With the same benefits as standard sanitary gaskets, these gaskets offer an outer lip to grip the sanitary coupling port -- once in place, no need to worry about maintaining the gasket's placement! An especially useful feature for hard-to-reach or vertical clamp joints, using the outer lip speeds assembly and reduces your line's down time.
Available in platinum-cured silicone, EPDM, Buna N and FKM (see material descriptions under standard sanitary fittings for more information). Sold in packs of 10.
Specifications
Product Type
Gaskets fittings
Material
EPDM
Manufacturer number
40MPFE-250
Model
40MPFE-250
Technical Resources
Related technical articles (Roll over for summary)
Sterilizing Fittings: The Critical Factors
Fittings Selection Guide
Show Less...
Related Searches for Sanitary Gasket, Flanged, Epdm, 2-1/2" Clamp Size (40MPFE-250)
Cole-Parmer Flanged Sanitary Gaskets
Cole-Parmer 1 2-Propanediol
Round Rubber Tubing 1 2
Wika 4 1/2" Industrial Gauges
Eutech Instruments 1/2-Din Ph/Orp Control
Pvdf 1 2 Id Tube
Tygothane 1 2 Id
Troy Mi 1 2 Steel Tubing
Stainless Steel Npt 1 2 Fittings
Cleanroom Labels Removable 2 X 2
Related Products for Sanitary Gasket, Flanged, Epdm, 2-1/2" Clamp Size (40MPFE-250)
Image
Product Detail
SKU
Sanitary Gasket, Flanged, Buna N, 1" Clamp Size (40MPFU-100)
Category: Gaskets for Sanitary Clamp Fittings
Description: Flanged sanitary gaskets
Brand:
Cole-Parmer
Model Number:
40MPFU-100
YO-30564-42
More Details
Sanitary Gasket, Flanged, Epdm, 2" Clamp Size (40MPFE-200)
Category: Gaskets for Sanitary Clamp Fittings
Description: Flanged sanitary gaskets
Brand:
Cole-Parmer
Model Number:
40MPFE-200
YO-30564-66
More Details
Sanitary Gasket, Flanged, Fkm, 2-1/2" Clamp Size (40MPFSFY-250)
Category: Gaskets for Sanitary Clamp Fittings
Description: Flanged sanitary gaskets
Brand:
Cole-Parmer
Model Number:
40MPFSFY-250
YO-30564-28
More Details
Sanitary Gasket, Flanged, Epdm, 1-1/2" Clamp Size (40MPFE-150)
Category: Gaskets for Sanitary Clamp Fittings
Description: Flanged sanitary gaskets
Brand:
Cole-Parmer
Model Number:
40MPFE-150
YO-30564-64
More Details
Sanitary Gasket, Flanged, Epdm, 1" Clamp Size (40MPFE-100)
Category: Gaskets for Sanitary Clamp Fittings
Description: Flanged sanitary gaskets
Brand:
Cole-Parmer
Model Number:
40MPFE-100
YO-30564-62
More Details
Sanitary Gasket, Flanged, Buna N, 1-1/2" Clamp Size (40MPFU-150)
Category: Gaskets for Sanitary Clamp Fittings
Description: Flanged sanitary gaskets
Brand:
Cole-Parmer
Model Number:
40MPFU-150
YO-30564-44
More Details
Sanitary Gasket, Flanged, Fkm, 2" Clamp Size (40MPFSFY-200)
Category: Gaskets for Sanitary Clamp Fittings
Description: Flanged sanitary gaskets
Brand:
Cole-Parmer
Model Number:
40MPFSFY-200
YO-30564-26
More Details
Sanitary Gasket, Buna N, 2 1/2", 10/pk (YO-30548-48)
Category: Gaskets for Sanitary Clamp Fittings
Description: Cole-Parmer Gaskets for Sanitary Lines
Brand:
Cole-Parmer
YO-30548-48
More Details
Sanitary Gasket, Flanged, Platinum-cured Silicone, 1-1/2" Clamp Size (40RXPX-F-150)
Category: Gaskets for Sanitary Clamp Fittings
Description: Flanged sanitary gaskets
Brand:
Cole-Parmer
Model Number:
40RXPX-F-150
YO-30564-04
More Details
Sanitary Gasket, Flanged, Platinum-cured Silicone, 2-1/2" Clamp Size (40RXPX-F-250)
Category: Gaskets for Sanitary Clamp Fittings
Description: Flanged sanitary gaskets
Brand:
Cole-Parmer
Model Number:
40RXPX-F-250
YO-30564-08
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Sanitary Gasket, Flanged, Epdm, 2-1/2" Clamp Size (40MPFE-250)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Sanitary Gasket, Flanged, Epdm, 1" Clamp Size (40MPFE-100)
|
Next: Sanitary Gasket, Flanged, Epdm, 2" Clamp Size (40MPFE-200)
31125329-p-content_2 31125329pcontent2