Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
BarbLockTM for x tube
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
YO-30604-06
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
BarbLock TM Assemblies
Share
|
½" BarbLockTM for ½" x ¾" tube
YO-30604-06
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Easy to install, three-piece locking mechanism ensures the tubing and hose barb won't disengage.
Safe to use with pressures up to 140 psi.
An excellent alternative to securing tubing and hose barbs with cable tiesno more crimps or tube distortion.
Easy to install, thre-piece locking mechanism ensures the tubing and hose barb won't disengage
Safe to use with pressures up to 140 psi or upper pressure limit of tubing
An excellent alternative to securing tubing and hose barbs with cable tiesno more crimps or tube distortion
BarbLock assemblies provide uniform 360° radial crimping for a leak-free, durable seal. The BarbLock assembly consists of three pieces: the sleeve, collet, and barbed fitting. To install the fitting, slide the sleeve and collet onto the tubing and then insert the barbed fitting into the tubing. Prelock the sleeve and collet by pressing them together until you hear a "click." Slide the sleeve and collet to the edge of the barbed fitting and lock with the BarbLock tool. Once assembled, the BarbLock assembly can only be unlocked with the universal removal tool. Assembly models 30603-00 through -06 can be used with standard barbed connections.BarbLock assemblies provide unifrom 360-degree radial crimping for a leak-free, durable seal. The BarbLock assembly consists of three pieces: the sleeve, collect, and barbed fittings. To install the fitting, slide the sleeve and collet onto the tubing and then inserts the barbed fitting into the tubing. Prelock the sleeve and collet by pressing them together until you hear a "click." Slide the sleeve and collet to the edge of the barbed fitting and lock with the BarbLock tool. Once assembled, the BarbLock assembly can only be unlocked with the removal tool.
Specifications
Product Type
BarbLock Assemblies
Brand
BarbLock
Manufacturer number
BL135500-250
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
13-14 International Catalog
Related technical articles (Roll over for summary)
Barblock Assemblies
Pure-Fit® Tru Valve System
Pure-Fit® TC Tube Clamp System
Show All...
BarbLock® Ultra-Secure Tubing Retainer
Show Less...
Related Searches for ½" BarbLockTM for ½" x ¾" tube
Universal 2 1 4 Tubing Clamps
Female Npt Adapter to 3 4
3 Ss Tubing Elbow
Fluoroware 2 Way Valve 1 4
Hose Clamps for 1 4 Tube
Jaco 1 4 Tubing Fittings
Bakersfield Ca 1 4 Tubing Fittings
Small Fittings for 1 4 Tubing
Copper 3 4
Polyethylene Tubing 3 4 in Od
Related Products for ½" BarbLockTM for ½" x ¾" tube
Image
Product Detail
SKU
Cole-Parmer Balance Draft Shield, Small 13"W x 13-1/2"H x 14-3/4"D (small)
Category: Draft Shields and Enclosures
Description: Cole-Parmer Balance Draft Shield
Brand:
Cole-Parmer
Model Number:
small
YO-01019-02
More Details
Storage bin; 16-1/2" x 7" 14-3/4", 6/pack (30-250 YELLOW)
Category: Plastic Storage Bins & Containers
Description: Polypropylene Storage Bins and Racks
Brand:
Cole-Parmer
Model Number:
30-250 YELLOW
YO-06811-50
More Details
Storage bin; 16-1/2" x 7" 14-3/4" (30-250 YELLOW)
Category: Plastic Storage Bins & Containers
Description: Polypropylene Storage Bins and Racks
Brand:
Cole-Parmer
Model Number:
30-250 YELLOW
YO-06811-56
More Details
Stainless steel utility tray, 13-1/2"L x 9-3/4"W
Description: Utility Trays
Brand:
Guaranteed In Stock Program
YO-07277-13
More Details
PVDF Sanitary Barbed Adapter, 1" Ladish Clamp Size x 3/4" Tubing ID (YO-31808-34)
Category: PVDF sanitary clamp fittings
Description: PVDF sanitary clamp to barbed adapters
Brand:
Cole-Parmer
YO-31808-34
More Details
Insul-Ice™ Sheet, 16-1/2"L x 14-3/4"W, 28 sheets/case
Category: Shippers
Description: Insul-Ice Sheet
Brand:
Insul-Ice
YO-06345-55
More Details
56C Frame AC Motor, 1725 rpm, 1/2 hp, totally enclosed fan-cooled (TEF (CL3504)
Category: Motors with 56C Frame
Description: 3/4 Hp and Smaller TEFC AC Motors, 56C Mount
Brand:
Masterflex
Model Number:
CL3504
YO-07129-20
More Details
Front View Distillation Apparatus, Groups 0, 1, 2, and 3, right side, (K45000)
Category: Petroleum Testing
Description: Front View Distillation Apparatus, Groups 0, 1, 2, and 3
Brand:
Koehler
Model Number:
K45000
YO-59873-00
More Details
Front View Distillation Apparatus, Groups 0, 1, 2, and 3, right side, (K45090)
Category: Petroleum Testing
Description: Front View Distillation Apparatus, Groups 0, 1, 2, and 3
Brand:
Koehler
Model Number:
K45090
YO-59873-05
More Details
Front View Distillation Apparatus, Groups 0, 1, 2, and 3, left side, 1
Category: Petroleum Testing
Description: Front View Distillation Apparatus, Groups 0, 1, 2, and 3
Brand:
Koehler
Model Number:
K45100
YO-59873-10
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
TM assembly tool" ProdReview="
(0)
">
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
½" BarbLockTM for ½" x ¾" tube
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: ½" BarbLockTM assembly tool
|
Next: ½" BarbLockTM for Colder couplings
31125329-p-content_2 31125329pcontent2