Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
YO-70100-10
Basic Variable-Speed DC Motor Controller for 1/4 to 2 hp motors
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
YO-70100-10
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
DC Motor Controllers
Share
|
Basic Variable-Speed DC Motor Controller for 1/4 to 2 hp motors
YO-70100-10
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Vary the speed of your DC motor with these controllers
Basic Controller
keeps 1/4- to 2-hp motors at constant speeds without additional resistors. The NEMA 1 (IP 10) housing guards against dust, light, and incidental splashing.
For thermal overload protection, use heat sink model 70100-50 with 90 VDC motors (3/4 hp or larger) and 180 VDC motors (1-1/2 hp or larger)—order separately.
What's Included:
Instruction operation manual
Specifications
Product Type
DC motor controllers
hp
1/4 to 1 hp @ 115 VAC
1/2 to 2 hp @ 230 VAC
Power
115/230 VAC
Dimensions
5"W x 8"H x 2-3/4"D
Accessories
Qty
/
Price
Availability
Heat sink for DC controller 70100-10
YO-70100-50
EACH
Call for availability.
Add to Cart
Technical Resources
Instruction Manual
Instruction Manual
Virtual Catalog (PDFs)
13-14 Catalog
13-14 International Catalog
Related technical articles (Roll over for summary)
Motor Enclosure Terminology
Show Less...
Related Searches for Basic Variable-Speed DC Motor Controller for 1/4 to 2 hp motors
Variable Speed Dc Motor Controller
Variable Speed Dc Transfer Pumps
Dc Variable Speed Drives
Speed Motor Control
Variable Speed Drives for Motors
Variable Speed Motors
Variable Speed Pump Motors
Variable Speed Pump Motor 6x6 10sel
Magnetek Variable Speed Pump Motors
Drives Variable Speed
Related Products for Basic Variable-Speed DC Motor Controller for 1/4 to 2 hp motors
Image
Product Detail
SKU
Advanced Variable-Speed DC Motor Drive, for 1/4 to 2 hp motors
Category: DC Motor Controllers
Description: Advanced Variable-Speed DC Motor Drive
Brand:
Cole-Parmer
Model Number:
PCM23401A
YO-70100-00
More Details
Variable-speed liquid piston pump, Class 1, Division 2 rated for hazar
Category: Piston Pumps
Description: Dual Solenoid Variable-speed Liquid and Gas Piston Pumps
Brand:
Cole-Parmer
Model Number:
PW2003E
YO-79306-10
More Details
Variable-speed gas piston pump, Class 1, Division 2 rated for hazardou
Category: Piston Pumps
Description: Dual Solenoid Variable-speed Liquid and Gas Piston Pumps
Brand:
Cole-Parmer
Model Number:
PW2071E
YO-79306-15
More Details
Variable-speed liquid piston pump, Class 1, Division 1 rated for hazar
Category: Piston Pumps
Description: Dual Solenoid Variable-speed Liquid and Gas Piston Pumps
Brand:
Cole-Parmer
Model Number:
PW2004E
YO-79306-20
More Details
Variable-speed gas piston pump, Class 1, Division 1 rated for hazardou
Category: Piston Pumps
Description: Dual Solenoid Variable-speed Liquid and Gas Piston Pumps
Brand:
Cole-Parmer
Model Number:
PW2072E
YO-79306-25
More Details
Masterflex L/S Precision Variable-Speed Drive with Remote Input; 1 to
Category: L/S Analog Variable-Speed Drives
Description: L/S Precision Variable-Speed Console Drives
Brand:
L/S
YO-07528-30
More Details
Waring variable-speed blender, 1-liter, glass 120v
Category: Waring One-Liter Blenders
Description: Waring Variable-Speed Blenders
Brand:
Waring
YO-04245-60
More Details
Waring variable-speed blender, 1-liter, SS, 120v
Category: Waring One-Liter Blenders
Description: Waring Variable-Speed Blenders
Brand:
Waring
YO-04245-62
More Details
Masterflex® L/S® variable-speed drive with 10-turn speed contr (77521-57)
Category: L/S
®
Variable-Speed Analog Drives
Description: L/S
Brand:
Masterflex
Model Number:
77521-57
YO-77521-57
More Details
1/4" Quick-Disconnect Brass Coupler with Shut-Off Valve, 1/4" NPT(F) (A184)
Category: Couplings
Description: 1/4" Quick-Disconnect Brass Couplings
Model Number:
A184
YO-79162-01
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Basic Variable-Speed DC Motor Controller for 1/4 to 2 hp motors
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Basic Statistics for Laboratories (0-471-28405-X)
|
Next: Basin Cover, HDPE for Basins 06019-10, -20
31125329-p-content_2 31125329pcontent2