Threaded plug, PFA, 1/8" male NPT (YO-31321-13)

YO-31321-13
Loading price...

Call for availability.
Qty
Product Type Compression Fitting
Material PFA
NPT(M) (in.) 0.125
Sterilization Ethylene Oxide (ETO) or Autoclave





Technical Resources
 
Related Products for Threaded plug, PFA, 1/8" male NPT (YO-31321-13)
Image
  Product Detail
SKU

Threaded plug, PFA, 1/4" male NPT (YO-31321-14)

Category: Threaded plugs
Description: Fluoropolymer Pipe Plugs NPT(M)
Brand: Parker Hannifin


Threaded plug, PFA, 3/8" male NPT (YO-31321-15)

Category: Threaded plugs
Description: Fluoropolymer Pipe Plugs NPT(M)
Brand: Parker Hannifin


Threaded plug, PFA, 1/2" male NPT (YO-31321-16)

Category: Threaded plugs
Description: Fluoropolymer Pipe Plugs NPT(M)
Brand: Parker Hannifin


Threaded plug, PFA, 1/8" male NPT (YO-06546-37)

Category: Threaded plugs
Description: Fluoropolymer Pipe Plugs NPT(M)
Brand: Cole-Parmer


Threaded plug, PFA, 3/4" male NPT (YO-31321-17)

Category: Threaded plugs
Description: Fluoropolymer Pipe Plugs NPT(M)
Brand: Parker Hannifin


Threaded plug, PFA, 1/4" male NPT (YO-06546-38)

Category: Threaded plugs
Description: Fluoropolymer Pipe Plugs NPT(M)
Brand: Cole-Parmer


Threaded plug, PFA, 3/8" male NPT (YO-06546-39)

Category: Threaded plugs
Description: Fluoropolymer Pipe Plugs NPT(M)
Brand: Cole-Parmer


Threaded plug, PFA, 1/2" male NPT (YO-06546-40)

Category: Threaded plugs
Description: Fluoropolymer Pipe Plugs NPT(M)
Brand: Cole-Parmer


PFA compression threaded adapter, 3/8" tubing OD x 1/8" male NPT (YO-31321-21)

Category: Male Threaded Adapters
Description: PFA Compression Threaded Adapters
Brand: Parker Hannifin


PFA compression threaded adapter, 4mm tubing OD x 1/8" male NPT (YO-31321-62)

Category: Male Threaded Adapters
Description: PFA Compression Threaded Adapters
Brand: Cole-Parmer


Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
Questions and Answers
Product Reviews

Threaded plug, PFA, 1/8" male NPT (YO-31321-13)
Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended
Popularity |  Favorites |  Title |  Price |  Part Number |  Category
Title |  Brand |  Category |  Price |  Popularity |  Best Sellers

Previous: Threaded plug, PFA, 1/8" male NPT (YO-06546-37)  |  Next: Threaded plug, PFA, 3/4" male NPT (YO-06546-41)