Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
YO-31501-41
Barbed fittings, NPT female pipe adapters, Clear PP,1/4" NPT x 1/8" ID, 1/16", 1-1/32", 5/8"; 25/pack
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
YO-31501-41
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Barbed to threaded adapters
Share
|
Barbed fittings, NPT female pipe adapters, Clear PP,1/4" NPT x 1/8" ID (YO-31501-41)
YO-31501-41
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Specifications
Product Type
Barbed fittings
Tubing ID (in.)
0.125
Dimensions orifice
1/16"
Sterilization
Autoclave or ETO
Dimensions B
5/8"
Material
Polypropylene
Dimensions A
1-1/32"
Quantity
25
Tubing ID
1/8"
NPT(F)
0.25
Manufacturer number
AP072518TF12P
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
13-14 International Catalog
Related technical articles (Roll over for summary)
Cole-Parmers Bioconnect Program
Sterilizing Fittings: The Critical Factors
Fittings Selection Guide
Show All...
Show Less...
Related Searches for Barbed fittings, NPT female pipe adapters, Clear PP,1/4" NPT x 1/8" ID (YO-31501-41)
Brass Fittings Tube to Pipe Adapters
Female Pipe Adapters
Male to Female Pipe Adapters
Fitting Tube Size 1 8 Id
Fittings Polypropylene Female
Npt Female to Female Adapter
In Female Barb Fitting
Female Npt Adapter to 3 4
Npt Female
Guaranteed I Male Pipe Adapters (Straight)
Related Products for Barbed fittings, NPT female pipe adapters, Clear PP,1/4" NPT x 1/8" ID (YO-31501-41)
Image
Product Detail
SKU
Barbed fittings, NPT female pipe adapters, Clear PP, 1/8" NPT x 1/8" I (YO-31501-40)
Category: Barbed to threaded adapters
Description: Straight barbed to female NPT threaded adapters
Brand:
Cole-Parmer
YO-31501-40
More Details
Barbed fittings, NPT male pipe adapter, Clear PP, 1/8" NPT x 1/16" ID, (YO-30506-20)
Category: Barbed to threaded adapters
Description: Straight barbed to male NPT threaded adapters
Brand:
Cole-Parmer
YO-30506-20
More Details
Barbed fittings, NPT male pipe adapter, Clear PP, 1/8" NPT x 3/32" ID, (YO-30506-21)
Category: Barbed to threaded adapters
Description: Straight barbed to male NPT threaded adapters
Brand:
Cole-Parmer
YO-30506-21
More Details
Barbed fittings, NPT male pipe adapter, Clear PP, 1/8" NPT x 1/8" ID, (YO-30506-22)
Category: Barbed to threaded adapters
Description: Straight barbed to male NPT threaded adapters
Brand:
Cole-Parmer
YO-30506-22
More Details
Barbed fittings, NPT male pipe adapter, Black PP, 1/4" OD x 3/16" NPT, (YO-06444-18)
Category: Barbed to threaded adapters
Description: Straight barbed to male NPT threaded adapters
Brand:
Parker Hannifin
YO-06444-18
More Details
Barbed fittings, NPT male pipe adapter, Natural PP, 1/4" NPT x 1/4" I (YO-30610-38)
Category: Straight barbed to male NPT threaded adapters
Description: Natural polypropylene straight barbed to male NPT threaded adapters
Brand:
Cole-Parmer
YO-30610-38
More Details
Barbed fittings, NPT male pipe adapter, Natural PP, 1/4" NPT x 3/8" ID (YO-30610-39)
Category: Straight barbed to male NPT threaded adapters
Description: Natural polypropylene straight barbed to male NPT threaded adapters
Brand:
Cole-Parmer
YO-30610-39
More Details
Barbed fittings, NPT male pipe adapter, Natural PP, 1/4" NPT x 1/2" (YO-30610-40)
Category: Straight barbed to male NPT threaded adapters
Description: Natural polypropylene straight barbed to male NPT threaded adapters
Brand:
Cole-Parmer
YO-30610-40
More Details
Barbed fittings, NPT male pipe adapter, Natural PP, 1/4" NPT x 3/8" I (YO-30640-39)
Category: Straight barbed to male NPT threaded adapters
Description: Natural polypropylene straight barbed to male NPT threaded adapters
Brand:
Cole-Parmer
YO-30640-39
More Details
Barbed fittings, NPT male pipe adapter, Natural PP, 1/4" NPT x 1/2" I (YO-30640-40)
Category: Straight barbed to male NPT threaded adapters
Description: Natural polypropylene straight barbed to male NPT threaded adapters
Brand:
Cole-Parmer
YO-30640-40
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Barbed fittings, NPT female pipe adapters, Clear PP,1/4" NPT x 1/8" ID (YO-31501-41)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Barbed fittings, NPT female pipe adapters, Clear PP, 1/8" NPT x 1/8" I (YO-31501-40)
|
Next: Barbed fittings, NPT female pipe adapters, Nylon, 1//8" NPT x 1/8" ID, (YO-31500-40)
31125329-p-content_2 31125329pcontent2