Home  >  Threaded nipples >
  

Pipe fittings, Nipple, PP, 10-32 UNF, 3/8", 1/4", 1/16"; 25/pack (YO-31503-10)

YO-31503-10
Loading price...

Call for availability.
Qty
Product Type Threaded Fitting
Material Polypropylene
Tubing ID (in.) 10-32 UNF(M)
Sterilization Autoclave or ETO
Dimensions A 3/8"
Dimensions C 1/16"
Dimensions B 1/4"
Quantity 25
Pipe size 10-32





Technical Resources
 
Related Products for Pipe fittings, Nipple, PP, 10-32 UNF, 3/8", 1/4", 1/16"; 25/pack (YO-31503-10)
Image
  Product Detail
SKU

Pipe fittings, Nipple, Nylon, 10-32 UNF, 3/8", 1/4", 1/16"; 25/pack (YO-31502-10)

Category: Threaded nipples
Description: UNF(M) Nipples
Brand: Cole-Parmer


Pipe fittings, Nipples UNF, Nylon, 10-32 UNF, 3/8", 1/4", 1/16"; 250/pack

Category: Threaded nipples
Description: UNF(M) Nipples
Brand: Cole-Parmer


Barbed fittings, UNF male pipe adapter, Clear PP, 10-32 UNF x 5/32" ID, 3/32", 13/16", 1/4"

Category: Barbed to threaded adapters
Description: Straight barbed to male UNF threaded adapters
Brand: Cole-Parmer


Barbed fittings, UNF male pipe adapter, Clear PP, 10-32 UNF x 5/32" ID, 3/32", 13/16", 1/4"; 25/pack

Category: Barbed to threaded adapters
Description: Straight barbed to male UNF threaded adapters
Brand: Cole-Parmer


Barbed fittings, UNF male pipe adapter, Clear PP, 10-32 UNF x 1/16" ID, 1/32", 5/8", 1/4"

Category: Barbed to threaded adapters
Description: Straight barbed to male UNF threaded adapters
Brand: Cole-Parmer


Barbed fittings, NPT male pipe adapter elbows, Natural PP, 3/8" NPT x 1/4" ID, 1/8", 1-3/8", 1-7/16"; 100/pack (L6-4NP(100/PK))

Category: Barbed to male NPT threaded elbow adapters
Description: Natural polypropylene barbed to male NPT threaded elbow adapters
Brand: Cole-Parmer


Barbed fittings, NPT male pipe adapter elbows, Natural PP, 3/8" NPT x 1/4" ID, 1/8", 1-3/8", 1-7/16"; 100/pack

Category: Barbed to threaded adapters
Description: Barbed to male NPT threaded elbow adapters
Brand: Cole-Parmer


Male Pipe Adapter, Polyethylene, 3/8" x 5/16", 100 PACK (YO-30625-58)

Category: Barbed to threaded adapters
Description: Straight barbed to male NPT threaded adapters
Brand: Cole-Parmer


Male Pipe Adapter, Kynar, 3/8" x 5/16", 50 PACK (YO-30625-59)

Category: Barbed to threaded adapters
Description: Straight barbed to male NPT threaded adapters
Brand: Cole-Parmer


Male Pipe Adapter, Nylon, 3/8" x 5/16", 100 PACK (YO-30625-60)

Category: Barbed to threaded adapters
Description: Straight barbed to male NPT threaded adapters
Brand: Cole-Parmer


Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
Questions and Answers
Product Reviews