Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Threaded adapter stainless steel 1 8 female NPT to 1 8 male BSPP
Click to enlarge
More Product Details
Specifications
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-31830-50
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
British Standard Pipe Tapered Threaded (BSPT) Adapters
Share
|
Threaded adapter, stainless steel, 1/8" female NPT to 1/8" male BSPP (YO-31830-50)
YO-31830-50
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Convert NPT-threaded systems to BSP threads—or visa versa—with these stainless steel fittings.
Use BSP taper (BSPT) fittings for connections where pressure-tight joints are made on the thread, BSP parallel (BSPP) fittings where pressure-tight joints are not made on the thread (as in connections with peripheral seals). Stainless steel fittings are chemically resistant.
Specifications
Product Type
Threaded fittings
NPT(M) (in.)
0.125
Material
Stainless Steel
NPT(F)
0.125
Brand
Parker Hannifin
Manufacturer number
2-2R-RA-SS
Technical Resources
Related technical articles (Roll over for summary)
Pipe Thread Types and Designations
Standard Pipe Thread Fittings
Sterilizing Fittings: The Critical Factors
Show All...
Fittings Selection Guide
Show Less...
Related Searches for Threaded adapter, stainless steel, 1/8" female NPT to 1/8" male BSPP (YO-31830-50)
Tubing Adapters Stainless Steel
8 Npt Stainless Steel Pipe
Parker Stainless Steel Straight Thread Fittings
Stainless Steel Tubing Od Sizes
British Pipe Thread Adapter
Stainless Steel Tubing Compression Fitting
Stainless Steel Flare Fitting
Stainless Steel Tubing Fittings Manufacturer
Metric Size Stainless Steel Compression Fittings
Stainless Steel 1 2 Npt Adapters
Related Products for Threaded adapter, stainless steel, 1/8" female NPT to 1/8" male BSPP (YO-31830-50)
Image
Product Detail
SKU
Threaded adapter, stainless steel, 1/8" female NPT to 1/8" male BSPT (YO-31830-40)
Category: BSP Adapters
Description: British Standard Pipe Tapered Threaded (BSPT) Adapters
Brand:
Parker Hannifin
YO-31830-40
More Details
Threaded adapter, stainless steel, 1/8" female BSPT to 1/8" male NPT (YO-31830-60)
Category: BSP Adapters
Description: British Standard Pipe Tapered Threaded (BSPT) Adapters
Brand:
Parker Hannifin
YO-31830-60
More Details
Threaded adapter, stainless steel, 1/8" female BSPP to 1/8" male NPT (YO-31830-70)
Category: British Standard Pipe Tapered Threaded (BSPT) Adapters
Description: British Standard Pipe Threaded (BSPP) Adapters
Brand:
Parker Hannifin
YO-31830-70
More Details
Compression Elbow Threaded Adapter, Stainless Steel, 1/8" x 1/8", Each (YO-31406-75)
Category: Male Threaded Adapter Elbows
Description: Stainless Steel Compression to NPT(M) Adapter Elbows
Brand:
Cole-Parmer
YO-31406-75
More Details
Compression Elbow Threaded Adapter, Stainless Steel, 1/8" x 1/4", Each (YO-31406-76)
Category: Male Threaded Adapter Elbows
Description: Stainless Steel Compression to NPT(M) Adapter Elbows
Brand:
Cole-Parmer
YO-31406-76
More Details
Threaded adapter, stainless steel, 3/8" female NPT to 3/8" male BSPP (YO-31830-52)
Category: British Standard Pipe Tapered Threaded (BSPT) Adapters
Description: British Standard Pipe Threaded (BSPP) Adapters
Brand:
Parker Hannifin
YO-31830-52
More Details
Threaded adapter, stainless steel, 1/2" female NPT to 1/2" male BSPP (YO-31830-53)
Category: British Standard Pipe Tapered Threaded (BSPT) Adapters
Description: British Standard Pipe Threaded (BSPP) Adapters
Brand:
Parker Hannifin
YO-31830-53
More Details
Compression pipe adapter, Stainless Steel, 1/4" x 1/8", Each (YO-31406-64)
Category: Male Threaded Adapters
Description: Stainless Steel Compression to NPT(M) Adapters
Brand:
Cole-Parmer
YO-31406-64
More Details
Compression pipe adapter, Stainless Steel, 3/8" x 1/4", Each (YO-31406-66)
Category: Male Threaded Adapters
Description: Stainless Steel Compression to NPT(M) Adapters
Brand:
Cole-Parmer
YO-31406-66
More Details
Compression pipe adapter, Stainless Steel, 3/8" x 1/2", Each (YO-31406-67)
Category: Male Threaded Adapters
Description: Stainless Steel Compression to NPT(M) Adapters
Brand:
Cole-Parmer
YO-31406-67
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Threaded adapter, stainless steel, 1/8" female NPT to 1/8" male BSPP (YO-31830-50)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Threaded adapter, stainless steel, 1/8" female BSPT to 1/8" male NPT (YO-31830-60)
|
Next: Threaded adapter, stainless steel, 1/8" female NPT to 1/8" male BSPT (YO-31830-40)
31125329-p-content_2 31125329pcontent2