Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Adapter 9 16 18 UNF x 1 4 NPT F
Click to enlarge
More Product Details
Specifications
Technical Resources
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-32775-51
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
General Accessories
Share
|
Adapter, 9/16"-18 UNF x 1/4" NPT(F) (YO-32775-51)
YO-32775-51
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Specifications
Product Type
Accessory
Technical Resources
Related Searches for Adapter, 9/16"-18 UNF x 1/4" NPT(F) (YO-32775-51)
Unf Thread 1 4 Adapters
Connection 1/4" Npt(F)
Plastic Ball Valve 1 4 Nptf
18 Unf Adapters
Unf 1 4 28
Npt 1 4 Pfa G Fittings
Quick Coupling 1 4 with Cord
To 1 4 Threaded Coupling
Npt 1 4 Straight Barb Fitting
Sanitary Fitting to 1 4 Pipe
Related Products for Adapter, 9/16"-18 UNF x 1/4" NPT(F) (YO-32775-51)
Image
Product Detail
SKU
Adapter, 9/16"-18 UNF x 1/2" hose barb (YO-32775-52)
Category: General Accessories
Description: Replacement Accessories
Brand:
Cole-Parmer
YO-32775-52
More Details
Adapter, 9/16"-18 UNF x 1/8" NPT(F) (YO-32775-50)
Category: General Accessories
Description: Replacement Accessories
Brand:
Cole-Parmer
YO-32775-50
More Details
Male Pipe Adapter, Polyethylene, 10-32 UNF x 1/8", 10 PACK (YO-30621-05)
Category: Barbed to threaded adapters
Description: Straight barbed to male UNF threaded adapters
Brand:
Cole-Parmer
YO-30621-05
More Details
Male Pipe Adapter, Nylon, 10-32 UNF x 1/8", 10 PACK (YO-30621-06)
Category: Barbed to threaded adapters
Description: Straight barbed to male UNF threaded adapters
Brand:
Cole-Parmer
YO-30621-06
More Details
Male Pipe Adapter, Polypropylene, 10-32 UNF x 1/8", 25 PACK (YO-30621-07)
Category: Straight barbed to male UNF threaded adapters
Description: Natural polypropylene straight barbed to male UNF threaded adapters
Brand:
Cole-Parmer
YO-30621-07
More Details
Male Pipe Adapter, Polyethylene, 10-32 UNF x 1/8", 100 PACK (YO-30625-05)
Category: Barbed to threaded adapters
Description: Straight barbed to male UNF threaded adapters
Brand:
Cole-Parmer
YO-30625-05
More Details
Male Pipe Adapter, Nylon, 10-32 UNF x 1/8", 100 PACK (YO-30625-06)
Category: Barbed to threaded adapters
Description: Straight barbed to male UNF threaded adapters
Brand:
Cole-Parmer
YO-30625-06
More Details
Male Pipe Adapter, Polypropylene, 10-32 UNF x 1/8", 250 PACK (YO-30625-07)
Category: Straight barbed to male UNF threaded adapters
Description: Natural polypropylene straight barbed to male UNF threaded adapters
Brand:
Cole-Parmer
YO-30625-07
More Details
PFA compression threaded adapter, 5/16" tubing OD x 1/4" male NPT (YO-31321-30)
Category: Male Threaded Adapters
Description: PFA Compression Threaded Adapters
Brand:
Cole-Parmer
YO-31321-30
More Details
Male Pipe Adapter, Polypropylene, 10-32 UNF x 1/8", 25 PACK (YO-40621-07)
Category: Barbed to threaded adapters
Description: Straight barbed to male UNF threaded adapters
Brand:
Cole-Parmer
YO-40621-07
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Adapter, 9/16"-18 UNF x 1/4" NPT(F) (YO-32775-51)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Adapter, 9/16"-18 UNF x 1/2" hose barb (YO-32775-52)
|
Next: Adapter, 9/16"-18 UNF x 1/8" NPT(F) (YO-32775-50)
31125329-p-content_2 31125329pcontent2