Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Horizontal Tube Furnace 36 1 4 Tube Length 1800C 208 240 VAC
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-33334-45
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Tube Furnaces
Share
|
Horizontal Tube Furnace, 36-1/4" Tube Length, 1800°C, 208 / 240 V (RHTH 120/600/18/C42)
YO-33334-45
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
For horizontal or vertical operatation up to 1800°C in gas atmosphere or vacuum applications
Insulation with vacuum-formed ceramic fiber plates provide fast heating time and greater energy efficiency
MoSi
2
heating elements, mounted vertically for easy replacement
Ceramic working tube with fiber plugs for applications in air
Furnaces with molybdenum disilicide (MoSi
2
) Heating Elements withstand oxidation at very high temperatures. High-quality insulation materials made of vacuum-formed fibre plates enable energy-saving operation and fast heating due to low heat storage and heat conductivity. These furnaces allow operation under gas atmosphere or vacuum. Tube furnaces operate up to 1800° C with either horizontal or vertical designs. Easy-to-use temperature controller features clear, blue-white LCD that shows status messages, power usage, operating time counter. Controller offers 9 programs with 18 segments per program.
What's Included:
an aluminum oxide process tube, type B thermocouple, and remote temperature controller with 9-ft (2.7-m) cable and 6-ft (1.8-m) power cord with standard NEMA plug.
Specifications
Product Type
Tube Furnace
Temperature range (º C) (high)
1800
Temp control accuracy (º C)
±1
Temp uniformity (º C)
±10°C
Chamber size (" L)
23.6
Chamber size (L (mm))
600
Power (VAC)
208/240
Overall dimensions (" W)
36.25
Overall dimensions (" H)
21-5/8
Overall dimensions (" D)
25.25
Power (Hz)
50/60
Power (watts)
2000
Shipping weight (lbs)
243
Shipping weight (kg)
110
Brand
Nabertherm
Model
RHTH 120/600/18/C42
Manufacturer number
RHTH 120/600/18/C42
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
13-14 International Catalog
Related technical articles (Roll over for summary)
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Related Searches for Horizontal Tube Furnace, 36-1/4" Tube Length, 1800°C, 208 / 240 V (RHTH 120/600/18/C42)
Tube Furnace 36
Universal 2 1 4 Tubing Clamps
Jaco 1 4 Tubing Fittings
Bakersfield Ca 1 4 Tubing Fittings
Hose Clamps for 1 4 Tube
Small Fittings for 1 4 Tubing
High Temperature Tube Furnace
Horizontal Tube Furnace
Low Temperature Tube Furnace
Nabertherm Compact Tube Furnaces with Silicon
Related Products for Horizontal Tube Furnace, 36-1/4" Tube Length, 1800°C, 208 / 240 V (RHTH 120/600/18/C42)
Image
Product Detail
SKU
Horizontal Tube Furnace, 18-1/2" Tube Length, 1800°C, 208 / 240 V (RHTH 120/150/18/C42)
Category: Tube Furnaces
Description: Nabertherm Horizontal and Vertical High Temperature Tube Furnaces
Brand:
Nabertherm
Model Number:
RHTH 120/150/18/C42
YO-33334-39
More Details
Horizontal Tube Furnace, 24-3/8" Tube Length, 1800°C, 208 / 240 V (RHTH 120/300/18/C42)
Category: Tube Furnaces
Description: Nabertherm Horizontal and Vertical High Temperature Tube Furnaces
Brand:
Nabertherm
Model Number:
RHTH 120/300/18/C42
YO-33334-42
More Details
Vertical Tube Furnace, 34-5/8" Tube Length, 1800°C, 208 / 240 VAC (RHTV 120/600/18/C42)
Category: Tube Furnaces
Description: Nabertherm Horizontal and Vertical High Temperature Tube Furnaces
Brand:
Nabertherm
Model Number:
RHTV 120/600/18/C42
YO-33334-54
More Details
Horizontal Tube Furnace, 36-1/4" Tube Length, 1600°C, 208 / 240 V (RHTH 120/600/16/C42)
Category: Tube Furnaces
Description: Nabertherm Horizontal and Vertical High Temperature Tube Furnaces
Brand:
Nabertherm
Model Number:
RHTH 120/600/16/C42
YO-33334-43
More Details
Horizontal Tube Furnace, 36-1/4" Tube Length, 1700°C, 208 / 240 V (RHTH 120/600/17/C42)
Category: Tube Furnaces
Description: Nabertherm Horizontal and Vertical High Temperature Tube Furnaces
Brand:
Nabertherm
Model Number:
RHTH 120/600/17/C42
YO-33334-44
More Details
Vertical Tube Furnace, 24-3/4" Tube Length, 1800°C, 208 / 240 VAC (RHTV 120/300/18/C42)
Category: Tube Furnaces
Description: Nabertherm Horizontal and Vertical High Temperature Tube Furnaces
Brand:
Nabertherm
Model Number:
RHTV 120/300/18/C42
YO-33334-51
More Details
Vertical Tube Furnace, 18-7/8" Tube Length, 1800°C, 208 / 240 VAC (RHTV 120/150/18/C42)
Category: Tube Furnaces
Description: Nabertherm Horizontal and Vertical High Temperature Tube Furnaces
Brand:
Nabertherm
Model Number:
RHTV 120/150/18/C42
YO-33334-48
More Details
Horizontal Tube Furnace, 18-1/2" Tube Length, 1600°C, 208 / 240 V (RHTH 120/150/16/C42)
Category: Tube Furnaces
Description: Nabertherm Horizontal and Vertical High Temperature Tube Furnaces
Brand:
Nabertherm
Model Number:
RHTH 120/150/16/C42
YO-33334-37
More Details
Horizontal Tube Furnace, 18-1/2" Tube Length, 1700°C, 208 / 240 V (RHTH 120/150/17/C42)
Category: Tube Furnaces
Description: Nabertherm Horizontal and Vertical High Temperature Tube Furnaces
Brand:
Nabertherm
Model Number:
RHTH 120/150/17/C42
YO-33334-38
More Details
Horizontal Tube Furnace, 24-3/8" Tube Length, 1600°C, 208 / 240 V (RHTH 120/300/16/C42)
Category: Tube Furnaces
Description: Nabertherm Horizontal and Vertical High Temperature Tube Furnaces
Brand:
Nabertherm
Model Number:
RHTH 120/300/16/C42
YO-33334-40
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Horizontal Tube Furnace, 36-1/4" Tube Length, 1800°C, 208 / 240 V (RHTH 120/600/18/C42)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Horizontal Tube Furnace, 36-1/4" Tube Length, 1700°C, 208 / 240 V (RHTH 120/600/17/C42)
|
Next: Horizontal Tube Furnace, 42-1/8" Tube Length, 1500°C, 208 / 240 V (RHTC 80-710/15/P330)
31125329-p-content_2 31125329pcontent2