Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, Fittings, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Steel Collecting Pan For Furnace Model Lt 9 L9 11 skm
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-33334-88
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Nabertherm Muffle Furnaces
Share
|
Steel Collecting Pan For Furnace Model Lt 9, L9/11/skm (691400147)
YO-33334-88
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Specifications
Product Type
Accessory
CE Compliance
Yes
Manufacturer number
691400147
Brand
Nabertherm
Model
691400147
Technical Resources
Virtual Catalog (PDFs)
11-12 Catalog
11-12 International Catalog
13-14 Catalog
13-14 International Catalog
Related technical articles (Roll over for summary)
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Related Searches for Steel Collecting Pan For Furnace Model Lt 9, L9/11/skm (691400147)
Stainless Steel Rectangular Pans
Stainless Steel Tube Pan
9 X 11 Metric Tubing
Ceramic Furnaces
Nabertherm Muffle Furnaces Accessories
Furnace Accessories
9 Liter Muffle Furnace
Furnace Insulation
Muffle Furnace Vertical Lift Door
Nabertherm Muffle Furnaces
Related Products for Steel Collecting Pan For Furnace Model Lt 9, L9/11/skm (691400147)
Image
Product Detail
SKU
Ceramic Ribbed Plate For Furnace Model Lt 9, L9/11/skm (691600509)
Category: Nabertherm Muffle Furnaces
Description: Nabertherm Muffle Furnaces Accessories
Brand:
Nabertherm
Model Number:
691600509
YO-33334-79
More Details
Steel Collecting Pan For Furnace Model Lt 3/11 (691400145)
Category: Nabertherm Muffle Furnaces
Description: Nabertherm Muffle Furnaces Accessories
Brand:
Nabertherm
Model Number:
691400145
YO-33334-86
More Details
Steel Collecting Pan For Furnace Model Lt 5/11 (691400146)
Category: Nabertherm Muffle Furnaces
Description: Nabertherm Muffle Furnaces Accessories
Brand:
Nabertherm
Model Number:
691400146
YO-33334-87
More Details
Steel Collecting Pan For Furnace Model Lt 15/11 (691400149)
Category: Nabertherm Muffle Furnaces
Description: Nabertherm Muffle Furnaces Accessories
Brand:
Nabertherm
Model Number:
691400149
YO-33334-89
More Details
Steel Collecting Pan For Furnace Model Lt 24/11 (691400626)
Category: Nabertherm Muffle Furnaces
Description: Nabertherm Muffle Furnaces Accessories
Brand:
Nabertherm
Model Number:
691400626
YO-33334-90
More Details
Steel Collecting Pan For Furnace Model Lt 40/11 (691400627)
Category: Nabertherm Muffle Furnaces
Description: Nabertherm Muffle Furnaces Accessories
Brand:
Nabertherm
Model Number:
691400627
YO-33334-91
More Details
Ceramic Collecting Plate For Furnace Model Lt 3 (691600510)
Category: Nabertherm Muffle Furnaces
Description: Nabertherm Muffle Furnaces Accessories
Brand:
Nabertherm
Model Number:
691600510
YO-33334-83
More Details
Ceramic Ribbed Plate For Furnace Model Lt 3 (691600507)
Category: Nabertherm Muffle Furnaces
Description: Nabertherm Muffle Furnaces Accessories
Brand:
Nabertherm
Model Number:
691600507
YO-33334-77
More Details
Ceramic Ribbed Plate For Furnace Model Lt 24 (691600874)
Category: Nabertherm Muffle Furnaces
Description: Nabertherm Muffle Furnaces Accessories
Brand:
Nabertherm
Model Number:
691600874
YO-33334-81
More Details
MUFFLE FURNACE, 540 CU.IN., 100-1100°C, 208 / 240 VAC (LT 9/11/P330)
Description: Nabertherm Muffle Furnaces
Brand:
Nabertherm
Model Number:
LT 9/11/P330
YO-33334-10
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Steel Collecting Pan For Furnace Model Lt 9, L9/11/skm (691400147)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Steel Collecting Pan For Furnace Model Lt 5/11 (691400146)
|
Next: Steel forceps with PTFE coating (37937)
31125329-p-content_2 31125329pcontent2