Steel Collecting Pan For Furnace Model Lt 9, L9/11/skm

YO-33334-88
Loading price...

Call for availability.
Qty
Product Type Accessory
CE Compliance Yes
Manufacturer number 691400147
Brand Nabertherm
Model 691400147





Technical Resources
 
Related Products for Steel Collecting Pan For Furnace Model Lt 9, L9/11/skm
Image
  Product Detail
SKU

Ceramic Ribbed Plate For Furnace Model Lt 9, L9/11/skm (691600509)

Category: Furnace Accessories
Description: Nabertherm Muffle Furnaces Accessories
Brand: Nabertherm


Steel Collecting Pan For Furnace Model Lt 3/11 (691400145)

Category: Furnace Accessories
Description: Nabertherm Muffle Furnaces Accessories
Brand: Nabertherm


Steel Collecting Pan For Furnace Model Lt 5/11 (691400146)

Category: Furnace Accessories
Description: Nabertherm Muffle Furnaces Accessories
Brand: Nabertherm


Steel Collecting Pan For Furnace Model Lt 15/11 (691400149)

Category: Furnace Accessories
Description: Nabertherm Muffle Furnaces Accessories
Brand: Nabertherm


Steel Collecting Pan For Furnace Model Lt 24/11 (691400626)

Category: Furnace Accessories
Description: Nabertherm Muffle Furnaces Accessories
Brand: Nabertherm


Steel Collecting Pan For Furnace Model Lt 40/11 (691400627)

Category: Furnace Accessories
Description: Nabertherm Muffle Furnaces Accessories
Brand: Nabertherm


Ceramic Collecting Plate For Furnace Model Lt 3 (691600510)

Category: Furnace Accessories
Description: Nabertherm Muffle Furnaces Accessories
Brand: Nabertherm


Ceramic Ribbed Plate For Furnace Model Lt 3 (691600507)

Category: Furnace Accessories
Description: Nabertherm Muffle Furnaces Accessories
Brand: Nabertherm


Ceramic Ribbed Plate For Furnace Model Lt 24 (691600874)

Category: Furnace Accessories
Description: Nabertherm Muffle Furnaces Accessories
Brand: Nabertherm


MUFFLE FURNACE, 540 CU.IN., 100-1100°C, 208 / 240 VAC (LT 9/11/P330)

Category: Muffle Furnaces
Description: Nabertherm Muffle Furnaces
Brand: Nabertherm


Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
Questions and Answers
Product Reviews

Steel Collecting Pan For Furnace Model Lt 9, L9/11/skm
Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended
Popularity |  Favorites |  Title |  Price |  Part Number |  Category
Title |  Brand |  Category |  Price |  Popularity |  Best Sellers

Previous: Steel Collecting Pan For Furnace Model Lt 5/11 (691400146)  |  Next: Steel Plated Beaker Tongs, 9"